Advance your cell and gene therapies with real-time PCR technologies
Real-time PCR (qPCR) and digital PCR (dPCR) play critical roles from the development to manufacture of cell therapies, including cell isolation, cell modification/gene modification, cell expansion, cell banking, cell identification, and contamination testing.
For viral vectors and plasmid DNA development for gene therapy, dPCR offers significant advantages for quantification of target DNA molecules in a range of analytical assays necessary for viral vector production and characterization. Unlike qPCR, dPCR can provide an absolute count of nucleic acids, enabling the precise quantification of AAV vectors, bacterial contaminants, and residual host cell DNA. No reference standard is required, improving precision and removing a source of variation. 
QualTrak cell and gene therapy workflow
The QualTrak cell and gene therapy workflow includes qPCR use cases across all steps of the development pipeline from discovery to GMP manufacturing.
Absolute Q Viral Titer dPCR assays
Absolute Q Viral Titer Digital PCR (dPCR) assays offer a precise, fast, and simple solution for accurate, absolute quantification of viral vectors in cell and gene therapy. 
Compliance documentation
QualTrak cGMP-compliant assays and reagents include quick and easy  access to documentation.
QualTrak workflow
qPCR use-cases in mRNA and protein-based cell and gene therapy pipelines
Discovery
Vector construct identification 
Viral titer measurement 
Vector construct analysis 
Residual plasmid DNA detection 
Quantification and quality monitoring of stored DNA 
Quantification of NGS libraries 
Cell counting 
Process development
Detection of microbial contamination 
Residual host cell contamination 
Confirmation of cell identity, modification,  and characteristics 
Viral titer measurement 
Pre-clinical and clinical trials
Confirmation of cell identity and functional characteristics 
Confirmation of transgene 
Efficacy studies 
cGMP manufacturing
Detection of microbial contamination 
Residual host cell contamination 
Viral titer measurement
Confirmation of product identity and characteristics 
1x1 image pixel for data collection