Advances in materials science
Innovative materials play essential roles in safety, clean energy, transportation, human health, and industrial productivity. To fuel continued innovation, researchers want to deepen their understanding of the physical and chemical properties of materials (morphological, structural, magnetic, thermal, and mechanical) from the macro- to nanoscale. Whether discovering new materials, solving analytical problems, improving processes, or assuring product quality, electron microscopy is capable of providing insight at all scales and modalities. The discoveries resulting from materials science research help enhance researchers’ ability to successfully correlate structural properties with functional performance. In turn, this insight helps commercial enterprises innovate products and processes to gain important time-to-market and cost advantages.
Materials characterization
Analytical solutions, including electron microscopy and spectroscopy, from Thermo Fisher Scientific can help you address your most pressing challenges, including;
  • Developing new functional materials that meet the demands of today’s unique social and economic challenges
  • Supporting the discovery of new materials with reproducible data from complementary techniques
  • Solving materials and method development challenges to improve processes and investigate product defects
  • Publishing groundbreaking discoveries, writing winning grant proposals, or patenting novel materials
  • Assuring defects are rejected before they reach customers
  • Taking your ideas to market quickly and keeping your company competitive
Video Player is loading.
Current Time 0:00
Duration 0:39
Loaded: 0%
Stream Type LIVE
Remaining Time 0:39
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    Defect analysis of a lithium ion battery cathode. Serial sectioning and imaging with Plasma FIB DualBeam followed by digital 3D reconstruction using Avizo software provides a highly detailed model of the sample.
     Scanning Electron Microscopy for Materials Science
      Scanning electron microscopy (SEM) is a powerful imaging technique that uses focused beams of electrons to produce high-resolution, three-dimensional images of sample surfaces at the nanoscale. This method is essential for researchers seeking detailed topographical, compositional, and morphological insights into a wide range of materials.
      Learn more
    Industrial manufacturing product selector
    The product selector is here to assist you in choosing the most suitable Scanning Electron Microscope (SEM) system and software for your research. Find out in a few minutes which SEM best suits your research application:
    Materials Science learning center
    Access a targeted collection of application notes, case studies, videos, webinars, and white papers covering a range of applications.
    Visit Materials science learning center
     TEM Sample Preparation
      Proper sample preparation is a critical step in the scientific experiment process—a step that can determine the ultimate quality and accuracy of the results. FIB-SEM instruments can be used to   facilitate fully automated in situ TEM sample preparation, enabling new chemistries and technologies while advancing research in batteries, polymers, metals, and more.
      Learn more
    Applications
    Process control using electron microscopy
    Process control using electron microscopy
    Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
    Quality control and failure analysis using electron microscopy
    Quality control and failure analysis
    Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
    Fundamental Materials Research_R&D_Thumb_274x180_144DPI
    Fundamental Materials Research
    Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
    Aluminum mineral grain found with SEM during parts cleanliness testing
    Technical Cleanliness
    More than ever, modern manufacturing necessitates reliable, quality components. With scanning electron microscopy, parts cleanliness analysis can be brought inhouse, providing you with a broad range of analytical data and shortening your production cycle.
    Style Sheet for Komodo Tabs
    Techniques
    (S)TEM Sample Preparation
    DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
    Learn more ›
    3D Materials Characterization
    Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
    Learn more ›
    Energy Dispersive X-Ray Spectroscopy Elemental Analysis
    Energy dispersive X-ray spectroscopy (EDS) techniques provide precise elemental analysis with high speed. Key methods include SEM EDS for integrated imaging and quantification and 3D EDS tomography for detailed nanoscale compositional data. These advancements streamline complex material analyses and enhance research efficiency.
    Learn more ›
    Sample preparation for materials science
    We provide comprehensive solutions for sample preparation in materials science, helping ensure precise and reliable results for electron microscopy and other analytical techniques. Our range of instruments allows you to perform cutting, grinding, polishing, and coating of samples, which are crucial steps for achieving high-quality analysis and characterization.
    Learn more ›
    Nanoscale Prototyping
    As technology continues to miniaturize, the demand for nanoscale devices and structures is ever increasing. 3D nanoprototyping with DualBeam instruments helps you to quickly design, create, and inspect micro- and nanoscale functional prototypes.
    Learn more ›
    3D EDS Tomography
    Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
    Learn more ›
    EDS Elemental Analysis
    Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
    Learn more ›
    Atomic-Scale Elemental Mapping with EDS
    Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
    Learn more ›
    Imaging using HRSTEM and HRTEM
    Transmission electron microscopy is invaluable for characterizing the structure of nanoparticles and nanomaterials. High-resolution STEM and TEM enable atomic-resolution data along with information on chemical composition.
    Learn more ›
    Differential Phase Contrast Imaging
    Modern electronics research relies on nanoscale analysis of electric and magnetic properties. Differential phase contrast STEM (DPC-STEM) can image the strength and distribution of magnetic fields in a sample and display the magnetic domain structure.
    Learn more ›
    Imaging Hot Samples
    Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
    Learn more ›
    Multi-technique surface analysis workflow
    To meet the need for extensive characterization of surfaces, we have established multi-technique workflows based on using either the Thermo Scientific ESCALAB CXi XPS Microprobe or the Thermo Scientific Nexsa Surface Analysis System. These instruments are designed as multi-technique workstations to provide comprehensive analyses in a timely and efficient manner. 
    Learn more ›
    X-Ray Photoelectron Spectroscopy
    X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
    Learn more ›
    Environmental SEM (ESEM)
    Environmental SEM allows materials to be imaged in their native state. This is ideally suited for academic and industrial researchers who need to test and analyze samples that are wet, dirty, reactive, outgassing or otherwise not vacuum compatible.
    Learn more ›
    Electron Energy Loss Spectroscopy
    Materials science research benefits from high-resolution EELS for a wide range of analytical applications. This includes high-throughput, high signal-to-noise-ratio elemental mapping, as well as probing of oxidation states and surface phonons.
    Learn more ›
    APT Sample Preparation
    Atom probe tomography (APT) provides atomic-resolution 3D compositional analysis of materials. Focused ion beam (FIB) microscopy is an essential technique for high-quality, orientation, and site-specific sample preparation for APT characterization.
    Learn more ›
    Cross-sectioning
    Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
    Learn more ›
    In Situ experimentation
    Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
    Learn more ›
    Particle analysis
    Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
    Learn more ›
    Cathodoluminescence
    Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.
    Learn more ›
    Multi-scale analysis
    Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
    Learn more ›
    SIMS
    The TOF-SIMS (time-of-flight secondary ion mass spectrometry) detector for focused ion beam scanning electron microscopy (FIB-SEM) tools enables high-resolution analytical characterization of all elements in the periodic table, even at low concentrations.
    Learn more ›
    EDS Analysis with ChemiSEM Technology
    Energy dispersive X-ray spectroscopy for materials characterization.
    Learn more ›
    Automated Particle Workflow
    The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
    Learn more ›
    Correlative Imaging and Surface Analysis Workflow
    The CISA Workflow’s combination of XPS with SEM analysis enables you to accurately add quantitative chemical composition information of the surface to the structural information from the microscope imagery​.
    Learn more ›
    (S)TEM Sample Preparation
    DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
    Learn more ›
    3D Materials Characterization
    Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
    Learn more ›
    Energy Dispersive X-Ray Spectroscopy Elemental Analysis
    Energy dispersive X-ray spectroscopy (EDS) techniques provide precise elemental analysis with high speed. Key methods include SEM EDS for integrated imaging and quantification and 3D EDS tomography for detailed nanoscale compositional data. These advancements streamline complex material analyses and enhance research efficiency.
    Learn more ›
    Sample preparation for materials science
    We provide comprehensive solutions for sample preparation in materials science, helping ensure precise and reliable results for electron microscopy and other analytical techniques. Our range of instruments allows you to perform cutting, grinding, polishing, and coating of samples, which are crucial steps for achieving high-quality analysis and characterization.
    Learn more ›
    Nanoscale Prototyping
    As technology continues to miniaturize, the demand for nanoscale devices and structures is ever increasing. 3D nanoprototyping with DualBeam instruments helps you to quickly design, create, and inspect micro- and nanoscale functional prototypes.
    Learn more ›
    3D EDS Tomography
    Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
    Learn more ›
    EDS Elemental Analysis
    Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
    Learn more ›
    Atomic-Scale Elemental Mapping with EDS
    Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
    Learn more ›
    Imaging using HRSTEM and HRTEM
    Transmission electron microscopy is invaluable for characterizing the structure of nanoparticles and nanomaterials. High-resolution STEM and TEM enable atomic-resolution data along with information on chemical composition.
    Learn more ›
    Differential Phase Contrast Imaging
    Modern electronics research relies on nanoscale analysis of electric and magnetic properties. Differential phase contrast STEM (DPC-STEM) can image the strength and distribution of magnetic fields in a sample and display the magnetic domain structure.
    Learn more ›
    Imaging Hot Samples
    Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
    Learn more ›
    Multi-technique surface analysis workflow
    To meet the need for extensive characterization of surfaces, we have established multi-technique workflows based on using either the Thermo Scientific ESCALAB CXi XPS Microprobe or the Thermo Scientific Nexsa Surface Analysis System. These instruments are designed as multi-technique workstations to provide comprehensive analyses in a timely and efficient manner. 
    Learn more ›
    X-Ray Photoelectron Spectroscopy
    X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
    Learn more ›
    Environmental SEM (ESEM)
    Environmental SEM allows materials to be imaged in their native state. This is ideally suited for academic and industrial researchers who need to test and analyze samples that are wet, dirty, reactive, outgassing or otherwise not vacuum compatible.
    Learn more ›
    Electron Energy Loss Spectroscopy
    Materials science research benefits from high-resolution EELS for a wide range of analytical applications. This includes high-throughput, high signal-to-noise-ratio elemental mapping, as well as probing of oxidation states and surface phonons.
    Learn more ›
    APT Sample Preparation
    Atom probe tomography (APT) provides atomic-resolution 3D compositional analysis of materials. Focused ion beam (FIB) microscopy is an essential technique for high-quality, orientation, and site-specific sample preparation for APT characterization.
    Learn more ›
    Cross-sectioning
    Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
    Learn more ›
    In Situ experimentation
    Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
    Learn more ›
    Particle analysis
    Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
    Learn more ›
    Cathodoluminescence
    Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.
    Learn more ›
    Multi-scale analysis
    Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
    Learn more ›
    SIMS
    The TOF-SIMS (time-of-flight secondary ion mass spectrometry) detector for focused ion beam scanning electron microscopy (FIB-SEM) tools enables high-resolution analytical characterization of all elements in the periodic table, even at low concentrations.
    Learn more ›
    EDS Analysis with ChemiSEM Technology
    Energy dispersive X-ray spectroscopy for materials characterization.
    Learn more ›
    Automated Particle Workflow
    The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
    Learn more ›
    Correlative Imaging and Surface Analysis Workflow
    The CISA Workflow’s combination of XPS with SEM analysis enables you to accurately add quantitative chemical composition information of the surface to the structural information from the microscope imagery​.
    Learn more ›
    Samples
    Battery Research
    Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
    Learn more ›
    Metals Research and Characterization
    Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
    Learn more ›
    Polymers Research
    Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
    Learn more ›
    Geological Research
    Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
    Learn more ›
    Oil and Gas
    As the demand for oil and gas continues, there is an ongoing need for efficient and effective extraction of hydrocarbons. Thermo Fisher Scientific offers a range of microscopy and spectroscopy solutions for a variety of petroleum science applications.
    Learn more ›
    Nanoparticles
    Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
    Learn more ›
    Forensics
    Micro-traces of crime scene evidence can be analyzed and compared using electron microscopy as part of a forensic investigation. Compatible samples include glass and paint fragments, tool marks, drugs, explosives, and GSR (gunshot residue).
    Learn more ›
    Catalysis Research
    Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
    Learn more ›
    Fibers and Filters
    The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
    Learn more ›
    2D Materials
    Novel materials research is increasingly interested in the structure of low-dimensional materials. Scanning transmission electron microscopy with probe correction and monochromation allows for high-resolution two-dimensional materials imaging.
    Learn more ›
    Automotive Materials Testing
    Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
    Learn more ›
    Food Development and Testing
    To ensure food safety, protect your brand, and ensure consumer satisfaction, analytical testing and material characterization instruments avert costly errors and product defects and help you deliver consistently delicious foods and beverages.
    Learn more ›
    Products
    FIB-SEM and Laser Ablation
    • All three beams have same coincident point for accurate and repeatable cut placement
    • Millimeter-scale cross sections with up to 15,000x faster material removal than a typical FIB
    • Statistically relevant deep subsurface and 3D data analysis
    Helios 5 Laser PFIB System
    • Fast, millimeter-scale cross sections
    • Statistically relevant deep subsurface and 3D data analysis
    • Shares all capabilities of the Helios 5 PFIB platform
    Thermo Scientific Helios Hydra plasma focused ion beam scanning electron microscope (DualBeam)
    Helios Hydra DualBeam
    • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
    • Ga-free TEM sample preparation
    • Extreme high resolution SEM imaging
    Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
    • Fully automated, high-quality, ultra-thin TEM sample preparation
    • High throughput, high resolution subsurface and 3D characterization
    • Rapid nanoprototyping capabilities
    Helios 5 PFIB DualBeam
    • Gallium-free STEM and TEM sample preparation
    • Multi-modal subsurface and 3D information
    • Next-generation 2.5 μA xenon plasma FIB column
    Thermo Scientific Scios 2 plasma focused ion beam scanning electron microscope (DualBeam)
    Scios 2 DualBeam
    • Full support of magnetic and non-conductive samples
    • High throughput subsurface and 3D characterization
    • Advanced ease of use and automation capabilities
    Iliad (S)TEM
    • Advanced integration of EELS and (S)TEM optics
    • Electrostatic beam blanker
    • High energy resolution electron source
    Spectra Ultra
    • New imaging and spectroscopy capabilities on the most beam sensitive materials
    • A leap forward in EDX detection with Ultra-X
    • Column designed to maintain sample integrity.
    Spectra 200
    • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
    • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
    • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
    Spectra 300
    • Highest-resolution structural and chemical information at the atomic level
    • Flexible high-tension range from 30-300 kV
    • Three lens condenser system
    Talos F200i TEM
    • Compact design with X-TWIN objective lens
    • Available with S-FEG, X-FEG, and X-CFEG
    • Flexible and fast EDS options for comprehensive elemental analysis
    Talos F200S TEM
    • Intuitive and easy-to-use automation software
    • Available with Super-X EDS for rapid quantitative chemical analysis
    • High-throughput with simultaneous multi-signal acquisition
    Talos F200X TEM
    • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
    • X-FEG and X-CFEG available for the highest brightness and energy resolution
    • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
    Thermo Scientific Talos L120C transmission electron microscope (TEM)
    Talos L120C TEM
    • High versatility and stability
    • 4k x 4k Ceta CMOS camera for speed and large FOV
    • TEM magnification range of 25X to 650kX
    • EDS and STEM options for compositional analyses
    Thermo Scientific Talos F200C transmission electron microscope (TEM)
    Talos F200C TEM
    • High-contrast and high-quality TEM and STEM imaging
    • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
    • Large pole piece gap and multiple in situ options
    Apreo ChemiSEM System
    • Integrated SEM imaging and chemical characterization
    • Enhanced automation to simplify workflows
    • Extended source lifetime and schedulable upgrades
    Axia ChemiSEM
    • Live quantitative elemental mapping
    • High fidelity scanning electron microscopy imaging
    • Flexible and easy to use, even for novice users
    • Easy maintenance
    Thermo Scientific Verios 5 XHR scanning electron microscope (SEM)
    Verios 5 XHR SEM
    • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
    • Easy access to beam landing energies as low as 20 eV
    • Excellent stability with piezo stage as standard
    Thermo Scientific Quattro E scanning electron microscope (SEM)
    Quattro ESEM
    • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
    • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
    Thermo Scientific Prisma E scanning electron microscope (SEM)
    Prisma E SEM
    • Entry-level SEM with excellent image quality
    • Easy and quick sample loading and navigation for multiple samples
    • Compatible with a wide range of materials thanks to dedicated vacuum modes
    Thermo Scientific VolumeScope 2 scanning electron microscope (SEM)
    VolumeScope 2 SEM
    • Isotropic 3D data from large volumes
    • High contrast and resolution in high and low vacuum modes
    • Simple switch between normal SEM use and serial block-face imaging
    Thermo Scientific Apreo 2 scanning electron microscope (SEM)
    Apreo 2 SEM
    • High-performance SEM for all-round nanometer or sub-nanometer resolution
    • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
    • Excellent performance at long working distance (10 mm)
    Phenom G6 Desktop SEMs
    • Durable high-brightness source
    • Compact design
    • Three models to suit your needs
    Phenom XL G2 Desktop SEM
    • For large samples (100x100 mm) and ideal for automation
    • <10 nm resolution and up to 200,000x magnification; 4.8 kV up to 20 kV acceleration voltage
    • Optional fully integrated EDS and BSE detector
    Phenom Pro G6 Desktop SEM
    • High performance desktop SEM
    • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
    • Optional SE detector
    Phenom ProX G6 Desktop SEM
    • High performance desktop SEM with integrated EDS detector
    • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
    • Optional SE detector
    Phenom Pure G6 Desktop SEM
    • Entry level desktop SEM
    • Resolution <15 nm; magnification up to 175,000x
    • Longlife CeB6 source
    Phenom ParticleX Battery Desktop SEM
    • Versatile solution for high-quality, in-house analysis
    • Automated system and analysis of multiple samples
    • Testing 10x faster
    Phenom ParticleX Steel Desktop SEM
    • SEM and EDS integrated
    • Ease of use
    • Sub-micrometer inclusions
    Phenom ParticleX TC Desktop SEM
    • Versatile desktop SEM with automation software for Technical Cleanliness
    • Resolution <10 nm; magnification up to 200,000x
    • Optional SE detector
    Phenom ParticleX GSR Desktop SEM
    • Dedicated automated GSR desktop SEM
    • Resolution <10 nm; magnification up to 200,000x
    • Longlife CeB6 source
    Phenom ParticleX AM Desktop SEM
    • Versatile desktop SEM with automation software for Additive Manufacturing
    • Resolution <10 nm; magnification up to 200,000x
    • Optional SE detector
    Phenom Pharos G2 Desktop FEG-SEM
    • FEG source with 1 – 20 kV acceleration voltage range
    • <2.0 nm (SE) and 3.0 nm (BSE) resolution @ 20 kV
    • Optional fully integrated EDS and SE detector
    K-Alpha XPS
    • High resolution XPS
    • Fast, efficient, automated workflow
    • Ion source for depth profiling
    ESCALAB QXi XPS
    • High spectral resolution
    • Multi-technique surface analysis
    • Extensive sample preparation and expansion options
    Nexsa G2 XPS
    • Micro-focus X-ray sources
    • Unique multi-technique options
    • Dual-mode ion source for monoatomic & cluster ion depth profiling
    CleanMill System
    • Broad ion beam milling and polishing
    • Wide acceleration voltage range
    • Cryogenic sample preparation
    • CleanConnect compatible
    Athena Software
    Imaging Data Management
    • Ensure traceability of images, data, metadata and experimental workflows
    • Simplify your imaging workflow​
    • Improve collaboration
    • Secure and manage data access​
    Avizo Software
    Materials Science
    • Support for multi-data/multi-view, multi-channel, time series, very large data
    • Advanced multi-mode 2D/3D automatic registration
    • Artifact reduction algorithms
    AutoTEM 5
    • Fully automated in situ S/TEM sample preparation
    • Support of top-down, planar and inverted geometry
    • Highly configurable workflow
    • Easy to use, intuitive user interface
    AutoScript 4
    • Improved reproducibility and accuracy
    • Unattended, high throughput imaging and patterning
    • Supported by Python 3.5-based scripting environment
    Velox
    • An experiments panel on the left side of the processing window.
    • Live quantitative mapping
    • Interactive detector layout interface for reproducible experiment control and setup
    Thermo Scientific Inspect 3D tomography software
    Inspect 3D Software
    • Image processing tools and filters for cross-correlation
    • Feature tracking for image alignment
    • Algebraic reconstruction technique for iterative projection comparison
    Thermo Scientific Maps electron microscopy software
    Maps Software
    • Acquire high-resolution images over large areas
    • Easily find regions of interest
    • Automate image acquisition process
    • Correlate data from different sources
    Nanobuilder
    • CAD-based prototyping
    • Fully automated job execution, stage navigation, milling, and deposition
    • Automated alignment and drift control
    ProSuite
    • Automated collection of images
    • Real-time remote control
    • Standard applications included: Automated Image Mapping + Remote User Interface
    PoroMetric
    • Correlate pore features such as area, aspect ratio, major and minor axis
    • Acquire images directly from the Desktop SEM
    • Statistical data with high-quality images
    ParticleMetric
    • Integrated software in ProSuite for online and offline analysis
    • Correlating particle features such as diameter, circularity, aspect ratio and convexity
    • Creating image datasets with Automated Image Mapping
    Quartz PCI/CFR
    • SEM imaging traceability compliant with 21 CFR Part 11
    • Compatible with the Phenom XL and Phenom Pro desktop SEMs
    • Windows 10 64-bit operating system support
    Thermo Scientific Auto Slice and View 4.0 serial section electron microscopy software
    Auto Slice and View 4.0 Software
    • Automated serial sectioning for DualBeam
    • Multi-modal data acquisition (SEM, EDS, EBSD)
    • On-the-fly editing capabilities
    • Edge based cut placement
    FiberMetric
    • Save time by automated measurements
    • Fast and automated collection of all statistical data
    • View and measure micro and nano fibers with unmatched accuracy
    3D Reconstruction
    • Intuitive user interface, maximum employability
    • Intuitive fully automated user interface
    • Based on 'shape from shading' technology, no stage tilt required
    Elemental Mapping
    • Fast and reliable information on the distribution of elements within the sample or the selected line
    • Easily exported and reported results
    Phenom Programming Interface
    • Customize your SEM to fit your workflow
    • Increase efficiency and save time with automated processes
    • Control imaging settings and stage navigation
    Service
    EM Services for DualBeam (FIB SEM) Microscope
    • Become an expert on all aspects of your workflow
    • Receive enhanced visibility to product performance
    • Enjoy comprehensive maintenance and committed uptime
    Style Sheet for Instrument Cards Original
    Style Sheet to change H2 style to p with em-h2-header class
    Contact us
    Style Sheet for Support and Service footer
    Style Sheet for Fonts
    Style Sheet for Cards
    Electron microscopy services for
    the materials science
    To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.