Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
L-Alanyl-Glutamine | 217.0 | 406.0 | 1.8709677 |
L-Arginine hydrochloride | 211.0 | 126.0 | 0.5971564 |
L-Cystine | 240.0 | 24.0 | 0.1 |
L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
L-Isoleucine | 131.0 | 52.0 | 0.39694658 |
L-Leucine | 131.0 | 52.0 | 0.39694658 |
L-Lysine hydrochloride | 183.0 | 73.0 | 0.3989071 |
L-Methionine | 149.0 | 15.0 | 0.10067114 |
L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
L-Threonine | 119.0 | 48.0 | 0.40336135 |
L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
L-Tyrosine | 181.0 | 36.0 | 0.19889502 |
L-Valine | 117.0 | 46.0 | 0.3931624 |
Vitamins | |||
Choline chloride | 140.0 | 1.0 | 0.007142857 |
D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
Folic Acid | 441.0 | 1.0 | 0.0022675737 |
Niacinamide | 122.0 | 1.0 | 0.008196721 |
Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
i-Inositol | 180.0 | 2.0 | 0.011111111 |
Inorganic Salts | |||
Calcium Chloride (CaCl2-2H2O) | 147.0 | 264.0 | 1.7959183 |
Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 200.0 | 0.8130081 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
Sodium Chloride (NaCl) | 58.0 | 6800.0 | 117.24138 |
Sodium Phosphate monobasic (NaH2PO4-2H2O) | 156.0 | 158.0 | 1.0128205 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
Phenol Red | 376.4 | 10.0 | 0.026567481 |
1. Biol. Med. Sci., 61:439. (rat astrocytes).
2. Eagle, H. (1959) Science 130:432.
3. Lolait, S.J., Harmer, J.H., Dahl, D., et al., (1983) Aust. J. Exp.
4. Norris, S.J. and Edmondson, D.G. (1986) Sex. Transm. Dis., 13:207 (Rabbit epithelial cells).
42360 - MEM, HEPES, GlutaMAX(TM)
Cat No:
42360024
,
42360032
,
42360081
,
42360099
32561 - MEM alpha, GlutaMAX(TM), no nucleosides
Cat No:
32561029
,
32561037
,
32561094
,
32561102
32571 - MEM alpha, GlutaMAX(TM), nucleosides
Cat No:
32571028
,
32571036
,
32571093
,
32571101