Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Inorganic Salts | |||
Sodium Chloride (NaCl) | 58.0 | 5000.0 | 86.206894 |
Other Components | |||
SELECT Peptone 140 (pancreatic digest of casein) | 10000.0 | Infinity | |
SELECT Yeast Extract autolyzed low sodium | 5000.0 | Infinity |
LB Agar, powder (Lennox L Agar)
Cat No:
22700025
,
22700041
LB Broth Base, powder (Lennox L Broth Base)
Cat No:
12780029
,
12780052