Technical Resources
2-YT Broth
INTENDED USE: For growth of hosts for replication of M13 vectors. INSTRUCTIONS FOR USE: Suspend 31 g in 1 L of demineralized water. Autoclave for 15 min at 121°C. RECOMMENDED STORAGE CONDITION: 15°C TO 30°C.
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Sodium Chloride (NaCl)5.0Infinity
Other Components
SELECT Peptone 140 (pancreatic digest of casein)16.0Infinity
SELECT Yeast Extract10.0Infinity
Reference:
Related Products:
Terrific Broth
Cat No: 22711022
Gibco® BG-11 Media: Optimized for Cyanobacteria
Cat No: a1379901 , A1379902
TAP Growth Media, optimized for Chlamydomonas culture
Cat No: A1379801 , A1379802