Search Thermo Fisher Scientific
Catalog Number(s)
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
L-Arginine hydrochloride | 211.0 | 126.0 | 0.5971564 |
L-Cystine 2HCl | 313.0 | 31.0 | 0.09904154 |
L-Glutamine | 146.0 | 292.0 | 2.0 |
L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
L-Isoleucine | 131.0 | 52.0 | 0.39694658 |
L-Leucine | 131.0 | 52.0 | 0.39694658 |
L-Lysine hydrochloride | 183.0 | 73.0 | 0.3989071 |
L-Methionine | 149.0 | 15.0 | 0.10067114 |
L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
L-Threonine | 119.0 | 48.0 | 0.40336135 |
L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
L-Tyrosine disodium salt dihydrate | 261.0 | 52.0 | 0.19923371 |
L-Valine | 117.0 | 46.0 | 0.3931624 |
Vitamins | |||
Choline chloride | 140.0 | 1.0 | 0.007142857 |
D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
Folic Acid | 441.0 | 1.0 | 0.0022675737 |
Niacinamide | 122.0 | 1.0 | 0.008196721 |
Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
i-Inositol | 180.0 | 2.0 | 0.011111111 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 140.0 | 1.2612612 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 98.0 | 0.81666666 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Potassium Phosphate monobasic (KH2PO4) | 136.0 | 60.0 | 0.44117647 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 350.0 | 4.1666665 |
Sodium Chloride (NaCl) | 58.0 | 8000.0 | 137.93103 |
Sodium Phosphate dibasic (Na2HPO4) anhydrous | 142.0 | 48.0 | 0.33802816 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
Phenol Red | 376.4 | 10.0 | 0.026567481 |
1. Eagle, H. (1959) Science 130:432.