Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Other Components | |||
Arachidonic Acid | 304.74 | 2.0 | 0.006562972 |
Cholesterol | 386.65 | 220.0 | 0.56899005 |
DL-alpha-Tocopherol Acetate | 472.74 | 70.0 | 0.14807294 |
Ethyl Alcohol 100% | confidential | n/a | |
Linoleic Acid | 280.45 | 10.0 | 0.035656977 |
Linolenic Acid | 278.44 | 10.0 | 0.03591438 |
Myristic Acid | 228.38 | 10.0 | 0.04378667 |
Oleic Acid | 282.47 | 10.0 | 0.03540199 |
Palmitic Acid | 256.43 | 10.0 | 0.038997 |
Palmitoleic Acid | 254.41 | 10.0 | 0.03930663 |
Pluronic F-68 | 7680.0 | 90000.0 | 11.71875 |
Stearic Acid | 284.48 | 10.0 | 0.035151854 |
Tween 80® | 1310.0 | 2200.0 | 1.6793894 |