Search Thermo Fisher Scientific
B-27 Supplements are 50X liquid serum-free supplements for growth and long-term viability of hippocampal neurons.
B-27 Supplement Minus AO is the complete B-27 Supplement without 5 antioxidants (vitamin E, vitamin E acetate, superoxide dismutase, catalase, and glutathione) that would interfere with studying freeradical damage to neurons.
Catalog Number(s)
10889038Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Vitamins | |||
Biotin | confidential | n/a | |
Vitamin A (acetate) | confidential | n/a | |
Proteins | |||
BSA, fatty acid free Fraction V | confidential | n/a | |
Human Recombinant Insulin | confidential | n/a | |
Human Transferrin | confidential | n/a | |
Other Components | |||
Corticosterone | confidential | n/a | |
D-Galactose | confidential | n/a | |
Ethanolamine HCl | confidential | n/a | |
L-Carnitine HCl | confidential | n/a | |
Linoleic Acid | confidential | n/a | |
Linolenic Acid | confidential | n/a | |
Progesterone | confidential | n/a | |
Putrescine 2HCl | confidential | n/a | |
Sodium Selenite | confidential | n/a | |
T3 (triodo-I-thyronine) | confidential | n/a |
B-27 Serum-Free Supplement (50X) liquid
Cat No:
17504001
,
17504044
B-27 Supplement Minus Vitamin A (50X) liquid
Cat No:
12587001
,
12587010