Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 200.0 | 2.6666667 |
L-Alanine | 89.0 | 225.0 | 2.52809 |
L-Arginine | 174.0 | 500.0 | 2.8735633 |
L-Asparagine | 132.0 | 250.0 | 1.8939394 |
L-Cysteine | 121.0 | 120.0 | 0.9917355 |
L-Glutamine | 146.0 | 300.0 | 2.0547945 |
L-Histidine | 155.0 | 250.0 | 1.6129032 |
L-Isoleucine | 131.0 | 250.0 | 1.908397 |
L-Leucine | 131.0 | 125.0 | 0.9541985 |
L-Lysine | 146.0 | 75.0 | 0.51369864 |
L-Methionine | 149.0 | 75.0 | 0.5033557 |
L-Phenylalanine | 165.0 | 125.0 | 0.75757575 |
L-Serine | 105.0 | 200.0 | 1.9047619 |
L-Threonine | 119.0 | 300.0 | 2.5210085 |
L-Tryptophan | 204.0 | 20.0 | 0.09803922 |
L-Tyrosine | 181.0 | 300.0 | 1.6574585 |
L-Valine | 117.0 | 100.0 | 0.85470086 |
Vitamins | |||
Choline chloride | 140.0 | 1.0 | 0.007142857 |
D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
Folic Acid | 441.0 | 1.0 | 0.0022675737 |
Niacinamide | 122.0 | 1.0 | 0.008196721 |
Pyridoxine hydrochloride | 206.0 | 1.0 | 0.004854369 |
Riboflavin 5'-phosphate Na | 478.0 | 0.1 | 2.0920503E-4 |
Thiamine monophosphate | 442.0 | 1.0 | 0.0022624435 |
i-Inositol | 180.0 | 2.0 | 0.011111111 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 140.0 | 1.2612612 |
Magnesium Chloride (anhydrous) | 95.0 | 93.7 | 0.9863158 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Potassium Phosphate monobasic (KH2PO4) | 136.0 | 60.0 | 0.44117647 |
Sodium Chloride (NaCl) | 58.0 | 8000.0 | 137.93103 |
Sodium Phosphate dibasic (Na2HPO4) anhydrous | 142.0 | 190.0 | 1.3380282 |
Other Components | |||
D+ Galactose | 180.0 | 900.0 | 5.0 |
Phenol Red | 398.0 | 10.0 | 0.025125628 |
Sodium Pyruvate | 110.0 | 550.0 | 5.0 |