Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 50.0 | 0.6666667 |
L-Alanine | 89.0 | 25.0 | 0.28089887 |
L-Arginine hydrochloride | 211.0 | 126.98 | 0.601801 |
L-Asparagine-H2O | 150.0 | 50.0 | 0.33333334 |
L-Aspartic acid | 133.0 | 30.0 | 0.22556391 |
L-Cysteine hydrochloride-H2O | 176.0 | 100.0 | 0.5681818 |
L-Cystine 2HCl | 313.0 | 31.0 | 0.09904154 |
L-Glutamic Acid | 147.0 | 75.0 | 0.5102041 |
L-Glutamine | 146.0 | 292.0 | 2.0 |
L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
L-Isoleucine | 131.0 | 52.4 | 0.4 |
L-Leucine | 131.0 | 52.0 | 0.39694658 |
L-Lysine hydrochloride | 183.0 | 73.0 | 0.3989071 |
L-Methionine | 149.0 | 15.0 | 0.10067114 |
L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
L-Proline | 115.0 | 40.0 | 0.3478261 |
L-Serine | 105.0 | 25.0 | 0.23809524 |
L-Threonine | 119.0 | 48.0 | 0.40336135 |
L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
L-Tyrosine disodium salt | 262.0 | 52.0 | 0.19847329 |
L-Valine | 117.0 | 46.0 | 0.3931624 |
Vitamins | |||
Ascorbic Acid | 176.0 | 50.0 | 0.2840909 |
Biotin | 244.0 | 0.1 | 4.0983607E-4 |
Choline chloride | 140.0 | 1.0 | 0.007142857 |
D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
Folic Acid | 441.0 | 1.0 | 0.0022675737 |
Niacinamide | 122.0 | 1.0 | 0.008196721 |
Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
Vitamin B12 | 1355.0 | 1.36 | 0.0010036901 |
i-Inositol | 180.0 | 2.0 | 0.011111111 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 200.0 | 1.8018018 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
Sodium Chloride (NaCl) | 58.0 | 6800.0 | 117.24138 |
Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 140.0 | 1.0144928 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
Lipoic Acid | 206.0 | 0.2 | 9.708738E-4 |
Phenol Red | 376.4 | 10.0 | 0.026567481 |
Sodium Pyruvate | 110.0 | 110.0 | 1.0 |
1. Simonsen, C.C. and Levinson, A.D. 1983. Isolation and expression of an altered mouse dihydrofolate reductase cDNA. Proc. Natl. Acad. Sci. U.S.A. 80:2495-2499.
12571 - MEM alpha, nucleosides
Cat No:
12571048
,
12571063
,
12571071
,
12571089
41061 - MEM alpha, nucleosides, no phenol red
Cat No:
41061029
,
41061037
22571 - MEM alpha, nucleosides
Cat No:
22571020
,
22571038
22561, MEM alpha, no nucleosides
Cat No:
22561021
,
22561054
A10490 - MEM alpha, nucleosides, no ascorbic acid
Cat No:
A1049001