Actin filaments of chick heart fibroblasts stained with rhodamine phalloidin
Actin filaments of chick heart fibroblasts stained with rhodamine phalloidin (Cat. no. R415). The subcompartments in the cytoskeleton are readily apparent and labeled as follows: sf, stress fiber; lam, lamellipodium; fil/ms, filipodium/microspike; am, actin meshwork; arc, dorsal arc. Figure reprinted from "Visualizing the Actin Cytoskeleton." J. Small et al. Microscopy Research and Technique 47, 3–17 (1999). Reprinted by permission of Wiley-Liss, Inc., a subsidiary of John Wiley & Sons, Inc., and J. Victor Small.
Actin filaments of chick heart fibroblasts stained with rhodamine phalloidin (Cat. no. R415). The subcompartments in the cytoskeleton are readily apparent and labeled as follows: <b>sf</b>, stress fiber; <b>lam</b>, lamellipodium; <b>fil/ms</b>, filipodium/microspike; <b>am</b>, actin meshwork; <b>arc</b>, dorsal arc. Figure reprinted from
Related Products
Catalog # Name Size Price (USD) Qty
R415 Rhodamine Phalloidin, Red-orange, 540/565 nm Each
605.65
Online Exclusive
624.00
Save 18.35 (3%)
Related Images