1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain.
A twofold dilution series of Escherichia coli 16S and 23S ribosomal RNA (rRNA) that has been electrophoresed on a nondenaturing 1% agarose gel and then stained with our SYBR® Green II RNA gel stain (Cat. no. S7564, S7568, S7586). Gel staining was visualized with 254 nm epi-illumination.
A twofold dilution series of <i>Escherichia coli</i> 16S and 23S ribosomal RNA (rRNA) that has been electrophoresed on a nondenaturing 1% agarose gel and then stained with our SYBR® Green II RNA gel stain (Cat. no. S7564, S7568, S7586).  Gel staining was visualized with 254 nm epi-illumination.
Related Products
Catalog # Name Size Price (USD) Qty
S7564 SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO, 500 μL Each
469.65
Online Exclusive
504.00
Save 34.35 (7%)
S7568 SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO, 1 mL Each
751.65
Online Exclusive
806.00
Save 54.35 (7%)
S7586 SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO, 20 x 50 μL Each
886.65
Online Exclusive
954.00
Save 67.35 (7%)
Related Images