Bone marrow aspirates provides unique and valuable research data, but cell numbers can be very low.  For such critical samples you need a quality medium that you can trust. MarrowMAX™ medium arrives in your lab complete and ready to use, fully supplemented with serum, a novel human stromal conditioned medium, antibiotics, and L-glutamine.  By eliminating the preparation steps and supplementation required with other bone marrow media, you save time, labor, money, and reduce the risk of contamination.
Product Information
MarrowMAX™ medium far outperforms commercially available formulations supplemented with giant cell tumor (GCT) conditioned media (Figure 1), providing consistent lot-to-lot performance (Figure 2) with a higher mitotic index and superior chromosome morphology.
 Figure 1 - Superior performance of MarrowMAX™ medium.  Cells were cultured in: basal medium without conditioned medium; MarrowMAX™ medium; supplier medium (GCT-CM 1); and supplier 2 medium (GCT-CM 2) (both supplier 1 and supplier 2 media contain GCT-conditioned medium).  Mitotic cells were assayed 24 hrs after plating.
 Figure 2 - Consistency of MarrowMAX™ medium.  Normal bone marrow mononuclear cells were seeded at 1x106 cells/ml in 4 different lots of MarrowMAX™ medium.  BrdU updake was measured by absorbance at 405 nm.  Results are mean ± SEM for n = 10.
Ordering information
Catalog # Name Size Price (USD) Qty
12260014 MarrowMAX™ Bone Marrow Medium, 100 mL Each
275.00
12260001 MarrowMAX™ Bone Marrow Medium, 100 mL Each
1,236.00
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions