Thermo Scientific Chromeleon 7.3.2 Chromatography Data System (CDS) was built to simplify your laboratory workflow and to give you superior insights into your data, delivering ease of use, comprehensive instrument control, automated data analysis, and flexible reporting. From constantly changing research and discovery methods, to routine testing and anything in between, Chromeleon CDS streamlines your entire workflow, delivering better results, faster, and with more confidence.
Learn how Chromeleon Software can boost your laboratory efficiency with one CDS designed to consolidate laboratory operations and deliver significant productivity and efficiency gains.
What are users saying?
"Due to policy, I can't tell much of a story. Let's just say I wish I'd adopted Chromeleon a few years ago before it became my Company's corporate standard."
- Senior Scientist, P&G
Benefits of Chromeleon 7.3.1. CDS for labs
  • Universal Instrument Control: Offering comprehensive, reliable control for over 550 different instrument modules (including CE drivers) and over 35 Mass Spectrometry instruments. Benefit from one software to operate your entire lab
  • Increased confidence in results: With dual sequence processing capabilities you can merge data to enable cross-confirmation between techniques. Speed up data processing, improve efficiency and reduce the risk of misreporting
  • Improved productivity: Use automation tools for fast optimization of your integration and achieve considerable efficiency gains with processing improvements for MS data
  • Ensure data integrity: Secure, administrator-controlled user access and permissions to enable compliance with GxP and 21 CFR Part 11
  • Minimized training requirements: Simplify data processing with a customizable and streamlined user interface. Guide operators step by step with tailored view settings to enable analysts to focus their time on critical tasks
  • Connected software environment: Super-charge Chromeleon CDS with the Ardia Platform and connect projects, people and instruments in one simple, easy-to-use interface
Get more "right first-time" results
Get more "right-first-time" results
Chromeleon CDS was designed with scientists and technicians in mind. It simplifies repetitive tasks, reduces errors and helps you minimize out of specification results. It’s based on something we call Operational Simplicity, improving your lab efficiency through use of:
  •  eWorkflow procedures allowing streamlined sequence creation and ensuring users follow SOPs while minimizing training requirements
  • Automated System Suitability Checks and  Intelligent Run Control enabling more "right-first-time" analyses with automated in-run actions to pass/fail decisions
  • Built in report templates and comprehensive, yet easy-to-customize Excel-style reporting engine enables the rendering of large reports and faster result review by flagging outliers via conditional formatting
Improve productivity by doing more with less
Improve productivity by doing more with less
Achieve considerable efficiency gains with one centrally implemented CDS for both chromatography and MS workflows. Chromeleon CDS streamlines laboratory operations making it easier to train lab staff and faster to implement, plus it enables access to all your resources, data and results from any location. Deliver results, faster by use of:
Connect your laboratory
Connect your laboratory
Network existing Chromeleon CDS stations to create a compliant, laboratory-wide solution. Utilize network resources and centralize administration and get enhanced integration to other laboratory software, such as:
Make compliance a competitive advantage
Make compliance a competitive advantage
Built for compliance from the ground up, Chromeleon CDS helps you meet the latest regulatory requirements and still improve productivity. It assists with achieving, maintaining and demonstrating compliance, enabling your laboratory to keep up with ever-evolving standards and regulations through:
  • Traceability with automatically-created comprehensive set of secure, computer-generated time-stamped audit trails
  • Secure, administrator-controlled user access and permissions
  • Electronic signatures
  • Versioning of data objects to understand and view the history of data and the impact on results
Chromeleon CDS – Driving Ease of Use
Setting your laboratory up for success requires selecting the right chromatography data system and ensuring you realize its full capabilities.
Learn how to drive efficiency while ensuring compliance, eliminate errors and streamline data analysis workflows using Chromeleon CDS.
Watch webinar ›
Built for the Lab without compromise
The laboratory is a centralized hub of activity from sample analysis through to result generation, so it is important that processes are efficient, errors are eliminated, and regulatory requirements are adhered to.
In this series, we discuss how a modern CDS can help you realize your laboratory’s potential with best practices, tips and tricks to help you work smarter and increase productivity while ensuring compliance.
View series
High Force Research streamlines operations with Chromeleon CDS
Chromeleon CDS is designed to drive lab productivity through the use of innovative software tools that are built for both IT and the lab.
High Force Research explains how Chromeleon CDS has increased lab productivity, maximized return on investment and delivered a solution that’s ready for future business expansion.
Download case study ›
What's new in Chromeleon 7.3.2 CDS
Chromeleon 7.3.2 CDS delivers vast improvements in performance, compliance, and usability while increasing the resilience of network operations.
Chromeleon CDS builds security, quality and trust
Australian-based Ego Pharmaceutical shares the process they used to select their laboratory software - Chromeleon CDS.
Ensure Business Continuity with Chromeleon CDS
Chromeleon Software delivers a solution that minimizes and protects against system outages and malicious attacks, ensuring uninterrupted access.
The No-Compromise Chromeleon CDS
Chromeleon Software bridges the needs of IT and the lab, offering better spend efficiency and easier, complete compliance plus increased efficiency.
* Required field
*
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Form Code v3.0
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD SchemaApp code