Be ready for tomorrow’s most challenging demands
Exceed experimental expectation with the MultiOmics, Structural Biology, and BioPharma Editions of the Thermo Scientific Orbitrap Ascend Editions Tribrid Mass Spectrometer.
With new capabilities for multiplexed quantitative proteomics, native protein characterization, and deep biopharmaceutical analyses, the three new Editions deliver ultimate experimental throughput, versatility, and ease-of-use for your specific needs.
Detect and characterize the most difficult protein complexes and native samples without losing low m/z instrument performance and flexibility with the new Native MS option, including world class Orbitrap detection up to m/z 16,000 and ion isolation in the ion trap and quadrupole up to m/z 8,000, with isolation widths down to m/z 100 and m/z 5, respectively.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Orbitrap Ascend Editions Tribrid MS
      Orbitrap Ascend MultiOmics Tribrid MS
      Discover more from comprehensive metabolomics to proteomics.
      Orbitrap Ascend Structural Biology Tribrid MS
      Take advantage of ultimate versatility for native protein characterization.
      Orbitrap Ascend BioPharma Tribrid MS
      Successfully characterize your most complex biotherapeutics.