Protocols
Introduction
Agarose 1000 is a specialized agarose that exhibits the following properties:
  • high resolution of PCR products and other short DNA fragments
  • better handling because of a stronger gel structure
  • improved clarity of the gel, enhancing visibility
  • excellent mechanical strength
Specifications                           4% Conc.
Gel Strength                              ≥ 1,400 g/cm2
Gel Point                                    ≤ 32.5–38°C
Melting Point                              ≥ 90°C
Usage
Refer to the following table for the recommended concentration of Agarose 1000 to resolve DNA fragments of the approximate listed range:
Fragment Size% Agarose (in 1X TAE)% Agarose (in 1X TBE)
500–1,00032
100–50043
10–10065
Note: TBE is preferred for small (<1 kb) DNA as it shows increased resolution of small DNA.
Ordering Information
Catalog # Name Size Price (USD) Qty
16550100 UltraPure™ Agarose-1000 Each
445.65
Online Exclusive
487.00
Save 41.35 (8%)
Dissolving Agarose 1000
Note:   Remember to take the thickness of the gel into account when determining the amount of agarose solution needed to cast your gel, as it affects both well volume and power requirements.
Method 1: Microwave (recommended for concentrations ≤3%)
  1. Determine the amount of agarose solution needed to cast your gel.
  2. Add chilled buffer (TAE or TBE) into a flask that can hold 2–4 times the volume of your agarose solution. Place a magnetic stir bar into the flask.
  3. Put the flask on a magnetic stirrer and slowly sprinkle the required amount of agarose powder into the flask as the solution mixes, to prevent the formation of agarose clumps.
  4. Remove the stir bar.
  5. Weigh the flask and solution before heating.
  6. Cover the mouth of the flask with plastic wrap, and pierce the wrap with a small hole for ventilation.
  7. Place the flask in the microwave oven and heat for two minutes.
  8. Remove the flask carefully, and swirl gently to resuspend any agarose particles. Exercise caution – microwaved solution may become superheated and foam over when agitated.
  9. Reheat the solution at 15–20 second intervals or until the solution comes to a boil, and all the agarose particles are dissolved.
  10. Remove the flask carefully and swirl gently to mix the solution.
  11. Place the flask on a scale, and bring it back to its initial weight (from Step 5) with warm distilled water.
  12. Mix gently and cool to 50–60°C (at room temperature for at least 20 minutes) before pouring the solution into the casting tray.
Method 2: Boiling water bath (for all concentrations, especially 4%–5%)
  1. Determine the amount of agarose solution needed to cast your gel.
  2. Add chilled buffer into a flask that can hold 2–4 times the volume of your agarose solution. Place a magnetic stir bar into the flask.
  3. Put the flask on a magnetic stirrer and slowly sprinkle the required amount of agarose powder into the flask as the solution mixes, to prevent the formation of agarose clumps.
  4. Weigh the flask and solution before heating.
  5. Cover the mouth of the flask with plastic wrap, and pierce the wrap with a small hole for ventilation.
  6. Bring the solution to a boil while stirring, and allow it to boil gently for approximately 10 minutes or until the agarose is completely dissolved.
  7. Place the flask on a scale, and bring it back to its initial weight (from Step 4) with warm distilled water.
  8. Mix gently and cool to 50-60°C (at room temperature for at least 20 minutes) before pouring the solution into the casting tray.
Method 3: Autoclave (for all concentrations, especially  ≥ 5%)
  1. Determine the amount of agarose solution needed to cast your gel.
  2. Add chilled buffer into a flask that can hold 2–4 times the volume of your agarose solution. Place a magnetic stir bar into the flask.
  3. Put the flask on a magnetic stirrer and slowly sprinkle the required amount of agarose powder into the flask as the solution mixes, to prevent the formation of agarose clumps.
  4. Remove the stir bar.
  5. Place the flask in the microwave oven and heat for two minutes.
  6. Cover the mouth of the flask with aluminum foil to prevent spillover and autoclave at 121°C for 15 minutes.
  7. Remove the flask from the autoclave and allow it to cool to 50–60°C before pouring the solution into the tray.
Visualization of DNA
For visualization of DNA in the gel, a fluorescent dye can be added to the agarose solution just prior to pouring, or the gel can be stained after electrophoresis. For the intercalating dye ethidium bromide, use a final concentration of 0.5 μg/ml. If more sensitive detection is required, use SYBR® Green I nucleic acid gel stain (Invitrogen Cat. no. S-7563), or SYBR® Safe DNA gel stain (Invitrogen Cat. no. S33102). Refer to the appropriate instructions for these products for in-gel staining, or post-staining protocols.
Dye Mobility
Refer to the following table for the migration of Bromophenol Blue and Xylene Cyanol tracking dyes in relation to DNA:
% Agarose
Bromophenol Blue
Xylene Cyanol
TAE
TBE
TAETBE
2.5130
70
950700
3 80
40
650500
4 40
20
350250
5
30
8
200
140
6
20
4
120
90
251053W       11-Nov-2008