Baculovirus Insect Expression System
The only optimized baculovirus insect expression system with superior protein yields: Gibco ExpiSf Expression System
Experience the first-ever chemically defined baculovirus expression system that can generate three times more protein compared to existing insect expression platforms. The ExpiSf Expression System enables greater consistency across multiple expression runs, all in less time. Put the newest member of the Expi system family to the test in your lab, from discovery to production.
Do you need guidance on a new project, troubleshooting expertise, or more information on a product?
Share your protein expression application needs with us, and one of our protein expression specialists will contact you to discuss the solutions we can provide.
Contact a specialist
Discover the Gibco ExpiSf Expression System
Video Player is loading.
Current Time 0:00
Duration 1:16
Loaded: 0%
Stream Type LIVE
Remaining Time 1:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    “The new ExpiSf Expression System offers a convenient virus generation and protein expression cell line in only two steps, for both secreted and intracellular proteins,we predict that using the ExpiSf system will save us about 30 percent in the time taken for protein expression and make cell maintenance simpler.”  Xuan Droz, Ph.D., project leader, Redbiotec AG
    The first-ever chemically defined baculovirus insect expression system
    Undefined insect cell media components like yeastolate make it very difficult to get consistent performance. The Gibco ExpiSf system is the first ever baculovirus expression system that includes both a chemically defined medium and protein expression enhancer for reliable results you can count on. This insect cell expression system also utilizes a non-engineered derivative of Sf9 insect cells that have been adapted for high-density suspension growth and a specialized vector for insect cells.
    Multiple components of the Gibco ExpiSf Expression System
    Included in the ExpiSf Expression System:
    1. Two vials of frozen ExpiSf9 Cells
    2. 1 L ExpiSf CD Medium
    3. One Bac-to-Bac Vector Kit*
    4. 4 mL ExpiSf Enhancer
    5. 0.5 mL MAX Efficiency DH10Bac Competent Cells*
    6. 1 mL ExpiFectamine Sf Transfection Reagent
    7. 500 mL Opti-MEM I Medium, with phenol red*
    8. 24 Nalgene Single-Use PETG vented 125-mL Erlenmeyer Flasks**
    *not included in China
    ** not included in all regions
    ONE system with remarkably consistent results
    The ExpiSf Expression System is designed to deliver consistent cell growth and protein expression, run after run. With our new Sf9 expression system, inconsistent results are a thing of the past. The superior performance of the chemically defined ExpiSf system delivers consistent results across expression runs and over multiple cell passages. One reliable system gives you one less variable to worry about.
    ONE system for superior protein yields
    It’s revolutionary—the ExpiSf system outperforms existing Sf9 expression platforms with 3x higher protein yield. The ExpiSf Expression System achieves protein yields that are vastly superior to traditional insect platforms. In performance tests against other suppliers’ media, the ExpiSf system demonstrated greater protein expression, delivering 3x more protein.
    Figure 1. Recombinant protein titers in ExpiSf and other baculovirus expression systems. Expression levels of green fluorescent protein (GFP), Fc fusion protein, and tumor necrosis factor alpha (TNF-a) in ExpiSf and other baculovirus-based insect cell protein expression products are shown. ExpiSf protein titers are >3, >4, or >5 times those of other supplier’s insect products, respectively.
    expression of three different protein categories using ExpiSf Expression Systems
    Figure 2. Expression of three different protein categories achieve expression levels >3x higher in the ExpiSf Expression System. Comparative western blotting analysis of a membrane protein (top panel), a receptor-type protein kinase (middle panel) and a tyrosine-protein kinase (bottom panel) produced in ExpiSf and control (Sf9 cells in Sf-900 II SFM) is shown. P0, P1, and P2 signify non-amplified (P0) or amplified (P1, P2) baculovirus stock used for the respective groups. *1.25x104 total cells/lane, **2.5x104 total cells/lane.
    ExpiSf System cost effectiveness
    Table 1. ExpiSf is cost effective compared to other products for baculovirus-based insect protein expression systems.
    NOTE: Average cost to express 100 mg of protein with ExpiSf System or a combination of competitor products. Cost show is based on list price in USD.
     Thermo FisherThermo FisherThermo FisherExpression SystemsMilliporeSigmaOxford Expression TechnologiesHyClone
    CellsExpiSf9Sf9Sf9Sf9Sf9Sf9Sf9
    MediumExpiSf CDSf-900 II SFMSf-900 III SFMESF 921™EX-CELL™ 420ESF 921™SFX-Insect™
    Transfection ReagentExpiFectamine SfCellfectin IICellfectin II
    Expres2 TRInsect GeneJuice™Expres2 TRCellfectin II
    Baculovirus SystemBac-to-BacBac-to-BacBac-to-BacBestBac™BacMagic™flashBAC™Bac-to-Bac
    Cost of 1L expression$334$235$260$186$332$110$243
    Culture volume (mL) for equivalent yield*300120017003100170031001500
    Number of 2L shake flasks needed1236363
    Total cost for same amount of protein$112$294$454$579$563$343$368
    Savings $65$226$351$335$115$139
    ONE optimized and fully integrated system
    System components work synergistically for maximum performance. Other suppliers offer separate reagents for protein production using the baculovirus expression vector system, but there is only one system that’s fully optimized with a streamlined workflow. The ExpiSf system contains integrated components that work together to deliver maximum protein yields.
    Schematic diagram showing optimized system components achieving 3x results
    ONE step from baculovirus production to protein expression
    The ExpiSf Expression System delivers protein in half the time. Bacmid DNA transfection in suspension using the ExpiSf system allows for efficient baculovirus generation without the need for virus amplification. That means you can get your protein in as few as six days.
    Standard protocol for baculovirus generation compared to shorter protocol with ExpiSf Expression System for baculovirus generation.
    ONE system that scales from discovery to production
    ExpiSf CD Media
    The ExpiSf system accelerates development from small to large scale. From deep-well plates to shake flasks, the ExpiSf system delivers the same high levels of volumetric yield, making the system excellent for both discovering new therapies and producing them in large quantities. We manufacture ExpiSf CD Medium in cGMP-compliant facilities and offer custom packaging to scale with your development, so you can be confident that our product quality enhances yours.
    The ExpiSf expression system delivers the same volumetric yield
    Ordering information
    Catalog # Name Size Price (USD) Qty
    A38841 ExpiSf™ Expression System Starter Kit, 1 kit Each
    2,360.00
    Reorder ExpiSf Expression System Kit components
    Catalog # Name Size Price (USD) Qty
    A3767805 ExpiSf™ CD Medium, 20 L Each
    2,274.65
    Online Exclusive
    2,345.00
    Save 70.35 (3%)
    A3767804 ExpiSf™ CD Medium, 10 L Each
    1,268.65
    Online Exclusive
    1,308.00
    Save 39.35 (3%)
    A3767803 ExpiSf™ CD Medium, 6 x 1000 mL Each
    807.65
    Online Exclusive
    832.00
    Save 24.35 (3%)
    A3767802 ExpiSf™ CD Medium, 1000 mL Each
    151.65
    Online Exclusive
    159.00
    Save 7.35 (5%)
    4115-0125 125mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 24 Case of 24
    328.65
    Online Exclusive
    369.00
    Save 40.35 (11%)
    A3767801 ExpiSf™ CD Medium, 500 mL Each
    85.65
    Online Exclusive
    87.75
    Save 2.10 (2%)
    A38915 ExpiFectamine™ Sf Transfection Reagent Each
    675.65
    Online Exclusive
    716.00
    Save 40.35 (6%)
    31985062 Opti-MEM™ I Reduced Serum Medium, 100 mL Each
    31.65
    Online Exclusive
    34.34
    Save 2.69 (8%)
    A3767809 ExpiSf™ Protein Production Kit, 6 x 1000 mL (ExpiSf CD Medium), 4 x 1 mL (ExpiSf Enhancer) Each
    996.65
    Online Exclusive
    1,074.00
    Save 77.35 (7%)
    A3767808 ExpiSf™ Protein Production Kit, 5 x 10 L (ExpiSf CD Medium), 5 x 40 mL (ExpiSf Enhancer) Each
    6,468.65
    Online Exclusive
    6,780.00
    Save 311.35 (5%)
    A3767807 ExpiSf™ Protein Production Kit, 10 L (ExpiSf CD Medium), 40 mL (ExpiSf Enhancer) Each
    1,610.65
    Online Exclusive
    1,688.00
    Save 77.35 (5%)
    A3767806 ExpiSf™ Protein Production Kit, 1 L (ExpiSf CD Medium), 4 x 1 mL (ExpiSf Enhancer) Each
    193.65
    Online Exclusive
    203.00
    Save 9.35 (5%)
    10361012 MAX Efficiency™ DH10Bac Competent Cells Each
    709.65
    Online Exclusive
    762.00
    Save 52.35 (7%)
    A35243 ExpiSf9™ Cells Each
    985.65
    Online Exclusive
    1,054.00
    Save 68.35 (6%)
    10360014 Bac-to-Bac™ Vector Kit Each
    1,214.00
    We offer a full array of innovative products and services, including fast and flexible cell line development, media and feeds, class-setting technologies, quality custom media development, and media optimizing services.
    Learn more about how we can customize your media formulation and format
    Cloning products
    Catalog # Name Size Price (USD) Qty
    11593027 Ampicillin, sodium salt, irradiated Each
    50.65
    Online Exclusive
    52.50
    Save 1.85 (4%)
    15750060 Gentamicin (50 mg/mL), 10 mL Each
    91.75
    C404003 One Shot™ TOP10 Chemically Competent E. coli, 21 x 50 μL Each
    502.65
    Online Exclusive
    532.00
    Save 29.35 (6%)
    15160054 Kanamycin Sulfate, 100 mL Each
    73.65
    Online Exclusive
    77.75
    Save 4.10 (5%)
    12532016 Platinum™ PCR SuperMix High Fidelity, 100 rxns Each
    403.65
    Online Exclusive
    429.00
    Save 25.35 (6%)
    15519028 Bluo-Gal Each
    533.65
    Online Exclusive
    564.00
    Save 30.35 (5%)
    15529019 IPTG Each
    92.65
    Online Exclusive
    98.00
    Save 5.35 (5%)
    Bac-to-Bac associated products
    Plasmid purification products
    Catalog # Name Size Price (USD) Qty
    K210004 PureLink™ HiPure Plasmid Midiprep Kit, 25 Preps Each
    352.65
    Online Exclusive
    366.00
    Save 13.35 (4%)
    K210008XP PureLink™ HiPure Expi Plasmid Megaprep Kit Each
    395.65
    Online Exclusive
    415.00
    Save 19.35 (5%)
    K210009XP PureLink™ HiPure Expi Plasmid Gigaprep Kit Each
    299.65
    Online Exclusive
    328.00
    Save 28.35 (9%)
    K210006 PureLink™ HiPure Plasmid Maxiprep Kit, 10 Preps Each
    313.65
    Online Exclusive
    329.00
    Save 15.35 (5%)
    Protein extraction and quantification products
    Catalog # Name Size Price (USD) Qty
    89802 I-PER™ Insect Cell Protein Extraction Reagent Each
    322.65
    Online Exclusive
    346.00
    Save 23.35 (7%)
    A37989 Pierce™ Protease Inhibitor XL Capsules, EDTA-free Each
    1,034.65
    Online Exclusive
    1,116.00
    Save 81.35 (7%)
    78437 Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X), 5 mL Each
    435.65
    Online Exclusive
    492.00
    Save 56.35 (11%)
    A32965 Pierce™ Protease Inhibitor Tablets, EDTA-free, 20 Tablets Each
    340.65
    Online Exclusive
    393.00
    Save 52.35 (13%)
    Protein purification products
    Catalog # Name Size Price (USD) Qty
    A36797 Pierce™ Anti-DYKDDDDK Magnetic Agarose, 4 mL Each
    482.65
    Online Exclusive
    538.00
    Save 55.35 (10%)
    1245906 POROS™ 50 PI Weak Anion Exchange Resin, 50 mL Each
    562.00
    A36801 Pierce™ Anti-DYKDDDDK Affinity Resin, 2 mL Each
    581.65
    Online Exclusive
    648.00
    Save 66.35 (10%)
    25215 HisPur™ Ni-NTA Superflow Agarose, 50 mL Each
    600.65
    Online Exclusive
    702.00
    Save 101.35 (14%)
    A36739 POROS™ CaptureSelect™ AAVX Affinity Resin, 10 mL Each
    1,854.00
    25236 Pierce™ Glutathione Superflow Agarose, 10 mL Each
    267.65
    Online Exclusive
    328.00
    Save 60.35 (18%)
    A50587 Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible, 500 mL Each
    5,046.65
    Online Exclusive
    5,620.00
    Save 573.35 (10%)
    A50584 Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible, 10 mL Each
    160.65
    Online Exclusive
    179.00
    Save 18.35 (10%)
    A50585 Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible, 50 mL Each
    644.65
    Online Exclusive
    724.00
    Save 79.35 (11%)
    78606 Pierce™ Ni-NTA Magnetic Agarose Beads, 20 mL Each
    1,099.65
    Online Exclusive
    1,222.00
    Save 122.35 (10%)
    A50586 Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible, 250 mL Each
    2,813.65
    Online Exclusive
    3,130.00
    Save 316.35 (10%)
    1335906 POROS™ 50 HS Strong Cation Exchange Resin, 50 mL Each
    564.00
    88222 HisPur™ Ni-NTA Resin, 100 mL Each
    927.65
    Online Exclusive
    1,062.00
    Save 134.35 (13%)
    82073 POROS™ XQ Strong Anion Exchange Resin, 10 mL Each
    255.00
    26181 Pierce™ Anti-HA Agarose, 1 mL Each
    686.65
    Online Exclusive
    756.00
    Save 69.35 (9%)
    Catalog # Name Size Price (USD) Qty
    4115-0500 500mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 12 Case of 12
    251.65
    Online Exclusive
    271.00
    Save 19.35 (7%)
    4115-0125 125mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 24 Case of 24
    328.65
    Online Exclusive
    369.00
    Save 40.35 (11%)
    4115-2800 2800mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 4 Case of 4
    259.65
    Online Exclusive
    277.00
    Save 17.35 (6%)
    4115-1000 1000mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 6 Case of 6
    163.65
    Online Exclusive
    181.00
    Save 17.35 (10%)
    4115-2000 2000mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 4 Case of 4
    125.65
    Online Exclusive
    148.00
    Save 22.35 (15%)
    4115-0250 250mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 12 Case of 12
    191.65
    Online Exclusive
    216.00
    Save 24.35 (11%)
    Try the Gibco MediaSelect Tool
    Easily find a media solution suited to your specific application. Use this interactive tool to select classical basal, protein expression, or viral vector media.
    Your selections will allow you to choose from a catalog menu of media – or customize your own formulation to fit your experimental setup and budget. Where available, you can compare media formulations side-by-side.
    Find your media with the Gibco MediaSelect Tool
    For Research Use Only. Not for use in diagnostic procedures.