Overview
The Invitrogen iBright Imaging Systems offer multiple powerful features to help make imaging and analyzing western blots and gels easy. The high-resolution 9.1 MP camera and suite of automated features helps you produce publication quality data fast. The touchscreen interface is carefully crafted to provide a smooth image capture experience. The system’s built in software and our standalone iBright Analysis Software are designed to streamline the image analysis process. Straightforward total lane protein and house-keeping protein normalization workflows allow for simpler image analysis from start to finish, no matter what your experience level. Maximize your investment with specialty applications and 21CFR part 11 compliance support. Explore more below to find out which iBright Imaging System is right for you.  
Video Player is loading.
Current Time 0:00
Duration 1:33
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:33
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Highlighted Features
    Convenient, intuitive interface and workflows
    Clear interface layout combined with a 12.1 inch capacitive touchscreen balances powerful functionality with practical ease of use.
    Powerful camera technology
    Capture crisp, clear images with a 9.1MP camera. Smart Exposure technology rapidly determines optimal exposure time, minimizing the need to repeat exposures to achieve the desired signal. For samples containing targets with widely varying expression levels, leveraging the Smart Range HDR (high dynamic range) feature can help extend the linear dynamic range of the data.
    Essential imaging modes and specialty applications
    Five imaging modes let you efficiently and easily capture data from protein gels, nucleic acid gels, chemiluminescent western blots, and fluorescent western blots. Specialty applications help enhance your investment.
    Fluorescent multiplexing for more data from your sample
    Multiplex and capture up to four proteins in a single blot for more meaningful and representative experiments.
    Streamlined image analysis
    Perform data analysis with on-instrument software or our iBright Analysis Software, available in desktop and cloud-based formats. Analysis capabilities include absolute and relative quantitation, molecular weight estimation normalization, and more.
    21 CFR Part 11 Compliance Support
    Optional package offers seamless integration of security, audit, and e-signature workflows with iBright Imaging Systems’ overall user-friendly imaging experience.