Overview
There are three models in the iBright Imaging Systems family: The iBright CL750 Imaging System, the iBright CL1500 Imaging System, and the iBright FL1500 Imaging System. The iBright CL750 Imaging System offers the core essential western blot and gel imaging functions and makes the transition from the darkroom and film easy. The iBright CL1500 Imaging System expands application support and has many of the high-performance specifications of our premier iBright FL1500 model. The iBright FL1500 Imaging System features maximum application support, including fluorescence western blot imaging with up to four fluorescence channels at a time.
Specifications
iBright CL750 Imaging System
iBright CL1500 Imaging System
iBright FL1500 Imaging System
Core essential western blot and gel imaging-transition from darkroom and film-based detection with ease
Expanded application support with many of the same high-performance specifications as the iBright FL1500 imager
Maximum application support including up to 4-channel multiplex western blot imaging
Core imaging applications
Colorimetric stained protein gels
Fluorescent stained protein gels
Fluorescent stained nucleic acid gels
Colorimetric stained membranes
Limited*
Chemiluminescent western blots
Colorimetric western blots
Fluorescent western blots
Specialty plate-based imaging applications
Fluorescent colonies (ex. GFP expressing)
Visible colonies (ex. crystal violet stained)
Qualitative visible imaging applications**
Opaque objects (e.g., 2D strips, TLC plates, leaf sections)
Camera
Detector Cooled 16-bit CCD, 65,535 shades of gray
Resolution 9.1 megapixels
Lens Fixed, 25 mm, f/0.95
Field of view 22.5 x 18.0 cm (W x D) (image up to 4 mini blots or gels)
Binning modes 1x1, 2x2, 4x4 (high-resolution/sensitivity settings)
1x1, 2x2, 3x3, 4x4, 5x5, 6x6, 8x8 (maximum flexibility for adjusting resolution and sensitivity based on needs)
Zoom 1–2x, digital (digital zoom reduces the effective resolution of the zoomed image) 1–8x (1–2x mechanical + 1–4x digital) (Mechanical zoom maximizes sensitivity by moving the camera closer to the sample stage, reducing focal length)
System interface
Touchscreen 12.1 inch capacitive LCD display, 1,024 x 768 pixels
Storage and connectivity
USB 2 USB (2.0)
Network Ethernet port, Connect (cloud-based) connectivity, optional Wi-Fi (adapter sold separately)
Image file format G2i (proprietary), TIFF, JPG, PNG
Hard drive 64 GB SSD 256 GB SSD
System Hardware
Sample drawer and stage Manually operated drawer with fixed stage
Automatic open/close drawer with automatic rotating stage (up to 10°)
Filter set 2 filters (0 excitation, 2 emission)
4 filters (2 excitation, 2 emission)
12 filter (6 excitation, 6 emission)
Illumination source
  • Green LED (470–550 nm) transilluminator
  • Green LED (470–550 nm) transilluminator
  • Epi white LED
  • Green LED (470–550 nm) transilluminator 
  • Epi white LED
  • Epi near-IR LED
System software
Automated features and algorithms
  • Automatic zoom
  • Automatic focus
  • Automatic exposure (Smart Exposure)
  • Automatic on-board image analysis
  • High dynamic range (HDR) image capture functionality (Smart Range HDR***)
Stand-alone analysis applications
iBright Analysis Software: desktop version (macOS™ or Windows™ operating systems) and Connect (cloud-based) version
iBright Analysis Software—Secure: (macOS™ or Windows™ operating systems) for 21 CFR part 11 compliance support
Shipping, warranty, and upgrades
Dimensions
(L x W x H): 68 x 38 x 60 cm
Weight
Approximately 47 kg (105 lb)
Approximately 50 kg (110 lbs)
Warranty 1 year from date of purchase
2 years from date of purchase
Upgradability No
Yes, to FL1500 imager
No.
* Membranes must be imaged when wet, using Thermo Scientific Pierce Reversible Protein Stain Kit for Nitrocellulose Membranes or Thermo Scientific Pierce Reversible Protein Stain Kit for PVDF Membranes.
** Applications provide a qualitative visualization of the object or confirmation of the presence of signal. Not recommended for quantitation.
*** Smart Range HDR available in chemiluminescent blot mode only.