Showcase Ion Channel Biology
Screen compounds that modulate ligand- and voltage-gated ion channels with our collection of antibodies,  immunoassays and cell-based assays to monitor potassium, chloride, calcium and membrane potentials.
Ion Channel Antibodies & Immunoassays
Find the right antibody you need for ion channel research. 
Cell-Based Ion Channel Assays
Perform functional studies for ion channels with our cell-based assays including the FluxOR™ assay for potassium,  the Premo™ Halide Sensor for chloride, Fluo-4 Direct™ for calcium and Voltage Sensor Probes for monitoring membrane potential changes.
For Research Use Only. Not for use in diagnostic procedures.