Search Thermo Fisher Scientific
We are constantly advancing CBRNE and bomb detection technology to protect against the threat of radiation and radioactive material with real-time detection and immediate results. From mobile detection devices to mobile and portal detection monitors, we have a solution for any scenario. Rely on us for:
Grants are available for radiation detection products and FREE assistance is available.
Thermo Scientific RadEye Spectroscopic Radiation Detectors are ideal for contamination monitoring in radiation controlled areas, contamination check points, and waste and material shipping surveys.
Scott Masiella, Global Safety & Security Market Manager, Radiation Measurement and Security Instruments, Thermo Fisher Scientifc, discusses this topic in Police One magazine.
American Security Today Guest Editorial by Scott Masiella, Global Safety & Security Market Manager, Radiation Measurement and Security Instruments, Thermo Fisher Scientifc.
Scott Masiella, Global Safety & Security Market Manager, Radiation Measurement and Security Instruments, talks to Homeland Security Today about radiological risks at hospitals and medical facilities.
Large sporting events are high-risk areas for malicious radiation threats. Learn how to secure events in your city.
With long routes through heavily populated areas, parades and races offer a challenging security scenario.
Ensure the safety of your loading dock by scanning arriving packages for potentially harmful sources of radiation.
Configurable services to help you optimize the performance of your radiation equipment. Select from a wide variety of service products to maximize the productivity of your assets while managing the high cost of unplanned maintenance and repair.
The natural background rejection (NBR) measurement method has been developed by Thermo Fisher Scientific for extremely fast discrimination between natural and artificial gamma radiation and is deployed on all Thermo Scientific™ radiation monitors. Using an instrument with NBR allows you to keep your alarm threshold extremely low without the need to worry about false alarms from non-threatening sources.
Frequently Asked Questions about Radiation Detection
Radiation can be found almost anywhere. Is it always a threat to public health and safety? How can it be monitored? What tools do I need to detect it? Are you trying to scare us? Get the answers to these and other questions on our Frequently Asked Questions about Radiation Detectionpage.