Search Thermo Fisher Scientific
Explore our protein gel options. Choose from precast polyacrylamide gel electrophoresis (PAGE) chemistries designed for specific applications including broad range, high, or low molecular weight separations, SDS-PAGE, native PAGE, or IEF. Mini gels and wider, higher throughput, midi gels are offered in a variety of well and cassette formats. Or select a system for pouring and casting your own gels.
Find the right gel for your protein
Convert from other suppliers' gels
Protein gels performance guarantee—Invitrogen precast protein gels are designed and tested to deliver superior performance, reliability, reproducibility, and consistency of quality. We guarantee it. Check out our Certificate of Analysis. No other supplier conducts such stringent quality testing.
Get started with our money-saving Protein Gel Welcome Packs
Restock with Protein Gel Resupply Packs
![]() Bis-Tris Gels | ![]() Tris-Glycine Gels | ![]() Tris-Acetate Gels | ![]() Tricine Gels | |
Available precast gels | NuPAGE Bis-Tris Gels, Bolt Bis-Tris Plus Gels | Novex Tris-Glycine | NuPAGE Tris-Acetate | Novex Tricine |
Protein sample type | Broad range MW (6–400 kDa) | Broad range MW (6–400 kDa) | High range MW (40–500 kDa) | Low range MW (2.5–40 kDa) |
Product benefits | Neutral pH helps prevent protein degradation for high sensitivity. Load up to 60 µL per well with Bolt gels. Fast 20–35 minute run times. Room temperature storage for up to 16 months. Two buffer options: Choose MOPS buffer for higher range separations or MES buffer for lower range separations. | Traditional Laemmli-style gels with larger loading capacity—load up to 60 µL per well. | Neutral pH for better separation, transfer, and analysis of high molecular weight proteins. | Better separation, transfer, and analysis of low molecular weight proteins. |
Recommended for | Western blotting, in-gel staining, mass spectrometry, post translationally modified proteins, dilute samples, and denaturing applications. | Western blotting, in-gel staining, samples containing detergent and high salt, and denaturing or native PAGE applications. | High molecular weight proteins, Western blotting, mass spectrometry, post translationally modified proteins, and denaturing or native PAGE applications. | Low molecular weight proteins, western blotting, in-gel staining, and denaturing applications. |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
Bolt Bis-Tris Plus |
| 8% | 6 to 400 kDa |
| 22–45 min | 16 months at room temperature |
NuPAGE Bis-Tris |
| 8% | 1.5 to 300 kDa |
| 35–50 min | 16 months at room temperature |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
|
| 6% 8% 10% 12% 4–20% 4–12% 14% 16% | 8 to 250 kDa |
| 90 min | 12 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
NuPAGE Tris-Acetate |
| 7% 3–8% | 36 to 500 kDa |
| 60 min | 8 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
Novex Tricine |
| 10% 16% | 2 to 200 kDa |
| 90 min | 1–2 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
| 3–12% 4–16% | 15 to 10,000 kDa |
| 90–120 min | 12 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
IEF |
| pH 3–7 pH 3–10 | pI 3.5 to pI 8.5 |
| 150 min | 2 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
Zymogram |
| 10% (with gelatin) | 10 to 220 kDa |
| 90 min | 2 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
E-PAGE |
| E-PAGE 96: 6% E-PAGE 48: 8% | 10 to 220 kDa |
| 14–25 min | 6 months |
![]() NativePAGE Bis-Tris Gels View all | ![]() Isoelectric Focusing Gels View all | ![]() Novex Zymogram Gels View all | ![]() E-PAGE Precast Gels View all | |
Gel chemistry | Bis-Tris | pH 3–7, pH 3–10 | Tris-glycine with gelatin | 8% or 6% |
Protein sample type | 15–10,000 kDa | pI 3.5–8.5 | 10–220 kDa | 10–200 kDa |
Product benefits | Separation of native proteins, neutral pH for high sensitivity applications | Separation of proteins based on isoelectric point | Separation of proteins based on size and activity | High-throughput gel system, that eliminates the need for buffers |
![]() SureCast Handcast System Explore | ![]() Empty Gel Cassettes Explore | ![]() Bolt Empty Cassettes Explore | |
Cassette type | Reusable, glass | Single use, pre-sealed | Single use, pre-sealed |
Product benefits |
|
|
|
Gel dimensions | Mini: 8x8 cm (1 mm thick) | Mini: 8x8 cm (1 or 1.5 mm thick) Midi: 8x13 cm (1 mm thick) | Mini: 8x8 cm (1 mm thick) |
Gel tank | Mini Gel Tank, Xcell SureLock Mini-Cell | Mini Gels: Mini Gel Tank or XCell SureLock Mini-Cell Midi Gels: XCell4 SureLock Midi-Cell or Bio-Rad Criterion (with adapters only) | Mini Gel Tank, Xcell SureLock Mini-Cell |
Reagents | SureCast reagents, other popular polyacrylamide reagents | SureCast reagents, other popular polyacrylamide reagents | SureCast reagents, other popular polyacrylamide reagents |
Combos available | 1-, 2-, 10-, 12-, and 15-wells | 1-, 2-, 5-, 10-, 12-, 15-, and 2D-wells | 10- or 12-wells (high capacity wells) |
![]() Bis-Tris Gels | ![]() Tris-Glycine Gels | ![]() Tris-Acetate Gels | ![]() Tricine Gels | |
Available precast gels | NuPAGE Bis-Tris Gels, Bolt Bis-Tris Plus Gels | Novex Tris-Glycine | NuPAGE Tris-Acetate | Novex Tricine |
Protein sample type | Broad range MW (6–400 kDa) | Broad range MW (6–400 kDa) | High range MW (40–500 kDa) | Low range MW (2.5–40 kDa) |
Product benefits | Neutral pH helps prevent protein degradation for high sensitivity. Load up to 60 µL per well with Bolt gels. Fast 20–35 minute run times. Room temperature storage for up to 16 months. Two buffer options: Choose MOPS buffer for higher range separations or MES buffer for lower range separations. | Traditional Laemmli-style gels with larger loading capacity—load up to 60 µL per well. | Neutral pH for better separation, transfer, and analysis of high molecular weight proteins. | Better separation, transfer, and analysis of low molecular weight proteins. |
Recommended for | Western blotting, in-gel staining, mass spectrometry, post translationally modified proteins, dilute samples, and denaturing applications. | Western blotting, in-gel staining, samples containing detergent and high salt, and denaturing or native PAGE applications. | High molecular weight proteins, Western blotting, mass spectrometry, post translationally modified proteins, and denaturing or native PAGE applications. | Low molecular weight proteins, western blotting, in-gel staining, and denaturing applications. |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
Bolt Bis-Tris Plus |
| 8% | 6 to 400 kDa |
| 22–45 min | 16 months at room temperature |
NuPAGE Bis-Tris |
| 8% | 1.5 to 300 kDa |
| 35–50 min | 16 months at room temperature |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
|
| 6% 8% 10% 12% 4–20% 4–12% 14% 16% | 8 to 250 kDa |
| 90 min | 12 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
NuPAGE Tris-Acetate |
| 7% 3–8% | 36 to 500 kDa |
| 60 min | 8 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
Novex Tricine |
| 10% 16% | 2 to 200 kDa |
| 90 min | 1–2 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
| 3–12% 4–16% | 15 to 10,000 kDa |
| 90–120 min | 12 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
IEF |
| pH 3–7 pH 3–10 | pI 3.5 to pI 8.5 |
| 150 min | 2 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
Zymogram |
| 10% (with gelatin) | 10 to 220 kDa |
| 90 min | 2 months |
Precast gels | When to use | Gel % available | Separation range | Formats available | Average run time | Shelf life |
---|---|---|---|---|---|---|
E-PAGE |
| E-PAGE 96: 6% E-PAGE 48: 8% | 10 to 220 kDa |
| 14–25 min | 6 months |
![]() NativePAGE Bis-Tris Gels View all | ![]() Isoelectric Focusing Gels View all | ![]() Novex Zymogram Gels View all | ![]() E-PAGE Precast Gels View all | |
Gel chemistry | Bis-Tris | pH 3–7, pH 3–10 | Tris-glycine with gelatin | 8% or 6% |
Protein sample type | 15–10,000 kDa | pI 3.5–8.5 | 10–220 kDa | 10–200 kDa |
Product benefits | Separation of native proteins, neutral pH for high sensitivity applications | Separation of proteins based on isoelectric point | Separation of proteins based on size and activity | High-throughput gel system, that eliminates the need for buffers |
![]() SureCast Handcast System Explore | ![]() Empty Gel Cassettes Explore | ![]() Bolt Empty Cassettes Explore | |
Cassette type | Reusable, glass | Single use, pre-sealed | Single use, pre-sealed |
Product benefits |
|
|
|
Gel dimensions | Mini: 8x8 cm (1 mm thick) | Mini: 8x8 cm (1 or 1.5 mm thick) Midi: 8x13 cm (1 mm thick) | Mini: 8x8 cm (1 mm thick) |
Gel tank | Mini Gel Tank, Xcell SureLock Mini-Cell | Mini Gels: Mini Gel Tank or XCell SureLock Mini-Cell Midi Gels: XCell4 SureLock Midi-Cell or Bio-Rad Criterion (with adapters only) | Mini Gel Tank, Xcell SureLock Mini-Cell |
Reagents | SureCast reagents, other popular polyacrylamide reagents | SureCast reagents, other popular polyacrylamide reagents | SureCast reagents, other popular polyacrylamide reagents |
Combos available | 1-, 2-, 10-, 12-, and 15-wells | 1-, 2-, 5-, 10-, 12-, 15-, and 2D-wells | 10- or 12-wells (high capacity wells) |
All Invitrogen precast protein gels are high quality polyacrylamide gels designed to deliver superior performance, reliability, reproducibility, and consistency of quality. Our extensive portfolio of precast protein gels provides a wide variety of gel options available for your research. Whether your protein is 2.5 kDa or as large as 500 kDa, or you’re looking for separation under native conditions or by isoelectric point—we have the right gel for your application.
Our precast protein polyacrylamide gels are offered in four different chemistries. The choice of whether to use one chemistry or another depends on the abundance of the protein you’re separating, the size of the protein, and your downstream application.
For denaturing gel electrophoresis or SDS PAGE there are many options available based on the application, and size of the protein of interest. For separation of a broad range of proteins, two chemistries, Bis-Tris and Tris-glycine, have been optimized for performance and long shelf life. Choose Bis-Tris gel chemistry when you have a low abundance of protein or when your downstream applications require high protein integrity, such as posttranslational modification analysis, mass spectrometry, or sequencing. Bis-Tris chemistry provides a neutral (pH 7.0) environment during electrophoresis, which may result in better sample integrity and stability of the gel. This helps reduce protein modifications and helps produce sharp band resolution. Bis-Tris gel chemistry is offered in our Bolt Bis-Tris Plus and NuPAGE Bis-Tris gels. Bolt Bis-Tris Plus mini gels build on the legacy of the highly published NuPAGE Bis-Tris gels, feature larger capacity wells- for loading up to 60 µl per well, and can be run in as little as 20 minutes.
To separate high-abundance proteins, select our robust Novex Tris-Glycine gel chemistry, which offers maximum protein separation performance and crisp, straight bands.
For separation of high molecular weight proteins, Tris-acetate gel chemistry is recommended, offered in NuPAGE Tris-Acetate gels.
For separation of low molecular weight proteins, use Novex Tricine gels which provide increased resolution of proteins with molecular weights as low as 2.5 kDa.
For separation of proteins under nondenaturing conditions or native page, three gel systems are recommended- NativePAGE Bis-Tris, Novex Tris-Glycine and NuPAGE Tris-Acetate. NativePAGE Bis-Tris gels, are based on the blue native polyacrylamide gel electrophoresis (BN-PAGE) which helps overcome the limitations of traditional native electrophoresis by providing a near-neutral operating pH and detergent compatibility. Both Novex Tris-Glycine and NuPAGE Tris-Acetate gels can be run under native and non-reducing conditions for separation of proteins by net charge, size and shape.
For separation of proteins by isoelectroctric point (pI), use Novex IEF gels. Novex IEF gels effectively create a pH gradient to separate proteins according to their unique pI.
This webinar will highlight the different protein modifications occurring during electrophoresis, the importance of pH in sample integrity, how to choose the right gel chemistry for specific research applications, and how to introduce the right new gel chemistry in your research workflow.
This video will show you how to separate proteins by using Invitrogen Mini Gel Tank and Invitrogen precast SDS-PAGE gels.
Find the exact gel and buffers needed for your electrophoresis experiments.
Find the right gel for your research needs based on molecular weight, downstream applications, and throughput requirements.
Learn how the right protein gel chemistry can help you achieve better protein separation and western blots.
For Research Use Only. Not for use in diagnostic procedures.