PSC Characterization Tools
Verifying the quality of your pluripotent stem cells (PSCs) is critical to moving your research goals forward. We have a variety of cellular and molecular methods to help you completely and cost-effectively characterize your PSCs. From the PluriTest compatible PrimeView Global Gene Expression Profile Assays, which provides quick verification of pluripotency, to TaqMan hPSC Scorecard Panel, which confirms trilineage differentiation potential, we have the tools you need to characterize with lines with confidence.
Request information
NEW  Review our stem cell services flyer for our characterization and pluripotency services today
Cellular and molecular PSC characterization methods
 Alkaline Phosphatase Live StainAntibody staining PluriTest-Compatible PrimeView Global Gene Expression Profile AssaysTaqMan hPSC Scorecard PanelKaryoStat and KaryoStat HD Assays
Product descriptionEasy identification of pluripotency without compromising cell integritySpecific and flexible identification of PSCsCost-effective global confirmation of pluripotency marker expressionEvaluates pluripotency and confirms trilineage differentiation potentialAlternative to g-banding karyotyping; offers whole-genome coverage for accurate detection of chromosomal abnormalities
How specific are the results?Low (stains stem and progenitor cells)Medium (stains human ES and iPS cells)High (whole-transcriptome gene expression profile)High (profiles expression of both PSC markers and early germ layer factors)High
Will the cells remain viable?YesNo [1]NoNoNo
How long before I see results?Stain PSCs typically in 20 minutes or lessStain PSCs typically in 90–120 minutes2 days6–8 hours3–4 days
Are data analysis tools included?NoNoYes, free online PluriTest analysis toolYes, free cloud-based softwareYes, free downloadable Chromosome Analysis Suite (ChAS) software
Is a reference standard included?NoNoYes [2]Yes [2]No
Are EVOS FLoid Cell Imager protocols available?YesYesNoN/ANo
Training and expertise requiredMinimalMinimalModerateModerateModerate
Unit size500 μL vial sufficient for staining twelve 6 cm dishes100 μg30 arrays (one sample/array) or 1, 16-sample array plate1, 384-well plate Kit (4 samples/plate) or 2, 96-well plates (1 sample/plate)24 arrays (one sample/array)
 OrderOrderOrderOrderLearn more
1. Non-destructive when used with antibodies qualified for live staining.; 2. Reference standard is included in FREE software.
PSC characterization services
With access to a complete portfolio of characterization tools and cutting-edge instruments our CellModel Services team can help.
Learn more
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.