Penicillium chrysogenum Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Penicillium is among the top three most common indoor airborne fungi (along with Aspergillus and Cladosporium).1,2 These same three molds as well as Alternaria alternata are the species most likely to cause allergy symptoms, which typically occur after allergic individuals inhale mold spores.3,4 Perhaps most familiar, however, is the Penicillium chrysogenum species, which is typically blue-green in color and can be found both indoors and outside.2
One of six species most commonly found in water-damaged environments, this mold is also a source of antibiotics such as penicillin.1,5 Closely associated with asthma, P. chrysogenum is well known as both an allergen and a pathogen (i.e., something that causes disease), and particularly in those with weakened immune systems due to an existing illness, it can trigger a variety of infections.5 Penicillium spores can be detected in the air throughout the year, but they can also show seasonal fluctuations. For example, in the United Kingdom, spores reach their peak in autumn, with highest levels during daytime hours.6
Penicillium chrysogenum is widespread in temperate and subtropical regions and is present practically anywhere organic material is available.4,5 Penicillium can be found outdoors in mediums such as soil, decaying plant debris, compost, grains, and rotting fruit.4 However, mold reproduces via spores, which can be transported by air, water, and insects.7,11 So even if a fungus originates outdoors, it often can enter your home through a variety of means, including doorways, windows, vents, and heating and air conditioning systems. Plus, it can hitch a ride inside via clothing, shoes, and pets.12 Therefore, Penicillium is also found indoors, where it usually grows on surfaces such as drywall, wood, painted materials, wallpaper, carpet, paint, fabrics, dust, and various other household contents.13,14 Plus, it sometimes colonizes leather products such as shoes, jackets, belts, and sofas. Penicillium chrysogenum can also develop on foods such as fruits and vegetables, cereal grains, cured meat, margarine, cheese, and other dairy products.5
Some people with Penicillium chrysogenum allergy may also experience symptoms when exposed to other types of mold. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.2
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
If you are allergic to mold, your healthcare provider may recommend a plan that includes the following.3,7,16,17
Your healthcare provider may direct you to take one of the following medications to improve your allergy symptoms:
Mold allergy symptoms can range from mild to severe and vary person to person.3 Reactions can happen almost immediately after exposure, or they can be delayed. Symptoms are most common in mid-summer to early fall, but since molds grow both indoors and out, allergic reactions can occur all year.7
Symptoms typically include one or more of the following:3,8
Mold sensitization is also a major risk factor for developing upper and lower respiratory diseases such as allergic rhinitis (aka hay fever).4,9 In addition, other reactions to Penicillium can include hypersensitivity pneumonitis (also known as extrinsic allergic alveolitis), which can lead to a chronic disease with irreversible lung damage.9
A small percentage of asthmatics with inhaled mold allergy can also develop allergic urticaria (aka hives) when they eat or drink anything containing yeast or mold.10 In addition to causing allergic reactions, molds can also lead to infections along with toxic reactions.1
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are allergic to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
There are no credible reports in medical literature documenting indoor exposure to molds as a cause of anaphylaxis.15 However, exposure and sensitization to fungal allergens can promote the development and worsening of allergic diseases such as allergic rhinitis and asthma.2