Timothy Grass Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
The most abundant airborne grass pollen comes from tall meadow grasses such as Timothy grass, which is prevalent in temperate climates.1,2 With aliases that include common cat's tail, herd grass, meadow cat's tail, and herd's grass, Timothy grass is a short-lived perennial that grows up to approximately 150 centimeters (5 feet) tall and features flat leaves with a dense spike-like flower cluster.3,4 Releasing its wind-borne pollen from spring to fall, Timothy grass (Phleum genus) is among the worst grass genera that cause hay fever in early summer.4,5 This species is second only to orchard grass in allergenicity among grasses, making it severely allergenic.4
Timothy grass is widely distributed in temperate climates, such as those in Europe, Asia, North America, and South America, where it prefers meadows, pastures, parkland, and roadsides.2,5,8
Many patients with Timothy grass allergy can experience symptoms when exposed to other allergens such as tree, weed, or grass pollens, making it difficult to determine which pollen is causing the symptoms, especially when pollen seasons are overlapping. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.9 Other respiratory allergens that may cause reactions associated with Timothy grass are other grasses, including wheat, along with tree and weed pollens.9
If you experience an itchy mouth or throat after eating fresh fruit or raw vegetables, you may suffer from Oral Allergy Syndrome (OAS), sometimes called Pollen Food Allergy Syndrome (PFAS). This condition is caused by your immune system's reaction to similar proteins, or components, found in different allergens. It is quite common, with up to 25 percent of children with allergic rhinitis (i.e., hay fever) also suffering from OAS.10 Common plant foods involved in OAS for Timothy grass include melon, watermelon, citrus, banana, pineapple, persimmon, zucchini, tomato, hazelnut, peanut, and many more.9
Knowing the proteins, or components, within each allergen that are triggering your symptoms can help guide your management plan. With that in mind, and based on your symptom history, your healthcare provider may suggest something called a specific IgE component test, which can help reveal other pollens and foods you may react to. Results from this test can also help your healthcare provider decide if allergen immunotherapy may reduce your symptoms.9
Already have your specific IgE component test results?
Your component test results will include the name of the components (a series of letters and numbers). Your healthcare provider will likely review the results with you, but here you'll find an at-a-glance breakdown you can use as a reference. Simply match the component names to the list below to see what they mean in terms of symptom management.9
rPhl p 1, rPhl p 5b
rPhl p 7
rPhl p 12
MUXF3 (CCD)
Test results should be interpreted by your healthcare provider in the context of your clinical history. Final diagnosis and decision on further management is made by your healthcare provider.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
The management of allergic rhinitis includes avoidance of relevant allergens, symptomatic treatment, and allergen immunotherapy.10,12,13
Timothy grass allergy symptoms can be similar to many other pollen allergies and may include:6
If you're sensitized to Timothy grass and have asthma, grass pollen may trigger or worsen asthma symptoms, such as coughing and wheezing.6
While symptoms are usually limited to the throat, nose, and eyes, some people who are severely allergic to grass may also develop hives (urticaria) upon contact with its pollen.7
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are sensitized to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Timothy grass pollinates from spring to fall.4
Responses close to anaphylaxis have occurred in the most dangerous reaction cases.5 Plus, anaphylaxis has been reported in cases where abraded skin contacted allergenic grass.10