Dust Mite Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Dust mites are an important cause of allergies worldwide, and those allergic to them may experience symptoms after inhaling dust mite particles.1 Estimates indicate that 10 percent of the world's population is allergic to dust mites, but up to 90 percent of people with allergic asthma in some regions are sensitized to them.2 And for those exposed to high levels of these allergens, the risk of developing asthma increases roughly five fold.3 Along with inhaling dust mite particles, eating foods made with mite contaminated wheat flour can also cause reactions, including anaphylaxis.1
Dust mites are a type of arachnid, much like spiders and ticks; however, unlike their relatives, mites don't bite humans.2,4 Instead, these tiny blind creatures dine on microscopic skin scales and fungi.3 And given their size, a single gram of dust can contain up to 19,000 mites.2 But it's not so much the mites themselves that are the problem. Rather, it's their excrement, as 95 percent of dust-mite allergens are a result of mite fecal particles.1 The most recognized house dust mites species are Dermatophagoides pteronyssinus, Dermatophagoides farinae, and Blomia tropicalis.4
Optimal temperatures for dust mite growth are roughly 18 to 27 C (approximately 65 to 80 F).5 Additionally, dust mites require a humidity level of at least 40 percent to survive.2 So temperature and humidity levels can significantly impact dust mite populations.3 Additionally, dust mite allergens typically increase in the summer when humidity levels are higher, and they remain elevated through the winter before dropping off late winter and spring.6
While dust mites are almost nonexistent in homes in arid climates, they thrive in warm humid environments, where they can cause year-round symptoms.2,6,10 In fact, roughly 84 percent of U.S. homes have detectable levels of dust mites, and older homes and those whose occupants have lower income levels generally have higher dust mite concentrations.9 Within dwellings, mites thrive in mediums such as mattresses, pillows, bedding, stuffed animals, upholstered furniture, and carpet.3,10
Many patients with dust mite allergy can experience symptoms when exposed to other allergens such as different species of dust mites, cockroaches, shellfish, and helminths (e.g., Ascaris lumbricoides). This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.4
Knowing the proteins, or components, within each allergen that are triggering your symptoms can help guide your management plan. With that in mind, and based on your symptom history, your healthcare provider may suggest something called a specific IgE component test, which can help reveal your specific risk profile concerning dust mite. Results from this test can also help your healthcare provider decide if allergen immunotherapy may reduce your symptoms.4
Already have your specific IgE component test results?
Your component test results will include the name of the components (a series of letters and numbers). Your healthcare provider will likely review the results with you, but here you'll find an at-a-glance breakdown you can use as a reference. Simply match the component names to the list below to see what they mean in terms of symptom management.4
rDer p 1, rDer p 2, rDer p 23
rDer p 10
Test results should be interpreted by your healthcare provider in the context of your clinical history. Final diagnosis and decision on further management is made by your healthcare provider.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
If you are allergic to dust mites, your healthcare provider may recommend a plan that includes the following.3,4,10-13
Dramatically decreasing exposure to dust mite allergens can help both related asthma and rhinitis symptoms.4
Your healthcare provider may direct you to employ one of the following therapies to improve your allergy symptoms:
If you're with someone who's having an allergic reaction and shows signs of shock, act fast. Look for pale, cool, and clammy skin; a weak, rapid pulse; trouble breathing; confusion; and loss of consciousness. Do the following immediately:
Symptoms of dust mite allergy can include:7
In addition, dust-mite allergy is an especially important risk factor for patients with rhinitis and asthma.4 In fact, if you have asthma, you may also experience symptoms such as:7
Oral mite anaphylaxis can occur when those allergic to dust mites consume mite contaminated wheat flour. The condition is most often triggered by pancakes and as such has been referred to as pancake syndrome. Other foods incriminated in this syndrome include sponge cakes, beignets, okonomiyaki (bonito and mackerel covered with flour), cornmeal cakes, polenta, pizzas, pastas, breads, parmigiana steaks (prepared with grated bread), and white sauces. Plus, mites can sometimes contaminate products stored at room temperature, such as cheese, ham, chorizo, and salami.8
Symptoms of pancake syndrome can include:8
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are allergic to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
In severe cases, dust mite allergy can be chronic, resulting in persistent sneezing, cough, congestion, facial pressure, or severe asthma attack.7 Most often, dust mite allergy symptoms are caused by inhaling dust mite particles. However, eating foods made with mite-contaminated wheat flour can also cause reactions, including anaphylaxis.1