Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
Antibody detects endogenous levels of total ADCY7.
This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate cyclase type 7; Adenylate cyclase type VII; Adenylyl cyclase 7; adenylyl cyclase type VII; ATP pyrophosphate-lyase 7; FLJ36387
Gene Aliases: AA407758; AC7; ADCY7; KIAA0037
UniProt ID: (Human) P51828, (Mouse) P51829
Entrez Gene ID: (Human) 113, (Rat) 84420, (Mouse) 11513
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support