Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Contact Us
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • C10orf99 Antibodies

            Invitrogen

            C10orf99 Polyclonal Antibody

            Advanced Verification
            2 References
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite C10orf99 Polyclonal Antibody

            • Antibody Testing Data (5)
            • Advanced Verification (1)
            Application
            C10orf99 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 6

            C10orf99 Antibody (PA5-62266) in IHC (P)

            Immunohistochemical analysis of C10orf99 in human pancreas using C10orf99 Polyclonal Antibody (Product # PA5-62266) shows no positivity in exocrine glandular cells as expected.
            View Product

            C10orf99 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            C10orf99 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            C10orf99 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            C10orf99 Antibody in Immunohistochemistry (IHC)
            C10orf99 Antibody in Immunohistochemistry (IHC)
            C10orf99 Antibody

            Product Details

            PA5-62266

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:200-1:500
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant protein corresponding to Human C10orf99. Recombinant protein control fragment (Product #RP-90605).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.20 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.2, with 40% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2638683

            Product Specific Information

            Immunogen sequence: KRRPAKAWSG RRTRLCCHRV PSPNSTNLKG HHVRLCKPCK LEPEPRLWVV PGALPQV

            Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.

            Target Information

            GPR15L is a newly identified ligand for GPR15, a member of the G protein-coupled receptor (GPCR) family. Upon ligation, GPR15L acts as a potent chemoattractant for GPR15-expressing T cells and together they mediate lymphocyte recruitment to the large intestine and skin. GPR15L is constitutively expressed by colon epithelial cells where its expression is minimally altered by intestinal inflammation. Conversely, GPR15L is nearly undetectable in adult epidermis but highly upregulated during wound healing and inflammation, particularly in psoriasis. Significant expression of GPR15L is also seen in additional mucosal epithelial cells, including those of the stomach, esophagus, and urinary tract. While maintaining similar expression patterns and intramolecular disulfide cysteine bridges found in members of the CC chemokine family, GPR15L differs from classic CC and CXC chemokines, whose active sites are found on the N-terminus, in that it relies on its C-terminus for receptor interaction. GPR15L was first identified as an antimicrobial peptide (AMP) due to its broad spectrum of antimicrobial activity, a property shared with many chemokines. GPR15L binds to an additional receptor, SUSD2, and early studies have indicated that both are downregulated in colon cancer tissue resulting in inhibited colon cancer growth.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: anti-microbial peptide with 57 amino acid residues; Antimicrobial peptide with 57 amino acid residues; antimicrobial peptide-57; AP-57; Colon-derived SUSD2 binding factor; CSBF; Protein GPR15 ligand; Protein GPR15L; Protein GPR15LG; RLLV1833; Secreted protein C10orf99

            View more View less

            Gene Aliases: AP-57; C10orf99; CSBF; GPR15L; GPR15LG; UNQ1833; UNQ1833/PRO3446

            View more View less

            UniProt ID: (Human) Q6UWK7

            View more View less

            Entrez Gene ID: (Human) 387695

            View more View less

            Function(s)

            cytokine activity

            Process(es)

            defense response to Gram-positive bacterium defense response to fungus negative regulation of cell division negative regulation of cell cycle G1/S phase transition
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            • Social Media
            • Contact Us
            • Report a Site Issue
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Argentina flag icon
            Argentina

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-r4nq5:80/100.66.79.32:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0