Product References
Regorafenib plus nivolumab in unresectable hepatocellular carcinoma: the phase 2 RENOBATE trial.
Nature medicine
Kim HD,Jung S,Lim HY,Ryoo BY,Ryu MH,Chuah S,Chon HJ,Kang B,Hong JY,Lee HC,Moon DB,Kim KH,Kim TW,Tai D,Chew V,Lee JS,Finn RS,Koh JY,Yoo C
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2024
Translation efficiency driven by CNOT3 subunit of the CCR4-NOT complex promotes leukemogenesis.
Nature communications
Ghashghaei M,Liu Y,Ettles J,Bombaci G,Ramkumar N,Liu Z,Escano L,Miko SS,Kim Y,Waldron JA,Do K,MacPherson K,Yuen KA,Taibi T,Yue M,Arsalan A,Jin Z,Edin G,Karsan A,Morin GB,Kuchenbauer F,Perna F,Bushell M,Vu LP
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Mar 15 00:00:00 UTC 2024
Spontaneous Osteoclastogenesis, a risk factor for bone metastasis in advanced luminal A-type breast cancer patients.
Frontiers in oncology
Fernández Vallone V,Borzone FR,Martinez LM,Giorello MB,Choi H,Dimase F,Feldman L,Bordenave RH,Chudzinski-Tavassi AM,Batagelj E,Chasseing NA
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Mar 10 00:00:00 UTC 2023
Local Myeloid-Derived Suppressor Cells Impair Progression of Experimental Autoimmune Uveitis by Alleviating Oxidative Stress and Inflammation.
Investigative ophthalmology & visual science
Lee JY,Kim S,Sohn HJ,Kim CH,Kim TG,Lee HS
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Tue Oct 03 00:00:00 UTC 2023
Scalable generation of functional human iPSC-derived CAR-macrophages that efficiently eradicate CD19-positive leukemia.
Journal for immunotherapy of cancer
Abdin SM,Paasch D,Kloos A,Oliveira MC,Jang MS,Ackermann M,Stamopoulou A,Mroch PJ,Falk CS,von Kaisenberg CS,Schambach A,Heuser M,Moritz T,Hansen G,Morgan M,Lachmann N
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Dec 22 00:00:00 UTC 2023
Saharan dust induces NLRP3-dependent inflammatory cytokines in an alveolar air-liquid interface co-culture model.
Particle and fibre toxicology
Bredeck G,Dobner J,Stahlmecke B,Fomba KW,Herrmann H,Rossi A,Schins RPF
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Oct 20 00:00:00 UTC 2023
LAL deficiency induced myeloid-derived suppressor cells as targets and biomarkers for lung cancer.
Journal for immunotherapy of cancer
Zhao T,Liu S,Hanna NH,Jalal S,Ding X,Wan J,Yan C,Du H
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
SARS-CoV-2 Delta (B.1.617.2) variant replicates and induces syncytia formation in human induced pluripotent stem cell-derived macrophages.
PeerJ
Thaweerattanasinp T,Wanitchang A,Saenboonrueng J,Srisutthisamphan K,Wanasen N,Sungsuwan S,Jongkaewwattana A,Chailangkarn T
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2023
Heliangin acts as a covalent ligand of RPS2 that disrupts pre-rRNA metabolic processes in NPM1-mutated acute myeloid leukemia.
Acta pharmaceutica Sinica. B
Feng Y,Han Y,Hu A,Qu Y,Hu Y,Wu H,Wang X,He L
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2023
Pharmacological inhibition of LSD1 triggers myeloid differentiation by targeting GSE1 oncogenic functions in AML.
Oncogene
Nicosia L,Boffo FL,Ceccacci E,Conforti F,Pallavicini I,Bedin F,Ravasio R,Massignani E,Somervaille TCP,Minucci S,Bonaldi T
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Tue Feb 01 00:00:00 UTC 2022
TGF-β orchestrates the phenotype and function of monocytic myeloid-derived suppressor cells in colorectal cancer.
Cancer immunology, immunotherapy : CII
Gneo L,Rizkalla N,Hejmadi R,Mussai F,de Santo C,Middleton G
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Jul 01 00:00:00 UTC 2022
Early macrophage response to obesity encompasses Interferon Regulatory Factor 5 regulated mitochondrial architecture remodelling.
Nature communications
Orliaguet L,Ejlalmanesh T,Humbert A,Ballaire R,Diedisheim M,Julla JB,Chokr D,Cuenco J,Michieletto J,Charbit J,Lindén D,Boucher J,Potier C,Hamimi A,Lemoine S,Blugeon C,Legoix P,Lameiras S,Baudrin LG,Baulande S,Soprani A,Castelli FA,Fenaille F,Riveline JP,Dalmas E,Rieusset J,Gautier JF,Venteclef N,Alzaid F
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Immunocytochemistry
Tue Aug 30 00:00:00 UTC 2022
A Matrix-Assisted Laser Desorption/Ionization Time-of-Flight Assay Identifies Nilotinib as an Inhibitor of Inflammation in Acute Myeloid Leukemia.
Journal of medicinal chemistry
Marín-Rubio JL,Peltier-Heap RE,Dueñas ME,Heunis T,Dannoura A,Inns J,Scott J,Simpson AJ,Blair HJ,Heidenreich O,Allan JM,Watt JE,Martin MP,Saxty B,Trost M
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Sep 22 00:00:00 UTC 2022
Decoding the pathogenesis of Diamond-Blackfan anemia using single-cell RNA-seq.
Cell discovery
Wang B,Wang C,Wan Y,Gao J,Ma Y,Zhang Y,Tong J,Zhang Y,Liu J,Chang L,Xu C,Shen B,Chen Y,Jiang E,Kurita R,Nakamura Y,Lim KC,Engel JD,Zhou J,Cheng T,Zhu X,Zhu P,Shi L
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Tue May 10 00:00:00 UTC 2022
Withdrawn: Parecoxib inhibits inflammatory responses in a mouse model of sepsis.
FEBS open bio
No authors listed
14-0149 was used in Flow cytometry/Cell sorting to suggest that Parecoxib inhibits the inflammatory response and has protective effects against sepsis in mice, and may have potential as a novel therapeutic method for treating sepsis.
Sat Jan 01 00:00:00 UTC 2022
SARS-CoV-2 infection paralyzes cytotoxic and metabolic functions of the immune cells.
Heliyon
Singh Y,Trautwein C,Fendel R,Krickeberg N,Berezhnoy G,Bissinger R,Ossowski S,Salker MS,Casadei N,Riess O
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Tue Jun 01 00:00:00 UTC 2021
Exposure of a specific pleioform of multifunctional glyceraldehyde 3-phosphate dehydrogenase initiates CD14-dependent clearance of apoptotic cells.
Cell death & disease
Chaudhary S,Patidar A,Dhiman A,Chaubey GK,Dilawari R,Talukdar S,Modanwal R,Raje M
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Neutralization
Thu Sep 30 00:00:00 UTC 2021
The long non-coding RNA Cancer Susceptibility 15 (CASC15) is induced by isocitrate dehydrogenase (IDH) mutations and maintains an immature phenotype in adult acute myeloid leukemia.
Haematologica
Grasedieck S,Ruess C,Krowiorz K,Lux S,Pochert N,Schwarzer A,Klusmann JH,Jongen-Lavrencic M,Herold T,Bullinger L,Pollack JR,Rouhi A,Kuchenbauer F
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
Gene expression network analyses during infection with virulent and avirulent Trypanosoma cruzi strains unveil a role for fibroblasts in neutrophil recruitment and activation.
PLoS pathogens
Oliveira AER,Pereira MCA,Belew AT,Ferreira LRP,Pereira LMN,Neves EGA,Nunes MDCP,Burleigh BA,Dutra WO,El-Sayed NM,Gazzinelli RT,Teixeira SMR
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Sat Aug 01 00:00:00 UTC 2020
A functional antibody cross-reactive to both human and murine cytotoxic T-lymphocyte-associated protein 4 via binding to an N-glycosylation epitope.
mAbs
Li D,Li J,Chu H,Wang Z
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Feb 12 00:00:00 UTC 2021
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Tue Oct 06 00:00:00 UTC 2020
MiR-103 protects from recurrent spontaneous abortion via inhibiting STAT1 mediated M1 macrophage polarization.
International journal of biological sciences
Zhu X,Liu H,Zhang Z,Wei R,Zhou X,Wang Z,Zhao L,Guo Q,Zhang Y,Chu C,Wang L,Li X
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Nov 12 00:00:00 UTC 2021
Patient iPSC-Derived Macrophages to Study Inborn Errors of the IFN-γ Responsive Pathway.
Cells
Haake K,Neehus AL,Buchegger T,Kühnel MP,Blank P,Philipp F,Oleaga-Quintas C,Schulz A,Grimley M,Goethe R,Jonigk D,Kalinke U,Boisson-Dupuis S,Casanova JL,Bustamante J,Lachmann N
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Wed Feb 19 00:00:00 UTC 2020
Chemerin enhances the adhesion and migration of human endothelial progenitor cells and increases lipid accumulation in mice with atherosclerosis.
Lipids in health and disease
Jia J,Yu F,Xiong Y,Wei W,Ma H,Nisi F,Song X,Yang L,Wang D,Yuan G,Zhou H
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Sun Sep 20 00:00:00 UTC 2020
Phase I Study of Ficlatuzumab and Cetuximab in Cetuximab-Resistant, Recurrent/Metastatic Head and Neck Cancer.
Cancers
Bauman JE,Ohr J,Gooding WE,Ferris RL,Duvvuri U,Kim S,Johnson JT,Soloff AC,Wallweber G,Winslow J,Gaither-Davis A,Grandis JR,Stabile LP
14-0149-82 was used in Flow Cytometry to establish the recommended Phase II dose (RP2D). Secondary objectives included overall response rate (ORR), progression-free survival (PFS), and overall survival (OS).
Thu Jun 11 00:00:00 UTC 2020
NLRP3 Inflammasome Activation in Hemodialysis and Hypertensive Patients with Intact Kidney Function.
Toxins
Ulrich C,Wildgrube S,Fiedler R,Seibert E,Kneser L,Fick S,Schäfer C,Markau S,Trojanowicz B,Girndt M
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Oct 26 00:00:00 UTC 2020
LILRB3 (ILT5) is a myeloid cell checkpoint that elicits profound immunomodulation.
JCI insight
Yeboah M,Papagregoriou C,Jones DC,Chan HTC,Hu G,McPartlan JS,Schiött T,Mattson U,Mockridge CI,Tornberg UC,Hambe B,Ljungars A,Mattsson M,Tews I,Glennie MJ,Thirdborough SM,Trowsdale J,Frendeus B,Chen J,Cragg MS,Roghanian A
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
NLRP3 inflammasome expression in peripheral blood monocytes of coronary heart disease patients and its modulation by rosuvastatin.
Molecular medicine reports
Zhu J,Wu S,Hu S,Li H,Li M,Geng X,Wang H
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Aug 01 00:00:00 UTC 2019
Novel lineage depletion preserves autologous blood stem cells for gene therapy of Fanconi anemia complementation group A.
Haematologica
Adair JE,Chandrasekaran D,Sghia-Hughes G,Haworth KG,Woolfrey AE,Burroughs LM,Choi GY,Becker PS,Kiem HP
14-0149 was used in Flow cytometry/Cell sorting to demonstrate an altered ratio of CD34Hi to CD34Lo cells in Fanconi patients relative to healthy donors, with exclusive in vitro repopulating ability in only CD34Hi cells, underscoring a need for novel strategies to preserve limited CD34+ cells.
Thu Nov 01 00:00:00 UTC 2018
Characterizing the Role of Monocytes in T Cell Cancer Immunotherapy Using a 3D Microfluidic Model.
Frontiers in immunology
Lee SWL,Adriani G,Ceccarello E,Pavesi A,Tan AT,Bertoletti A,Kamm RD,Wong SC
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Jun 24 00:00:00 UTC 2019
AURKA Suppresses Leukemic THP-1 Cell Differentiation through Inhibition of the KDM6B Pathway.
Molecules and cells
Park JW,Cho H,Oh H,Kim JY,Seo SB
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu May 31 00:00:00 UTC 2018
Butyrate upregulates the TLR4 expression and the phosphorylation of MAPKs and NK-κB in colon cancer cell in vitro.
Oncology letters
Xiao T,Wu S,Yan C,Zhao C,Jin H,Yan N,Xu J,Wu Y,Li C,Shao Q,Xia S
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Oct 01 00:00:00 UTC 2018
Impaired efferocytosis by monocytes in multiple myeloma.
Oncology letters
Liang YY,Schwarzinger I,Simonitsch-Klupp I,Agis H,Oehler R
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Sun Jul 01 00:00:00 UTC 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
Comprehensive characterization of chorionic villi-derived mesenchymal stromal cells from human placenta.
Stem cell research & therapy
Ventura Ferreira MS,Bienert M,Müller K,Rath B,Goecke T,Opländer C,Braunschweig T,Mela P,Brümmendorf TH,Beier F,Neuss S
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2018
Cellular metabolism constrains innate immune responses in early human ontogeny.
Nature communications
Kan B,Michalski C,Fu H,Au HHT,Lee K,Marchant EA,Cheng MF,Anderson-Baucum E,Aharoni-Simon M,Tilley P,Mirmira RG,Ross CJ,Luciani DS,Jan E,Lavoie PM
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Nov 16 00:00:00 UTC 2018
The pro-inflammatory phenotype of the human non-classical monocyte subset is attributed to senescence.
Cell death & disease
Ong SM,Hadadi E,Dang TM,Yeap WH,Tan CT,Ng TP,Larbi A,Wong SC
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Feb 15 00:00:00 UTC 2018
Macrophages are exploited from an innate wound healing response to facilitate cancer metastasis.
Nature communications
Muliaditan T,Caron J,Okesola M,Opzoomer JW,Kosti P,Georgouli M,Gordon P,Lall S,Kuzeva DM,Pedro L,Shields JD,Gillett CE,Diebold SS,Sanz-Moreno V,Ng T,Hoste E,Arnold JN
14-0149 was used in Immunohistochemistry-immunofluorescence to show that tumours can exploit this response to facilitate transendothelial migration and metastatic spread of the disease, which can be pharmacologically targeted using a clinically relevant HO-1 inhibitor.
Fri Jul 27 00:00:00 UTC 2018
A therapeutic T cell receptor mimic antibody targets tumor-associated PRAME peptide/HLA-I antigens.
The Journal of clinical investigation
Chang AY,Dao T,Gejman RS,Jarvis CA,Scott A,Dubrovsky L,Mathias MD,Korontsvit T,Zakhaleva V,Curcio M,Hendrickson RC,Liu C,Scheinberg DA
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Jun 30 00:00:00 UTC 2017
Machine-learning algorithms define pathogen-specific local immune fingerprints in peritoneal dialysis patients with bacterial infections.
Kidney international
Zhang J,Friberg IM,Kift-Morgan A,Parekh G,Morgan MP,Liuzzi AR,Lin CY,Donovan KL,Colmont CS,Morgan PH,Davis P,Weeks I,Fraser DJ,Topley N,Eberl M
14-0149 was used in Flow cytometry/Cell sorting to demonstrate that different groups of bacteria induce qualitatively distinct local immune fingerprints.
Sat Jul 01 00:00:00 UTC 2017
Membrane nanoclusters of FcγRI segregate from inhibitory SIRPα upon activation of human macrophages.
The Journal of cell biology
Lopes FB,Bálint Š,Valvo S,Felce JH,Hessel EM,Dustin ML,Davis DM
14-0149 was used in Flow cytometry/Cell sorting to investigate the distribution of FcγRI, FcγRII, and SIRPα on the surface of macrophages.
Mon Apr 03 00:00:00 UTC 2017
Soluble CD83 Inhibits T Cell Activation by Binding to the TLR4/MD-2 Complex on CD14+ Monocytes.
Journal of immunology (Baltimore, Md. : 1950)
Horvatinovich JM,Grogan EW,Norris M,Steinkasserer A,Lemos H,Mellor AL,Tcherepanova IY,Nicolette CA,DeBenedette MA
14-0149 was used in Flow cytometry/Cell sorting to investigate the mechanism of action underlying the tolerogenic function of soluble CD83.
Wed Mar 15 00:00:00 UTC 2017
Toll-like receptor 3 activation selectively reverses HIV latency in microglial cells.
Retrovirology
Alvarez-Carbonell D,Garcia-Mesa Y,Milne S,Das B,Dobrowolski C,Rojas R,Karn J
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Immunofluorescence
Mon Feb 06 00:00:00 UTC 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2017
Kaposi's Sarcoma-Associated Herpesvirus Increases PD-L1 and Proinflammatory Cytokine Expression in Human Monocytes.
mBio
Host KM,Jacobs SR,West JA,Zhang Z,Costantini LM,Stopford CM,Dittmer DP,Damania B
14-0149 was used in Flow cytometry/Cell sorting to demonstrate that primary human monocytes infected with kaposi's sarcoma-associated herpesvirus (KSHV) upregulate PD-L1 transcription and protein expression.
Tue Oct 10 00:00:00 UTC 2017
Pulmonary sarcoidosis is associated with high-level inducible co-stimulator (ICOS) expression on lung regulatory T cells--possible implications for the ICOS/ICOS-ligand axis in disease course and resolution.
Clinical and experimental immunology
Sakthivel P,Grunewald J,Eklund A,Bruder D,Wahlström J
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2016
Thrombomodulin regulates monocye differentiation via PKCδ and ERK1/2 pathway in vitro and in atherosclerotic artery.
Scientific reports
Tsai CS,Lin YW,Huang CY,Shih CM,Tsai YT,Tsao NW,Lin CS,Shih CC,Jeng H,Lin FY
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Dec 02 00:00:00 UTC 2016
Thrombomodulin regulates monocye differentiation via PKCδ and ERK1/2 pathway in vitro and in atherosclerotic artery.
Scientific reports
Tsai CS,Lin YW,Huang CY,Shih CM,Tsai YT,Tsao NW,Lin CS,Shih CC,Jeng H,Lin FY
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri Dec 02 00:00:00 UTC 2016
The prognostic effects of tumor infiltrating regulatory T cells and myeloid derived suppressor cells assessed by multicolor flow cytometry in gastric cancer patients.
Oncotarget
Choi HS,Ha SY,Kim HM,Ahn SM,Kang MS,Kim KM,Choi MG,Lee JH,Sohn TS,Bae JM,Kim S,Kang ES
14-0149 was used in Flow cytometry/Cell sorting to demonstrate that in gastric cancer, a high frequency of Tregs among tumour-infiltrating CD4+ T cells is favourable, but an increased proportion of MDSCs is an adverse independent prognostic factor.
Tue Feb 16 00:00:00 UTC 2016
MicroRNA-941 Expression in Polymorphonuclear Granulocytes Is Not Related to Granulomatosis with Polyangiitis.
PloS one
Svendsen JB,Baslund B,Cramer EP,Rapin N,Borregaard N,Cowland JB
14-0149 was used in Magnetic cell separation to find that PRTN3 mRNA, KDM6B mRNA, and miR-941 expression levels in PMNs do not differ between GPA patients and healthy controls.
Wed May 31 00:00:00 UTC 2017
The Effect of Chronic Hepatitis B Virus Infection on BDCA3+ Dendritic Cell Frequency and Function.
PloS one
van der Aa E,Buschow SI,Biesta PJ,Janssen HL,Woltman AM
14-0149 was used in Flow cytometry/Cell sorting to investigate the role of BDCA3+ DCs in immune response against the hepititis B virus, showing that BDCA3+ DCs are available in the blood and on site in HBV infected livers.
Wed Aug 02 00:00:00 UTC 2017
STK4 regulates TLR pathways and protects against chronic inflammation-related hepatocellular carcinoma.
The Journal of clinical investigation
Li W,Xiao J,Zhou X,Xu M,Hu C,Xu X,Lu Y,Liu C,Xue S,Nie L,Zhang H,Li Z,Zhang Y,Ji F,Hui L,Tao W,Wei B,Wang H
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Nov 02 00:00:00 UTC 2015
Brief Report: IFIH1 Mutation Causes Systemic Lupus Erythematosus With Selective IgA Deficiency.
Arthritis & rheumatology (Hoboken, N.J.)
Van Eyck L,De Somer L,Pombal D,Bornschein S,Frans G,Humblet-Baron S,Moens L,de Zegher F,Bossuyt X,Wouters C,Liston A
14-0149 was used in Flow cytometry/Cell sorting to determine the underlying genetic defect responsible for severe early onset and refractory systemic lupus erythematosus.
Mon Jun 01 00:00:00 UTC 2015
Thymic HIV-2 infection uncovers posttranscriptional control of viral replication in human thymocytes.
Journal of virology
Nunes-Cabaço H,Matoso P,Foxall RB,Tendeiro R,Pires AR,Carvalho T,Pinheiro AI,Soares RS,Sousa AE
14-0149 was used in Flow cytometry/Cell sorting to assess the direct impact of HIV-2 infection on the human thymus.
Sun Feb 01 00:00:00 UTC 2015
Liposomal Glutathione Supplementation Restores TH1 Cytokine Response to Mycobacterium tuberculosis Infection in HIV-Infected Individuals.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Ly J,Lagman M,Saing T,Singh MK,Tudela EV,Morris D,Anderson J,Daliva J,Ochoa C,Patel N,Pearce D,Venketaraman V
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2015
Investigating the causes for decreased levels of glutathione in individuals with type II diabetes.
PloS one
Lagman M,Ly J,Saing T,Kaur Singh M,Vera Tudela E,Morris D,Chi PT,Ochoa C,Sathananthan A,Venketaraman V
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Wed Dec 30 00:00:00 UTC 2015
A novel antibody-drug conjugate targeting SAIL for the treatment of hematologic malignancies.
Blood cancer journal
Kim SY,Theunissen JW,Balibalos J,Liao-Chan S,Babcock MC,Wong T,Cairns B,Gonzalez D,van der Horst EH,Perez M,Levashova Z,Chinn L,D'Alessio JA,Flory M,Bermudez A,Jackson DY,Ha E,Monteon J,Bruhns MF,Chen G,Migone TS
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Fri May 29 00:00:00 UTC 2015
A cell-based systems biology assessment of human blood to monitor immune responses after influenza vaccination.
PloS one
Hoek KL,Samir P,Howard LM,Niu X,Prasad N,Galassie A,Liu Q,Allos TM,Floyd KA,Guo Y,Shyr Y,Levy SE,Joyce S,Edwards KM,Link AJ
14-0149 was used in Immunocytochemistry to monitor immune responses after influenza vaccination as part of a cell-based systems biology assessment of the blood of human subjects.
Thu Dec 03 00:00:00 UTC 2015
A lung-on-a-chip array with an integrated bio-inspired respiration mechanism.
Lab on a chip
Stucki AO,Stucki JD,Hall SR,Felder M,Mermoud Y,Schmid RA,Geiser T,Guenat OT
14-0149 was used in Flow cytometry/Cell sorting to report a novel lung-on-a-chip that mimics the pulmonary parenchymal environment, including the alveolar barrier and the 3D cyclic strain induced by breathing movements.
Sat Mar 07 00:00:00 UTC 2015
Comparative transcriptional and functional profiling defines conserved programs of intestinal DC differentiation in humans and mice.
Nature immunology
Watchmaker PB,Lahl K,Lee M,Baumjohann D,Morton J,Kim SJ,Zeng R,Dent A,Ansel KM,Diamond B,Hadeiba H,Butcher EC
14-0149 was used in Flow cytometry/Cell sorting to characterise human gut dendritic cell populations, and define their relationship to previously studied human and mouse dendritic cells.
Wed Jan 01 00:00:00 UTC 2014
The role of transforming growth factor β signaling in fibroblast-like synoviocytes from patients with oligoarticular juvenile idiopathic arthritis: dysregulation of transforming growth factor β signaling, including overexpression of bone morphogenetic protein 4, may lead to a chondrocyte phenotype and may contribute to bony hypertrophy.
Arthritis & rheumatology (Hoboken, N.J.)
Brescia AC,Simonds MM,McCahan SM,Fawcett PT,Rose CD
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Immunofluorescence
Thu May 01 00:00:00 UTC 2014
Genuine Immunomodulation With dSLIM.
Molecular therapy. Nucleic acids
Kapp K,Kleuss C,Schroff M,Wittig B
14-0149 was used in Flow cytometry/Cell sorting to describe the structure and biologic profile of dSLIM, showing that it has immunomodulation properties.
Tue Jun 24 00:00:00 UTC 2014
Microbe-specific unconventional T cells induce human neutrophil differentiation into antigen cross-presenting cells.
Journal of immunology (Baltimore, Md. : 1950)
Davey MS,Morgan MP,Liuzzi AR,Tyler CJ,Khan MWA,Szakmany T,Hall JE,Moser B,Eberl M
14-0149 was used in Immunofluorescence to investigate how unconventional T cells represent key controllers of neutrophil-driven innate and adaptive responses to a broad range of pathogens.
Wed Oct 01 00:00:00 UTC 2014
Human mesenchymal stem cells possess different biological characteristics but do not change their therapeutic potential when cultured in serum free medium.
Stem cell research & therapy
Wang Y,Wu H,Yang Z,Chi Y,Meng L,Mao A,Yan S,Hu S,Zhang J,Zhang Y,Yu W,Ma Y,Li T,Cheng Y,Wang Y,Wang S,Liu J,Han J,Li C,Liu L,Xu J,Han ZB,Han ZC
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Dec 04 00:00:00 UTC 2014
Biologically active polymers from spontaneous carotenoid oxidation: a new frontier in carotenoid activity.
PloS one
Johnston JB,Nickerson JG,Daroszewski J,Mogg TJ,Burton GW
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Jun 25 00:00:00 UTC 2015
Biologically active polymers from spontaneous carotenoid oxidation: a new frontier in carotenoid activity.
PloS one
Johnston JB,Nickerson JG,Daroszewski J,Mogg TJ,Burton GW
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Jun 25 00:00:00 UTC 2015
Monocytes from chronic HBV patients react in vitro to HBsAg and TLR by producing cytokines irrespective of stage of disease.
PloS one
Boltjes A,Groothuismink ZM,van Oord GW,Janssen HL,Woltman AM,Boonstra A
14-0149 was used in Flow cytometry/Cell sorting to investigate the in vitro and in vivo effects of hepatitis B virus surface antigen on monocytes from chronic HBV patients.
Tue Jan 06 00:00:00 UTC 2015
Immune response to RB1-regulated senescence limits radiation-induced osteosarcoma formation.
The Journal of clinical investigation
Kansara M,Leong HS,Lin DM,Popkiss S,Pang P,Garsed DW,Walkley CR,Cullinane C,Ellul J,Haynes NM,Hicks R,Kuijjer ML,Cleton-Jansen AM,Hinds PW,Smyth MJ,Thomas DM
14-0149 was used in Flow cytometry/Cell sorting to indicate that overcoming the immune response to senescence is a rate-limiting step in the formation of IR-induced osteosarcoma.
Sun Dec 01 00:00:00 UTC 2013
Lipoxin A4 regulates natural killer cell and type 2 innate lymphoid cell activation in asthma.
Science translational medicine
Barnig C,Cernadas M,Dutile S,Liu X,Perrella MA,Kazani S,Wechsler ME,Israel E,Levy BD
14-0149 was used in Flow cytometry/Cell sorting to identify the potential of innate lymphoid cells to decrease airway inflammation.
Wed Feb 27 00:00:00 UTC 2013
Somatostatin expression in human hair follicles and its potential role in immune privilege.
The Journal of investigative dermatology
Breitkopf T,Lo BK,Leung G,Wang E,Yu M,Carr N,Zloty D,Cowan B,Shapiro J,McElwee KJ
14-0149 was used in Flow cytometry/Cell sorting to demonstrate and quantify the functional immune privilege properties apparent in human hair follicles.
Mon Jul 01 00:00:00 UTC 2013
Monocytes and γδ T cells control the acute-phase response to intravenous zoledronate: insights from a phase IV safety trial.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Welton JL,Morgan MP,Martí S,Stone MD,Moser B,Sewell AK,Turton J,Eberl M
14-0149 was used in Flow cytometry/Cell sorting to investigate the acute-phase response of otherwise healthy individuals to first-time intravenous treatment with aminobisphosphonates zoledronate.
Fri Mar 01 00:00:00 UTC 2013
Noncanonical dendritic cell differentiation and survival driven by a bacteremic pathogen.
Journal of leukocyte biology
Miles B,Scisci E,Carrion J,Sabino GJ,Genco CA,Cutler CW
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Aug 01 00:00:00 UTC 2013
Dectin-1/TLR2 and NOD2 agonists render dendritic cells susceptible to infection by X4-using HIV-1 and promote cis-infection of CD4(+) T cells.
PloS one
Côté SC,Plante A,Tardif MR,Tremblay MJ
14-0149 was used in Flow cytometry/Cell sorting to highlight the crucial role of dendritic cells at different stages of HIV-1 infection.
Thu Jan 23 00:00:00 UTC 2014
CD137L-stimulated dendritic cells are more potent than conventional dendritic cells at eliciting cytotoxic T-cell responses.
Oncoimmunology
Harfuddin Z,Kwajah S,Chong Nyi Sim A,Macary PA,Schwarz H
14-0149 was used in Flow cytometry/Cell sorting to develop improved dendritic cell-based immunotherapy using CD137 signalling.
Fri Nov 01 00:00:00 UTC 2013
HIV-1 Tat protein binds to TLR4-MD2 and signals to induce TNF-α and IL-10.
Retrovirology
Ben Haij N,Leghmari K,Planès R,Thieblemont N,Bahraoui E
14-0149 was used in Inhibition experiments to provide the first evidence for TLR4 as a potential candidate receptor for HIV-1 Tat protein.
Mon Oct 28 00:00:00 UTC 2013
Induction of autophagy is essential for monocyte-macrophage differentiation.
Blood
Zhang Y,Morgan MJ,Chen K,Choksi S,Liu ZG
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Thu Mar 22 00:00:00 UTC 2012
Dysfunctional B-cell activation in cirrhosis resulting from hepatitis C infection associated with disappearance of CD27-positive B-cell population.
Hepatology (Baltimore, Md.)
Doi H,Iyer TK,Carpenter E,Li H,Chang KM,Vonderheide RH,Kaplan DE
14-0149 was used in Blocking experiments to investigate the effect of viral infection, cirrhosis, and liver cancer on memory B-cell frequency and function in the spectrum of hepatitis C virus disease.
Thu Mar 01 00:00:00 UTC 2012
Cytomegalovirus-specific T-cell reactivity in biliary atresia at the time of diagnosis is associated with deficits in regulatory T cells.
Hepatology (Baltimore, Md.)
Brindley SM,Lanham AM,Karrer FM,Tucker RM,Fontenot AP,Mack CL
14-0149 was used in Flow cytometry/Cell sorting to investigate the viral-induced pathogenesis of BA and mechanisms for its associated autoimmunity.
Sun Apr 01 00:00:00 UTC 2012
Negative regulation of JAK2 by H3K9 methyltransferase G9a in leukemia.
Molecular and cellular biology
Son HJ,Kim JY,Hahn Y,Seo SB
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2012
Ulcerative colitis impairs the acylethanolamide-based anti-inflammatory system reversal by 5-aminosalicylic acid and glucocorticoids.
PloS one
Suárez J,Romero-Zerbo Y,Márquez L,Rivera P,Iglesias M,Bermúdez-Silva FJ,Andreu M,Rodríguez de Fonseca F
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Immunofluorescence
Thu Dec 13 00:00:00 UTC 2012
Role of toll like receptors in irritable bowel syndrome: differential mucosal immune activation according to the disease subtype.
PloS one
Belmonte L,Beutheu Youmba S,Bertiaux-Vandaële N,Antonietti M,Lecleire S,Zalar A,Gourcerol G,Leroi AM,Déchelotte P,Coëffier M,Ducrotté P
14-0149 was used in Flow cytometry/Cell sorting to investigate the molecular pathophysiology of irritable bowel syndrome.
Mon Jan 21 00:00:00 UTC 2013
Upregulation of programmed death-1 on T cells and programmed death ligand-1 on monocytes in septic shock patients.
Critical care (London, England)
Zhang Y,Li J,Lou J,Zhou Y,Bo L,Zhu J,Zhu K,Wan X,Cai Z,Deng X
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Wed Dec 07 00:00:00 UTC 2011
Human neutrophil clearance of bacterial pathogens triggers anti-microbial γδ T cell responses in early infection.
PLoS pathogens
Davey MS,Lin CY,Roberts GW,Heuston S,Brown AC,Chess JA,Toleman MA,Gahan CG,Hill C,Parish T,Williams JD,Davies SJ,Johnson DW,Topley N,Moser B,Eberl M
14-0149 was used in Flow cytometry/Cell sorting to demonstrate that Vγ9/Vδ2 T cells provide potent survival signals resulting in neutrophil activation and the release of the neutrophil chemoattractant CXCL8.
Sun May 01 00:00:00 UTC 2011
Generation of bivalent chromatin domains during cell fate decisions.
Epigenetics & chromatin
De Gobbi M,Garrick D,Lynch M,Vernimmen D,Hughes JR,Goardon N,Luc S,Lower KM,Sloane-Stanley JA,Pina C,Soneji S,Renella R,Enver T,Taylor S,Jacobsen SE,Vyas P,Gibbons RJ,Higgs DR
14-0149 was used in Flow cytometry/Cell sorting to investigate the patterns and levels of histone modifications at bivalent genes, showing the generation of bivalent chromatin domains during cell fate decisions.
Mon Jun 06 00:00:00 UTC 2011
Comparison of gene expression profiles between human and mouse monocyte subsets.
Blood
Ingersoll MA,Spanbroek R,Lottaz C,Gautier EL,Frankenberger M,Hoffmann R,Lang R,Haniffa M,Collin M,Tacke F,Habenicht AJ,Ziegler-Heitbrock L,Randolph GJ
14-0149 was used in Flow cytometry/Cell sorting to identify how human and mouse monocyte subsets are far more broadly conserved than currently recognised.
Thu Jan 21 00:00:00 UTC 2010
Glycosidic Tn-based vaccines targeting dermal dendritic cells favor germinal center B-cell development and potent antibody response in the absence of adjuvant.
Blood
Freire T,Zhang X,Dériaud E,Ganneau C,Vichier-Guerre S,Azria E,Launay O,Lo-Man R,Bay S,Leclerc C
14-0149 was used in Flow cytometry/Cell sorting to study dendritic cell antigen uptake and presentation by targeting the antigen receptor, macrophage galactose-type lectin (MGL), with tumor-associated Tn antigens.
Thu Nov 04 00:00:00 UTC 2010
Nutrient modification of the innate immune response: a novel mechanism by which saturated fatty acids greatly amplify monocyte inflammation.
Arteriosclerosis, thrombosis, and vascular biology
Schwartz EA,Zhang WY,Karnik SK,Borwege S,Anand VR,Laine PS,Su Y,Reaven PD
14-0149 was used in Flow cytometry/Cell sorting to investigate whether metabolism of saturated fatty acids cooperatively amplifies Toll-like receptor-4-mediated inflammation.
Thu Apr 01 00:00:00 UTC 2010
CD137 ligand signaling induces human monocyte to dendritic cell differentiation.
European journal of immunology
Kwajah M M S,Schwarz H
14-0149 was used in Flow cytometry/Cell sorting to deduce that peripheral monocytes can be differentiated into mature inflammatory dendritic cells by the single factor CD137.
Thu Jul 01 00:00:00 UTC 2010
Ro60-associated single-stranded RNA links inflammation with fetal cardiac fibrosis via ligation of TLRs: a novel pathway to autoimmune-associated heart block.
Journal of immunology (Baltimore, Md. : 1950)
Clancy RM,Alvarez D,Komissarova E,Barrat FJ,Swartz J,Buyon JP
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Feb 15 00:00:00 UTC 2010
Ligand of scavenger receptor class A indirectly induces maturation of human blood dendritic cells via production of tumor necrosis factor-alpha.
Blood
Jin JO,Park HY,Xu Q,Park JI,Zvyagintseva T,Stonik VA,Kwak JY
14-0149 was used in Flow cytometry/Cell sorting to demonstrate a potential pathway for SR-A to enhance T-cell stimulatory capacity.
Thu Jun 04 00:00:00 UTC 2009
Severe loss of invariant NKT cells exhibiting anti-HTLV-1 activity in patients with HTLV-1-associated disorders.
Blood
Azakami K,Sato T,Araya N,Utsunomiya A,Kubota R,Suzuki K,Hasegawa D,Izumi T,Fujita H,Aratani S,Fujii R,Yagishita N,Kamijuku H,Kanekura T,Seino K,Nishioka K,Nakajima T,Yamano Y
14-0149 was used in Flow cytometry/Cell sorting to assess the contribution of iNKT cells to the immune defence against human T-lymphotropic virus type 1 and associated disorders.
Thu Oct 08 00:00:00 UTC 2009
B cells and monocytes from patients with active multiple sclerosis exhibit increased surface expression of both HERV-H Env and HERV-W Env, accompanied by increased seroreactivity.
Retrovirology
Brudek T,Christensen T,Aagaard L,Petersen T,Hansen HJ,Møller-Larsen A
Published figure using CD14 monoclonal antibody (Product # 14-0149-82) in Flow Cytometry
Mon Nov 16 00:00:00 UTC 2009
C-terminal repeats of Clostridium difficile toxin A induce production of chemokine and adhesion molecules in endothelial cells and promote migration of leukocytes.
Infection and immunity
Yeh CY,Lin CN,Chang CF,Lin CH,Lien HT,Chen JY,Chia JS
14-0149 was used in Flow cytometry/Cell sorting to investigate how Clostridium difficile toxin A may bind to multiple carbohydrate motifs to exert its biological activity on human endothelial cells.
Sat Mar 01 00:00:00 UTC 2008
C1q and MBL, components of the innate immune system, influence monocyte cytokine expression.
Journal of leukocyte biology
Fraser DA,Bohlson SS,Jasinskiene N,Rawal N,Palmarini G,Ruiz S,Rochford R,Tenner AJ
14-0149 was used in Flow cytometry/Cell sorting to show that C1q and MBL modulate cytokine production at the mRNA and protein levels.
Sat Jul 01 00:00:00 UTC 2006
Endocarditis-associated oral streptococci promote rapid differentiation of monocytes into mature dendritic cells.
Infection and immunity
Hahn CL,Schenkein HA,Tew JG
14-0149 was used in Neutralization experiments to investigate the mechanism of pathogenesis of oral-streptococci-induced endocarditis, showing that they promote rapid differentiation of monocytes into mature dendritic cells.
Mon Aug 01 00:00:00 UTC 2005