Product References
Flow pattern-dependent mitochondrial dynamics regulates the metabolic profile and inflammatory state of endothelial cells.
JCI insight
Hong SG,Shin J,Choi SY,Powers JC,Meister BM,Sayoc J,Son JS,Tierney R,Recchia FA,Brown MD,Yang X,Park JY
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunohistochemistry
Thu Sep 22 00:00:00 UTC 2022
Monomeric C-reactive protein via endothelial CD31 for neurovascular inflammation in an ApoE genotype-dependent pattern: A risk factor for Alzheimer's disease?
Aging cell
Zhang Z,Na H,Gan Q,Tao Q,Alekseyev Y,Hu J,Yan Z,Yang JB,Tian H,Zhu S,Li Q,Rajab IM,Blusztajn JK,Wolozin B,Emili A,Zhang X,Stein T,Potempa LA,Qiu WQ
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Western Blot
Mon Nov 01 00:00:00 UTC 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Tue May 11 00:00:00 UTC 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Tue May 11 00:00:00 UTC 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Tue May 11 00:00:00 UTC 2021
VE-cadherin endocytosis controls vascular integrity and patterning during development.
The Journal of cell biology
Grimsley-Myers CM,Isaacson RH,Cadwell CM,Campos J,Hernandes MS,Myers KR,Seo T,Giang W,Griendling KK,Kowalczyk AP
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunohistochemistry
Mon May 04 00:00:00 UTC 2020
VE-cadherin endocytosis controls vascular integrity and patterning during development.
The Journal of cell biology
Grimsley-Myers CM,Isaacson RH,Cadwell CM,Campos J,Hernandes MS,Myers KR,Seo T,Giang W,Griendling KK,Kowalczyk AP
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunohistochemistry
Mon May 04 00:00:00 UTC 2020
Interleukin 35 Delays Hindlimb Ischemia-Induced Angiogenesis Through Regulating ROS-Extracellular Matrix but Spares Later Regenerative Angiogenesis.
Frontiers in immunology
Fu H,Sun Y,Shao Y,Saredy J,Cueto R,Liu L,Drummer C,Johnson C,Xu K,Lu Y,Li X,Meng S,Xue ER,Tan J,Jhala NC,Yu D,Zhou Y,Bayless KJ,Yu J,Rogers TJ,Hu W,Snyder NW,Sun J,Qin X,Jiang X,Wang H,Yang X
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Fri Jul 09 00:00:00 UTC 2021
Cellular cartography of the organ of Corti based on optical tissue clearing and machine learning.
eLife
Urata S,Iida T,Yamamoto M,Mizushima Y,Fujimoto C,Matsumoto Y,Yamasoba T,Okabe S
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunohistochemistry
Fri Jan 18 00:00:00 UTC 2019
CNS lymphatic drainage and neuroinflammation are regulated by meningeal lymphatic vasculature.
Nature neuroscience
Louveau A,Herz J,Alme MN,Salvador AF,Dong MQ,Viar KE,Herod SG,Knopp J,Setliff JC,Lupi AL,Da Mesquita S,Frost EL,Gaultier A,Harris TH,Cao R,Hu S,Lukens JR,Smirnov I,Overall CC,Oliver G,Kipnis J
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunocytochemistry
Mon Oct 01 00:00:00 UTC 2018
CNS lymphatic drainage and neuroinflammation are regulated by meningeal lymphatic vasculature.
Nature neuroscience
Louveau A,Herz J,Alme MN,Salvador AF,Dong MQ,Viar KE,Herod SG,Knopp J,Setliff JC,Lupi AL,Da Mesquita S,Frost EL,Gaultier A,Harris TH,Cao R,Hu S,Lukens JR,Smirnov I,Overall CC,Oliver G,Kipnis J
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunocytochemistry
Mon Oct 01 00:00:00 UTC 2018
Role of Notch1 in the arterial specification and angiogenic potential of mouse embryonic stem cell-derived endothelial cells.
Stem cell research & therapy
Park JK,Lee TW,Do EK,Moon HJ,Kim JH
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Wed Jul 18 00:00:00 UTC 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Tue Mar 20 00:00:00 UTC 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Tue Mar 20 00:00:00 UTC 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Tue Mar 20 00:00:00 UTC 2018
Tumour ischaemia by interferon-γ resembles physiological blood vessel regression.
Nature
Kammertoens T,Friese C,Arina A,Idel C,Briesemeister D,Rothe M,Ivanov A,Szymborska A,Patone G,Kunz S,Sommermeyer D,Engels B,Leisegang M,Textor A,Fehling HJ,Fruttiger M,Lohoff M,Herrmann A,Yu H,Weichselbaum R,Uckert W,Hübner N,Gerhardt H,Beule D,Schreiber H,Blankenstein T
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunocytochemistry
Thu May 04 00:00:00 UTC 2017
Activation of the TGFβ pathway impairs endothelial to haematopoietic transition.
Scientific reports
Vargel Ö,Zhang Y,Kosim K,Ganter K,Foehr S,Mardenborough Y,Shvartsman M,Enright AJ,Krijgsveld J,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Fri Feb 19 00:00:00 UTC 2016
Suppression of ischemia in arterial occlusive disease by JNK-promoted native collateral artery development.
eLife
Ramo K,Sugamura K,Craige S,Keaney JF,Davis RJ
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Western Blot
Tue Aug 09 00:00:00 UTC 2016
EphrinB2 regulates the emergence of a hemogenic endothelium from the aorta.
Scientific reports
Chen II,Caprioli A,Ohnuki H,Kwak H,Porcher C,Tosato G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Thu Jun 02 00:00:00 UTC 2016
Endothelial fibroblast growth factor receptor signaling is required for vascular remodeling following cardiac ischemia-reperfusion injury.
American journal of physiology. Heart and circulatory physiology
House SL,Castro AM,Lupu TS,Weinheimer C,Smith C,Kovacs A,Ornitz DM
13-1441-82 was used in Flow Cytometry to suggest the potential for therapeutic benefit from activation of endothelial FGFR pathways following ischemic injury to the heart.
Tue Mar 01 00:00:00 UTC 2016
Angiopoietin 2 regulates the transformation and integrity of lymphatic endothelial cell junctions.
Genes & development
Zheng W,Nurmi H,Appak S,Sabine A,Bovay E,Korhonen EA,Orsenigo F,Lohela M,D'Amico G,Holopainen T,Leow CC,Dejana E,Petrova TV,Augustin HG,Alitalo K
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunofluorescence
Tue Jul 15 00:00:00 UTC 2014
Sox7 is regulated by ETV2 during cardiovascular development.
Stem cells and development
Behrens AN,Zierold C,Shi X,Ren Y,Koyano-Nakagawa N,Garry DJ,Martin CM
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Mon Sep 01 00:00:00 UTC 2014
The expression of Sox17 identifies and regulates haemogenic endothelium.
Nature cell biology
Clarke RL,Yzaguirre AD,Yashiro-Ohtani Y,Bondue A,Blanpain C,Pear WS,Speck NA,Keller G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Flow Cytometry
Wed May 01 00:00:00 UTC 2013
HIF-1α activation results in actin cytoskeleton reorganization and modulation of Rac-1 signaling in endothelial cells.
Cell communication and signaling : CCS
Weidemann A,Breyer J,Rehm M,Eckardt KU,Daniel C,Cicha I,Giehl K,Goppelt-Struebe M
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunofluorescence
Mon Oct 21 00:00:00 UTC 2013
ER71 specifies Flk-1+ hemangiogenic mesoderm by inhibiting cardiac mesoderm and Wnt signaling.
Blood
Liu F,Kang I,Park C,Chang LW,Wang W,Lee D,Lim DS,Vittet D,Nerbonne JM,Choi K
13-1441 was used in Flow cytometry/Cell sorting to investigate whether ER71 could modulate the haemangiogenic or cardiogenic outcome of the Flk-1(+) mesoderm.
Thu Apr 05 00:00:00 UTC 2012
Discontinuous LYVE-1 expression in corneal limbal lymphatics: dual function as microvalves and immunological hot spots.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Nakao S,Zandi S,Faez S,Kohno R,Hafezi-Moghadam A
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunofluorescence
Wed Feb 01 00:00:00 UTC 2012
Endothelial-myofibroblast transition contributes to the early development of diabetic renal interstitial fibrosis in streptozotocin-induced diabetic mice.
The American journal of pathology
Li J,Qu X,Bertram JF
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 13-1441-82) in Immunofluorescence
Thu Oct 01 00:00:00 UTC 2009