Product References
Nonredundancy of IL-1α and IL-1β is defined by distinct regulation of tissues orchestrating resistance versus tolerance to infection.
Science advances
Eislmayr K,Bestehorn A,Morelli L,Borroni M,Vande Walle L,Lamkanfi M,Kovarik P
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Fri Mar 04 00:00:00 UTC 2022
Mapping and role of T cell response in SARS-CoV-2-infected mice.
The Journal of experimental medicine
Zhuang Z,Lai X,Sun J,Chen Z,Zhang Z,Dai J,Liu D,Li Y,Li F,Wang Y,Zhu A,Wang J,Yang W,Huang J,Li X,Hu L,Wen L,Zhuo J,Zhang Y,Chen D,Li S,Huang S,Shi Y,Zheng K,Zhong N,Zhao J,Zhou D,Zhao J
45-0161 was used in Flow cytometry/Cell sorting to demonstrate cross-reactive T cell responses between SARS-CoV and SARS-CoV-2, but not MERS-CoV, in mice.
Mon Apr 05 00:00:00 UTC 2021
Aged hematopoietic stem cells are refractory to bloodborne systemic rejuvenation interventions.
The Journal of experimental medicine
Ho TT,Dellorusso PV,Verovskaya EV,Bakker ST,Flach J,Smith LK,Ventura PB,Lansinger OM,Hérault A,Zhang SY,Kang YA,Mitchell CA,Villeda SA,Passegué E
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Mon Jul 05 00:00:00 UTC 2021
Generation of a Broadly Useful Model for COVID-19 Pathogenesis, Vaccination, and Treatment.
Cell
Sun J,Zhuang Z,Zheng J,Li K,Wong RL,Liu D,Huang J,He J,Zhu A,Zhao J,Li X,Xi Y,Chen R,Alshukairi AN,Chen Z,Zhang Z,Chen C,Huang X,Li F,Lai X,Chen D,Wen L,Zhuo J,Zhang Y,Wang Y,Huang S,Dai J,Shi Y,Zheng K,Leidinger MR,Chen J,Li Y,Zhong N,Meyerholz DK,McCray PB,Perlman S,Zhao J
45-0161 was used in Flow cytometry/Cell sorting to describe a murine model of immediate utility to investigate COVID-19 pathogenesis and new therapies to treat it.
Thu Aug 06 00:00:00 UTC 2020
C/EBPα and GATA-2 Mutations Induce Bilineage Acute Erythroid Leukemia through Transformation of a Neomorphic Neutrophil-Erythroid Progenitor.
Cancer cell
Di Genua C,Valletta S,Buono M,Stoilova B,Sweeney C,Rodriguez-Meira A,Grover A,Drissen R,Meng Y,Beveridge R,Aboukhalil Z,Karamitros D,Belderbos ME,Bystrykh L,Thongjuea S,Vyas P,Nerlov C
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Mon May 11 00:00:00 UTC 2020
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Mon Mar 02 00:00:00 UTC 2020
tagHi-C Reveals 3D Chromatin Architecture Dynamics during Mouse Hematopoiesis.
Cell reports
Zhang C,Xu Z,Yang S,Sun G,Jia L,Zheng Z,Gu Q,Tao W,Cheng T,Li C,Cheng H
45-0161-82 was used in Flow Cytometry to introduce a low-input tagmentation-based Hi-C (tagHi-C) method to capture the chromatin structures of hundreds of cells.
Tue Sep 29 00:00:00 UTC 2020
Androgen Receptor Signaling Positively Regulates Monocytic Development.
Frontiers in immunology
Consiglio CR,Gollnick SO
45-0161-82 was used in Flow Cytometry to investigate the role of androgen signaling in monocytic differentiation with a genetic mouse model of myeloid androgen receptor (AR) knockout (MARKO) and pharmacological inhibition of AR.
Thu Apr 29 00:00:00 UTC 2021
LUBAC is essential for embryogenesis by preventing cell death and enabling haematopoiesis.
Nature
Peltzer N,Darding M,Montinaro A,Draber P,Draberova H,Kupka S,Rieser E,Fisher A,Hutchinson C,Taraborrelli L,Hartwig T,Lafont E,Haas TL,Shimizu Y,Böiers C,Sarr A,Rickard J,Alvarez-Diaz S,Ashworth MT,Beal A,Enver T,Bertin J,Kaiser W,Strasser A,Silke J,Bouillet P,Walczak H
45-0161-82 was used in Flow cytometry/Cell sorting to demonstrate that both linear ubiquitin chain assembly complex components HOIP and HOIL-1, are required for embryogenesis by preventing cell death.
Tue May 01 00:00:00 UTC 2018
miR-143/145 differentially regulate hematopoietic stem and progenitor activity through suppression of canonical TGFβ signaling.
Nature communications
Lam J,van den Bosch M,Wegrzyn J,Parker J,Ibrahim R,Slowski K,Chang L,Martinez-Høyer S,Condorelli G,Boldin M,Deng Y,Umlandt P,Fuller M,Karsan A
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Wed Jun 20 00:00:00 UTC 2018
The creatine kinase pathway is a metabolic vulnerability in EVI1-positive acute myeloid leukemia.
Nature medicine
Fenouille N,Bassil CF,Ben-Sahra I,Benajiba L,Alexe G,Ramos A,Pikman Y,Conway AS,Burgess MR,Li Q,Luciano F,Auberger P,Galinsky I,DeAngelo DJ,Stone RM,Zhang Y,Perkins AS,Shannon K,Hemann MT,Puissant A,Stegmaier K
45-0161 was used in Flow cytometry/Cell sorting to investigate the effect of MECOM on cellular metabolism.
Wed Mar 01 00:00:00 UTC 2017
Myeloid progenitor cluster formation drives emergency and leukaemic myelopoiesis.
Nature
Hérault A,Binnewies M,Leong S,Calero-Nieto FJ,Zhang SY,Kang YA,Wang X,Pietras EM,Chu SH,Barry-Holson K,Armstrong S,Göttgens B,Passegué E
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Thu Apr 06 00:00:00 UTC 2017
Ubiquitination of hnRNPA1 by TRAF6 links chronic innate immune signaling with myelodysplasia.
Nature immunology
Fang J,Bolanos LC,Choi K,Liu X,Christie S,Akunuru S,Kumar R,Wang D,Chen X,Greis KD,Stoilov P,Filippi MD,Maciejewski JP,Garcia-Manero G,Weirauch MT,Salomonis N,Geiger H,Zheng Y,Starczynowski DT
45-0161 was used in Flow cytometry/Cell sorting to examine the significance of ubiquitin signalling in coordinating RNA processing by Toll-like receptor pathways during an immune response and in premalignant haematologic diseases.
Wed Feb 01 00:00:00 UTC 2017
Rational combination of oncolytic vaccinia virus and PD-L1 blockade works synergistically to enhance therapeutic efficacy.
Nature communications
Liu Z,Ravindranathan R,Kalinski P,Guo ZS,Bartlett DL
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Mon Mar 27 00:00:00 UTC 2017
CD36 receptor regulates malaria-induced immune responses primarily at early blood stage infection contributing to parasitemia control and resistance to mortality.
The Journal of biological chemistry
Thylur RP,Wu X,Gowda NM,Punnath K,Neelgund SE,Febbraio M,Gowda DC
Fri Jun 02 00:00:00 UTC 2017
Gliotoxin Suppresses Macrophage Immune Function by Subverting Phosphatidylinositol 3,4,5-Trisphosphate Homeostasis.
mBio
Schlam D,Canton J,Carreño M,Kopinski H,Freeman SA,Grinstein S,Fairn GD
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Tue Apr 05 00:00:00 UTC 2016
Agonist anti-GITR monoclonal antibody and stereotactic radiation induce immune-mediated survival advantage in murine intracranial glioma.
Journal for immunotherapy of cancer
Patel MA,Kim JE,Theodros D,Tam A,Velarde E,Kochel CM,Francica B,Nirschl TR,Ghasemzadeh A,Mathios D,Harris-Bookman S,Jackson CC,Jackson C,Ye X,Tran PT,Tyler B,Coric V,Selby M,Brem H,Drake CG,Pardoll DM,Lim M
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Wed May 18 00:00:00 UTC 2016
Endothelial Cell-Selective Adhesion Molecule Expression in Hematopoietic Stem/Progenitor Cells Is Essential for Erythropoiesis Recovery after Bone Marrow Injury.
PloS one
Sudo T,Yokota T,Okuzaki D,Ueda T,Ichii M,Ishibashi T,Isono T,Habuchi Y,Oritani K,Kanakura Y
45-0161 was used in Flow cytometry/Cell sorting to determine the physiological importance of Endothelial Cell-Selective Adhesion Molecule (ESAM) in erythroid recovery, showing that ESAM is essential for erthryopoiesis recovery following bone marrow injury.
Tue Mar 14 00:00:00 UTC 2017
The GIY-YIG Type Endonuclease Ankyrin Repeat and LEM Domain-Containing Protein 1 (ANKLE1) Is Dispensable for Mouse Hematopoiesis.
PloS one
Braun J,Meixner A,Brachner A,Foisner R
45-0161 was used in Flow cytometry/Cell sorting to test whether ANKLE1 may contribute to Holliday junction resolution during DNA repair in mice.
Fri Aug 12 00:00:00 UTC 2016
Targeting megakaryocytic-induced fibrosis in myeloproliferative neoplasms by AURKA inhibition.
Nature medicine
Wen QJ,Yang Q,Goldenson B,Malinge S,Lasho T,Schneider RK,Breyfogle LJ,Schultz R,Gilles L,Koppikar P,Abdel-Wahab O,Pardanani A,Stein B,Gurbuxani S,Mullally A,Levine RL,Tefferi A,Crispino JD
45-0161 was used in Flow cytometry/Cell sorting to demonstrate that megakaryocytes drive fibrosis in PMF and that targeting them with AURKA inhibitors has the potential for therapeutic benefit.
Tue Dec 01 00:00:00 UTC 2015
Telomerase Deficiency Causes Alveolar Stem Cell Senescence-associated Low-grade Inflammation in Lungs.
The Journal of biological chemistry
Chen R,Zhang K,Chen H,Zhao X,Wang J,Li L,Cong Y,Ju Z,Xu D,Williams BR,Jia J,Liu JP
45-0161 was used in Flow cytometry/Cell sorting to elucidate the mechanisms by which TERC and TERT participate in lung diseases.
Fri Dec 25 00:00:00 UTC 2015
The miR-126-VEGFR2 axis controls the innate response to pathogen-associated nucleic acids.
Nature immunology
Agudo J,Ruzo A,Tung N,Salmon H,Leboeuf M,Hashimoto D,Becker C,Garrett-Sinha LA,Baccarini A,Merad M,Brown BD
45-0161 was used in Flow cytometry/Cell sorting to identify the miR-126-VEGFR2 axis as an important regulator of the innate response that operates through multiscale control of pDCs.
Wed Jan 01 00:00:00 UTC 2014
Diet-induced obesity promotes myelopoiesis in hematopoietic stem cells.
Molecular metabolism
Singer K,DelProposto J,Morris DL,Zamarron B,Mergian T,Maley N,Cho KW,Geletka L,Subbaiah P,Muir L,Martinez-Santibanez G,Lumeng CN
45-0161 was used in Flow cytometry/Cell sorting to demonstrate how metabolic signals potentiate leukocyte production and dietary priming of haematopoietic progenitors contributes to adipose tissue inflammation.
Mon Sep 01 00:00:00 UTC 2014
Wnt5a regulates hematopoietic stem cell proliferation and repopulation through the Ryk receptor.
Stem cells (Dayton, Ohio)
Povinelli BJ,Nemeth MJ
45-0161 was used in Flow cytometry/Cell sorting to study whether Ryk protein regulates the response of haematopoietic stem cells to Wnt5a.
Wed Jan 01 00:00:00 UTC 2014
Notch2-dependent classical dendritic cells orchestrate intestinal immunity to attaching-and-effacing bacterial pathogens.
Nature immunology
Satpathy AT,Briseño CG,Lee JS,Ng D,Manieri NA,Kc W,Wu X,Thomas SR,Lee WL,Turkoz M,McDonald KG,Meredith MM,Song C,Guidos CJ,Newberry RD,Ouyang W,Murphy TL,Stappenbeck TS,Gommerman JL,Nussenzweig MC,Colonna M,Kopan R,Murphy KM
45-0161 was used in Flow cytometry/Cell sorting to elucidate the cellular source of essential IL-23 production required for the mucosal response to attaching-and-effacing bacterial infection.
Sun Sep 01 00:00:00 UTC 2013
Bcl11a controls Flt3 expression in early hematopoietic progenitors and is required for pDC development in vivo.
PloS one
Wu X,Satpathy AT,Kc W,Liu P,Murphy TL,Murphy KM
45-0161 was used in Flow cytometry/Cell sorting to investigate the role of Bcl11a in the development of dendritic cells, showing that it controls Flt3 expression in early hematopoietic progenitors and is required for pDC development.
Fri Jan 10 00:00:00 UTC 2014
Zbtb46 expression distinguishes classical dendritic cells and their committed progenitors from other immune lineages.
The Journal of experimental medicine
Satpathy AT,KC W,Albring JC,Edelson BT,Kretzer NM,Bhattacharya D,Murphy TL,Murphy KM
45-0161 was used in Flow cytometry/Cell sorting to identify Zbtb46 as a transcription factor selectively expressed by classical DCs and their committed progenitors.
Mon Jun 04 00:00:00 UTC 2012
Morphine decreases bacterial phagocytosis by inhibiting actin polymerization through cAMP-, Rac-1-, and p38 MAPK-dependent mechanisms.
The American journal of pathology
Ninković J,Roy S
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Thu Mar 01 00:00:00 UTC 2012
Morphine decreases bacterial phagocytosis by inhibiting actin polymerization through cAMP-, Rac-1-, and p38 MAPK-dependent mechanisms.
The American journal of pathology
Ninković J,Roy S
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Thu Mar 01 00:00:00 UTC 2012
In vivo inhibition of c-MYC in myeloid cells impairs tumor-associated macrophage maturation and pro-tumoral activities.
PloS one
Pello OM,Chèvre R,Laoui D,De Juan A,Lolo F,Andrés-Manzano MJ,Serrano M,Van Ginderachter JA,Andrés V
Published figure using CD16/CD32 monoclonal antibody (Product # 45-0161-82) in Flow Cytometry
Mon Feb 25 00:00:00 UTC 2013
IFN-γ- and IL-10-expressing virus epitope-specific Foxp3(+) T reg cells in the central nervous system during encephalomyelitis.
The Journal of experimental medicine
Zhao J,Zhao J,Fett C,Trandem K,Fleming E,Perlman S
45-0161 was used in Flow cytometry/Cell sorting to show that pathogen epitope-specific Foxp3(+) T reg cells can be identified on the basis of cytokine production.
Mon Aug 01 00:00:00 UTC 2011
Regulation of granulocyte and macrophage populations of murine bone marrow cells by G-CSF and CD137 protein.
PloS one
Jiang D,Schwarz H
45-0161 was used in Flow cytometry/Cell sorting to confirm the role of CD137 ligand signalling in regulating myelopoiesis, and demonstrate that this growth promoting influence is restricted to the monocytic lineage.
Mon Dec 13 00:00:00 UTC 2010