Product References
CD304 + adipose tissue-derived mesenchymal stem cell abundance in autologous fat grafts highly correlates with improvement of localized pain syndromes.
Pain
Rezzonico Jost T,Lozito A,Mangani D,Raimondi A,Klinger F,Morone D,Klinger M,Grassi F,Vinci V
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
Stimulated Human Umbilical Cord Mesenchymal Stem Cells Enhance the Osteogenesis and Cranial Bone Regeneration through IL-32 Mediated P38 Signaling Pathway.
Stem cells international
Zhang X,Zheng Y,Wang G,Liu Y,Wang Y,Jiang X,Liang Y,Zhao X,Li P,Zhang Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2024
Adipose-derived stem cell exosomes promote tumor characterization and immunosuppressive microenvironment in breast cancer.
Cancer immunology, immunotherapy : CII
Zhu Q,Cao Y,Yuan J,Hu Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Jan 31 00:00:00 UTC 2024
Tumor-derived mesenchymal progenitor cell-related genes in the regulation of breast cancer proliferation.
Gland surgery
Chen Y,Zhu L,Wang Y,Hu J,Zhang H,Zhu J,Gong W,Liu X,Xiao F,Li X
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Mar 27 00:00:00 UTC 2024
Osimertinib Covalently Binds to CD34 and Eliminates Myeloid Leukemia Stem/Progenitor Cells.
Cancer research
Xia L,Liu JY,Yang MY,Zhang XH,Jiang Y,Yin QQ,Luo CH,Liu HC,Kang ZJ,Zhang CT,Gao BB,Zhou AW,Cai HY,Waller EK,Yan JS,Lu Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2024
Comparison of retinal degeneration treatment with four types of different mesenchymal stem cells, human induced pluripotent stem cells and RPE cells in a rat retinal degeneration model.
Journal of translational medicine
Liu Q,Liu J,Guo M,Sung TC,Wang T,Yu T,Tian Z,Fan G,Wu W,Higuchi A
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu Dec 14 00:00:00 UTC 2023
Exercise combined with administration of adipose-derived stem cells ameliorates neuropathic pain after spinal cord injury.
Neural regeneration research
Cheng X,Mao GP,Hu WJ,Yu ZR,Xu YY,Chen W,Li X,Zeng XL,Zhang WW,Chen JW,Wan Y,Wang L
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
Celecoxib induces adipogenic differentiation of hemangioma-derived mesenchymal stem cells through the PPAR-γ pathway in vitro and in vivo.
Experimental and therapeutic medicine
Wang Y,Kong L,Sun B,Cui J,Shen W
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Jun 01 00:00:00 UTC 2022
Mitochondrial oxidative phosphorylation is dispensable for survival of CD34+ chronic myeloid leukemia stem and progenitor cells.
Cell death & disease
Yan JS,Yang MY,Zhang XH,Luo CH,Du CK,Jiang Y,Dong XJ,Wang ZM,Yang LX,Li YD,Xia L,Lu Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Apr 20 00:00:00 UTC 2022
Human placental mesenchymal stem cells regulate inflammation via the NF‑κB signaling pathway.
Experimental and therapeutic medicine
Liu Y,Zhang X,Hu Y,Kang M,Wu Y,Wang Y,Deng C
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Nov 01 00:00:00 UTC 2022
Angiogenic stem cell delivery platform to augment post-infarction neovasculature and reverse ventricular remodeling.
Scientific reports
Shin HS,Thakore A,Tada Y,Pedroza AJ,Ikeda G,Chen IY,Chan D,Jaatinen KJ,Yajima S,Pfrender EM,Kawamura M,Yang PC,Wu JC,Appel EA,Fischbein MP,Woo Y,Shudo Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu Oct 20 00:00:00 UTC 2022
Decoding the pathogenesis of Diamond-Blackfan anemia using single-cell RNA-seq.
Cell discovery
Wang B,Wang C,Wan Y,Gao J,Ma Y,Zhang Y,Tong J,Zhang Y,Liu J,Chang L,Xu C,Shen B,Chen Y,Jiang E,Kurita R,Nakamura Y,Lim KC,Engel JD,Zhou J,Cheng T,Zhu X,Zhu P,Shi L
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue May 10 00:00:00 UTC 2022
LIM Mineralization Protein-1 Enhances the Committed Differentiation of Dental Pulp Stem Cells through the ERK1/2 and p38 MAPK Pathways and BMP Signaling.
International journal of medical sciences
Mu R,Chen B,Bi B,Yu H,Liu J,Li J,He M,Rong L,Liu B,Liu K,Zhu L,Shi X,Shuai Y,Jin L
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Mon Aug 08 00:00:00 UTC 2022
Astragaloside IV - mediated endothelial progenitor cell exosomes promote autophagy and inhibit apoptosis in hyperglycemic damaged endothelial cells via miR-21/PTEN axis.
Folia histochemica et cytobiologica
Xiong W,Huang XL,Li CY,Wang XL,Lan HW,Wang TT,Chen ZL,Yang QP,Hu AL,Xia YF,Zhu FR,Zhou ZZ
17-0349-42 was used in Flow cytometry/Cell sorting to demonstrate that AS-IV-mediated EPC-exos-miR-21 enhances autophagy and depresses apoptosis in HG-damaged endothelial cells via the miR-21/PTEN axis.
Mon Jan 02 00:00:00 UTC 2023
Single-cell Transcriptomic Analysis Reveals the Cellular Heterogeneity of Mesenchymal Stem Cells.
Genomics, proteomics & bioinformatics
Zhang C,Han X,Liu J,Chen L,Lei Y,Chen K,Si J,Wang TY,Zhou H,Zhao X,Zhang X,An Y,Li Y,Wang QF
17-0349-42 was used in Flow cytometry/Cell sorting to reveal five distinct subpopulations from bone marrow and Wharton's jelly.
Tue Feb 01 00:00:00 UTC 2022
Heterogeneous disease-propagating stem cells in juvenile myelomonocytic leukemia.
The Journal of experimental medicine
Louka E,Povinelli B,Rodriguez-Meira A,Buck G,Wen WX,Wang G,Sousos N,Ashley N,Hamblin A,Booth CAG,Roy A,Elliott N,Iskander D,de la Fuente J,Fordham N,O'Byrne S,Inglott S,Norfo R,Salio M,Thongjuea S,Rao A,Roberts I,Mead AJ
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
VAP-PLGA microspheres (VAP-PLGA) promote adipose-derived stem cells (ADSCs)-induced wound healing in chronic skin ulcers in mice via PI3K/Akt/HIF-1α pathway.
Bioengineered
Jiang W,Zhang J,Zhang X,Fan C,Huang J
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Dec 01 00:00:00 UTC 2021
Endothelial GABBR2 Regulates Post-ischemic Angiogenesis by Inhibiting the Glycolysis Pathway.
Frontiers in cardiovascular medicine
Zhang H,Zhou H,Yuan J,Nan Y,Liu J
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Aug 24 00:00:00 UTC 2021
Unsupervised discovery of dynamic cell phenotypic states from transmitted light movies.
PLoS computational biology
Nguyen P,Chien S,Dai J,Monnat RJ,Becker PS,Kueh HY
17-0349-42 was used in Flow Cytometry to identify distinct blood cell types in a heterogeneous dataset.
Wed Dec 01 00:00:00 UTC 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Aug 31 00:00:00 UTC 2021
Development, characterization, and hematopoietic differentiation of Griscelli syndrome type 2 induced pluripotent stem cells.
Stem cell research & therapy
Güney-Esken G,Erol ÖD,Pervin B,Gürhan Sevinç G,Önder T,Bilgiç E,Korkusuz P,Günel-Özcan A,Uçkan-Çetinkaya D,Aerts-Kaya F
17-0349-42 was used in Flow Cytometry to generate different iPSC clones from 3 GS-2 patients.
Thu May 13 00:00:00 UTC 2021
Development, characterization, and hematopoietic differentiation of Griscelli syndrome type 2 induced pluripotent stem cells.
Stem cell research & therapy
Güney-Esken G,Erol ÖD,Pervin B,Gürhan Sevinç G,Önder T,Bilgiç E,Korkusuz P,Günel-Özcan A,Uçkan-Çetinkaya D,Aerts-Kaya F
17-0349-42 was used in Flow Cytometry to generate different iPSC clones from 3 GS-2 patients.
Thu May 13 00:00:00 UTC 2021
Targeting the epichaperome as an effective precision medicine approach in a novel PML-SYK fusion acute myeloid leukemia.
NPJ precision oncology
Sugita M,Wilkes DC,Bareja R,Eng KW,Nataraj S,Jimenez-Flores RA,Yan L,De Leon JP,Croyle JA,Kaner J,Merugu S,Sharma S,MacDonald TY,Noorzad Z,Panchal P,Pancirer D,Cheng S,Xiang JZ,Olson L,Van Besien K,Rickman DS,Mathew S,Tam W,Rubin MA,Beltran H,Sboner A,Hassane DC,Chiosis G,Elemento O,Roboz GJ,Mosquera JM,Guzman ML
17-0349-42 was used in Flow Cytometry to develop a flow cytometry-based assay for evaluation and monitoring of epichaperome abundance at the single cell level.
Wed May 26 00:00:00 UTC 2021
Targeting the epichaperome as an effective precision medicine approach in a novel PML-SYK fusion acute myeloid leukemia.
NPJ precision oncology
Sugita M,Wilkes DC,Bareja R,Eng KW,Nataraj S,Jimenez-Flores RA,Yan L,De Leon JP,Croyle JA,Kaner J,Merugu S,Sharma S,MacDonald TY,Noorzad Z,Panchal P,Pancirer D,Cheng S,Xiang JZ,Olson L,Van Besien K,Rickman DS,Mathew S,Tam W,Rubin MA,Beltran H,Sboner A,Hassane DC,Chiosis G,Elemento O,Roboz GJ,Mosquera JM,Guzman ML
17-0349-42 was used in Flow Cytometry to develop a flow cytometry-based assay for evaluation and monitoring of epichaperome abundance at the single cell level.
Wed May 26 00:00:00 UTC 2021
Phenotypic and functional characterization of synovial fluid-derived fibroblast-like synoviocytes in rheumatoid arthritis.
Scientific reports
Køster D,Egedal JH,Lomholt S,Hvid M,Jakobsen MR,Müller-Ladner U,Eibel H,Deleuran B,Kragstrup TW,Neumann E,Nielsen MA
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Fri Nov 12 00:00:00 UTC 2021
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Oct 06 00:00:00 UTC 2020
Resveratrol rescues TNF‑α‑induced inhibition of osteogenesis in human periodontal ligament stem cells via the ERK1/2 pathway.
Molecular medicine reports
Yuan J,Wang X,Ma D,Gao H,Zheng D,Zhang J
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
Genome-Wide Transcriptional Regulation of the Long Non-coding RNA Steroid Receptor RNA Activator in Human Erythroblasts.
Frontiers in genetics
Sawaengdee W,Cui K,Zhao K,Hongeng S,Fucharoen S,Wongtrakoongate P
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Mon Sep 28 00:00:00 UTC 2020
Chemerin enhances the adhesion and migration of human endothelial progenitor cells and increases lipid accumulation in mice with atherosclerosis.
Lipids in health and disease
Jia J,Yu F,Xiong Y,Wei W,Ma H,Nisi F,Song X,Yang L,Wang D,Yuan G,Zhou H
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Sun Sep 20 00:00:00 UTC 2020
Apoptotic bodies derived from mesenchymal stem cells promote cutaneous wound healing via regulating the functions of macrophages.
Stem cell research & therapy
Liu J,Qiu X,Lv Y,Zheng C,Dong Y,Dou G,Zhu B,Liu A,Wang W,Zhou J,Liu S,Liu S,Gao B,Jin Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Fri Nov 27 00:00:00 UTC 2020
TNFAIP3 Plays a Role in Aging of the Hematopoietic System.
Frontiers in immunology
Smith MA,Culver-Cochran AE,Adelman ER,Rhyasen GW,Ma A,Figueroa ME,Starczynowski DT
17-0349-42 was used in Flow Cytometry to find that the expression of A20 is significantly reduced in aged hematopoietic stem and progenitor cells (HSPC) as compared to young HSPC.
Wed Apr 28 00:00:00 UTC 2021
The downregulated membrane expression of CD18 in CD34+ cells defines a primitive population of human hematopoietic stem cells.
Stem cell research & therapy
Mesa-Núñez C,Leon-Rico D,Aldea M,Damián C,Sanchez-Baltasar R,Sanchez R,Alberquilla O,Segovia JC,Bueren JA,Almarza E
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Apr 28 00:00:00 UTC 2020
Identification and characterization of the cellular subclones that contribute to the pathogenesis of mantle cell lymphoma.
Genes & diseases
Tang J,Zhang L,Zhou T,Sun Z,Kong L,Jing L,Xing H,Wu H,Liu Y,Zhou S,Li J,Chen M,Xu F,Tang J,Ma T,Hu M,Liu D,Guo J,Zhu X,Chen Y,Ye T,Wang J,Li X,Xing HR
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Sun Dec 01 00:00:00 UTC 2019
Overexpression of ERBB4 rejuvenates aged mesenchymal stem cells and enhances angiogenesis via PI3K/AKT and MAPK/ERK pathways.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Liang X,Ding Y,Lin F,Zhang Y,Zhou X,Meng Q,Lu X,Jiang G,Zhu H,Chen Y,Lian Q,Fan H,Liu Z
17-0349-42 was used in Flow Cytometry to investigate whether Erb-B2 receptor tyrosine kinase 4 ERBB4 rejuvenates aged mesenchymal stem cells MSC and how ERBB4 enhances therapeutic efficacy of aged MSC in treating myocardial infarction (MI).
Fri Mar 01 00:00:00 UTC 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu May 13 00:00:00 UTC 2021
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu May 02 00:00:00 UTC 2019
The suppressive effects of miR-508-5p on the odontogenic differentiation of human dental pulp stem cells by targeting glycoprotein non-metastatic melanomal protein B.
Stem cell research & therapy
Liu F,Wang X,Yang Y,Hu R,Wang W,Wang Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Jan 22 00:00:00 UTC 2019
Regnase-1-mediated post-transcriptional regulation is essential for hematopoietic stem and progenitor cell homeostasis.
Nature communications
Kidoya H,Muramatsu F,Shimamura T,Jia W,Satoh T,Hayashi Y,Naito H,Kunisaki Y,Arai F,Seki M,Suzuki Y,Osawa T,Akira S,Takakura N
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Mar 06 00:00:00 UTC 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Fri Dec 20 00:00:00 UTC 2019
Efficient gene transfer into T lymphocytes by fiber-modified human adenovirus 5.
BMC biotechnology
Lv Y,Xiao FJ,Wang Y,Zou XH,Wang H,Wang HY,Wang LS,Lu ZZ
17-0349 was used in Flow cytometry/Cell sorting to show the HAdV-11p fiber pseudotyped HAdV-5 could effectively transduce human T cells when human EF1a promoter was used to control the expression of transgene, suggesting its possible application in T cell immunocellular therapy.
Wed Apr 24 00:00:00 UTC 2019
Efficient gene transfer into T lymphocytes by fiber-modified human adenovirus 5.
BMC biotechnology
Lv Y,Xiao FJ,Wang Y,Zou XH,Wang H,Wang HY,Wang LS,Lu ZZ
17-0349 was used in Flow cytometry/Cell sorting to show the HAdV-11p fiber pseudotyped HAdV-5 could effectively transduce human T cells when human EF1a promoter was used to control the expression of transgene, suggesting its possible application in T cell immunocellular therapy.
Wed Apr 24 00:00:00 UTC 2019
Highly Efficient and Marker-free Genome Editing of Human Pluripotent Stem Cells by CRISPR-Cas9 RNP and AAV6 Donor-Mediated Homologous Recombination.
Cell stem cell
Martin RM,Ikeda K,Cromer MK,Uchida N,Nishimura T,Romano R,Tong AJ,Lemgart VT,Camarena J,Pavel-Dinu M,Sindhu C,Wiebking V,Vaidyanathan S,Dever DP,Bak RO,Laustsen A,Lesch BJ,Jakobsen MR,Sebastiano V,Nakauchi H,Porteus MH
17-0349 was used in Flow cytometry/Cell sorting to describe marker-free genome editing in human pluripotent stem cells.
Thu May 02 00:00:00 UTC 2019
A Human IPS Model Implicates Embryonic B-Myeloid Fate Restriction as Developmental Susceptibility to B Acute Lymphoblastic Leukemia-Associated ETV6-RUNX1.
Developmental cell
Böiers C,Richardson SE,Laycock E,Zriwil A,Turati VA,Brown J,Wray JP,Wang D,James C,Herrero J,Sitnicka E,Karlsson S,Smith AJH,Jacobsen SEW,Enver T
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2018
MLL leukemia induction by t(9;11) chromosomal translocation in human hematopoietic stem cells using genome editing.
Blood advances
Schneidawind C,Jeong J,Schneidawind D,Kim IS,Duque-Afonso J,Wong SHK,Iwasaki M,Breese EH,Zehnder JL,Porteus M,Cleary ML
17-0349 was used in Flow cytometry/Cell sorting to study the use of genome editing for the modelling of human acute MLL-rearranged leukaemia in vivo.
Tue Apr 24 00:00:00 UTC 2018
In Vitro Expansion and Characterization of Mesenchymal Stromal Cells from Peritoneal Dialysis Effluent in a Human Protein Medium.
Stem cells international
Han B,Zhou L,Guan Q,da Roza G,Wang H,Du C
17-0349 was used in Flow cytometry/Cell sorting to show that upon expansion in a human protein medium, pMSCs shows a differential MSC marker expression profile from those of bone marrow or adipose tissue-derived MSCs and could maintain the multipotency.
Wed Sep 30 00:00:00 UTC 2020
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
Improving hematopoietic recovery through modeling and modulation of the mesenchymal stromal cell secretome.
Stem cell research & therapy
Liu FD,Tam K,Pishesha N,Poon Z,Van Vliet KJ
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Oct 24 00:00:00 UTC 2018
Similar expression profiles in CD34+ cells from chronic phase chronic myeloid leukemia patients with and without deep molecular responses to nilotinib.
Oncotarget
Patel AB,Lange T,Pomicter AD,Conley CJ,Harrington CA,Reynolds KR,Kelley TW,O'Hare T,Deininger MW
17-0349 was used in Flow cytometry/Cell sorting to perform transcriptional profiling on CD34+ progenitor cells from newly diagnosed chronic phase chronic myeloid leukemia patients treated with nilotinib.
Tue Apr 03 00:00:00 UTC 2018
EDAG promotes the expansion and survival of human CD34+ cells.
PloS one
Zhao K,Zheng WW,Dong XM,Yin RH,Gao R,Li X,Liu JF,Zhan YQ,Yu M,Chen H,Ge CH,Ning HM,Yang XM,Li CY
17-0349 was used in Flow cytometry/Cell sorting to provide novel insight into EDAG in regulation of expansion and survival of human hematopoietic stem/progenitor cells.
Tue Feb 06 00:00:00 UTC 2018
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Tue Sep 04 00:00:00 UTC 2018
Rap1 deficiency-provoked paracrine dysfunction impairs immunosuppressive potency of mesenchymal stem cells in allograft rejection of heart transplantation.
Cell death & disease
Ding Y,Liang X,Zhang Y,Yi L,Shum HC,Chen Q,Chan BP,Fan H,Liu Z,Tergaonkar V,Qi Z,Tse HF,Lian Q
17-0349 was used in Flow cytometry/Cell sorting to indicate that Rap1 is essential to maintain the immunomodulatory function of MSCs.
Wed Mar 07 00:00:00 UTC 2018
Global survey of the immunomodulatory potential of common drugs.
Nature chemical biology
Vladimer GI,Snijder B,Krall N,Bigenzahn JW,Huber KVM,Lardeau CH,Sanjiv K,Ringler A,Berglund UW,Sabler M,de la Fuente OL,Knöbl P,Kubicek S,Helleday T,Jäger U,Superti-Furga G
17-0349 was used in Flow cytometry/Cell sorting to offer an attractive platform for the personalized identification and characterization of immunomodulatory therapeutics.
Thu Jun 01 00:00:00 UTC 2017
Bacillus anthracis lethal toxin negatively modulates ILC3 function through perturbation of IL-23-mediated MAPK signaling.
PLoS pathogens
Seshadri S,Allan DSJ,Carlyle JR,Zenewicz LA
17-0349 was used in Flow cytometry/Cell sorting to study the effects of MAPK signalling pathways in group 3 innate immune lymphocytes using Bacillus anthracis.
Sun Oct 01 00:00:00 UTC 2017
Activation of KLF1 Enhances the Differentiation and Maturation of Red Blood Cells from Human Pluripotent Stem Cells.
Stem cells (Dayton, Ohio)
Yang CT,Ma R,Axton RA,Jackson M,Taylor AH,Fidanza A,Marenah L,Frayne J,Mountford JC,Forrester LM
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2017
A high-yield isolation and enrichment strategy for human lung microvascular endothelial cells.
Pulmonary circulation
Gaskill C,Majka SM
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
Vitamin C plus hydrogel facilitates bone marrow stromal cell-mediated endometrium regeneration in rats.
Stem cell research & therapy
Yang H,Wu S,Feng R,Huang J,Liu L,Liu F,Chen Y
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Nov 21 00:00:00 UTC 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2017
Human Induced Pluripotent Stem Cell-Derived Macrophages Share Ontogeny with MYB-Independent Tissue-Resident Macrophages.
Stem cell reports
Buchrieser J,James W,Moore MD
17034942 was used in flow cytometry to use CRISPR-Cas9 to study human hematopoietic development
Tue Feb 14 00:00:00 UTC 2017
Etanercept-Synthesising Mesenchymal Stem Cells Efficiently Ameliorate Collagen-Induced Arthritis.
Scientific reports
Park N,Rim YA,Jung H,Kim J,Yi H,Kim Y,Jang Y,Jung SM,Lee J,Kwok SK,Park SH,Ju JH
17-0349 was used in Flow cytometry/Cell sorting to develop a novel method to produce synthetic biological drugs from engineered MSCs, and investigate the anti-arthritic effect of these cells in a collagen-induced arthritis mouse model.
Fri Jan 13 00:00:00 UTC 2017
UV-inactivated HSV-1 potently activates NK cell killing of leukemic cells.
Blood
Samudio I,Rezvani K,Shaim H,Hofs E,Ngom M,Bu L,Liu G,Lee JT,Imren S,Lam V,Poon GF,Ghaedi M,Takei F,Humphries K,Jia W,Krystal G
17-0349 was used in Flow cytometry/Cell sorting to support the preclinical development of UV light-inactivated HSV-1 as an adjuvant either alone or with IL-15, to treat acute myeloid leukemia.
Thu May 26 00:00:00 UTC 2016
Enforced Expression of HOXB4 in Human Embryonic Stem Cells Enhances the Production of Hematopoietic Progenitors but Has No Effect on the Maturation of Red Blood Cells.
Stem cells translational medicine
Jackson M,Ma R,Taylor AH,Axton RA,Easterbrook J,Kydonaki M,Olivier E,Marenah L,Stanley EG,Elefanty AG,Mountford JC,Forrester LM
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Jan 19 00:00:00 UTC 2016
Microvesicles from Mesenchymal Stromal Cells Are Involved in HPC-Microenvironment Crosstalk in Myelodysplastic Patients.
PloS one
Muntión S,Ramos TL,Diez-Campelo M,Rosón B,Sánchez-Abarca LI,Misiewicz-Krzeminska I,Preciado S,Sarasquete ME,de Las Rivas J,González M,Sánchez-Guijo F,Del Cañizo MC
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Jul 12 00:00:00 UTC 2016
A Simplified Method for the Aspiration of Bone Marrow from Patients Undergoing Hip and Knee Joint Replacement for Isolating Mesenchymal Stem Cells and In Vitro Chondrogenesis.
Bone marrow research
Juneja SC,Viswanathan S,Ganguly M,Veillette C
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Fri Apr 08 00:00:00 UTC 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
Pharmacological targeting of β-catenin in normal karyotype acute myeloid leukemia blasts.
Haematologica
Griffiths EA,Golding MC,Srivastava P,Povinelli BJ,James SR,Ford LA,Wetzler M,Wang ES,Nemeth MJ
17-0349 was used in Flow cytometry/Cell sorting to suggest a potential clinical benefit for WNT suppression.
Sun Feb 01 00:00:00 UTC 2015
Functional plasticity of the N-methyl-d-aspartate receptor in differentiating human erythroid precursor cells.
American journal of physiology. Cell physiology
Hänggi P,Telezhkin V,Kemp PJ,Schmugge M,Gassmann M,Goede JS,Speer O,Bogdanova A
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Mon Jun 15 00:00:00 UTC 2015
Differences in the Osteogenic Differentiation Capacity of Omental Adipose-Derived Stem Cells in Obese Patients With and Without Metabolic Syndrome.
Endocrinology
Oliva-Olivera W,Leiva Gea A,Lhamyani S,Coín-Aragüez L,Alcaide Torres J,Bernal-López MR,García-Luna PP,Morales Conde S,Fernández-Veledo S,El Bekay R,Tinahones FJ
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Tue Dec 01 00:00:00 UTC 2015
Oncogenic NRAS hyper-activates multiple pathways in human cord blood stem/progenitor cells and promotes myelomonocytic proliferation in vivo.
American journal of translational research
Wang T,Li C,Xia C,Dong Y,Yang D,Geng Y,Cai J,Zhang J,Zhang X,Wang J
17-0349 was used in Flow cytometry/Cell sorting to evaluate the effects of the NRASG12D mutation on human haematopoiesis and leukaemogenesis in vitro and in vivo.
Tue Dec 22 00:00:00 UTC 2015
Reprogramming human endothelial cells to haematopoietic cells requires vascular induction.
Nature
Sandler VM,Lis R,Liu Y,Kedem A,James D,Elemento O,Butler JM,Scandura JM,Rafii S
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu Jul 17 00:00:00 UTC 2014
JAK2/STAT5 inhibition by nilotinib with ruxolitinib contributes to the elimination of CML CD34+ cells in vitro and in vivo.
Blood
Gallipoli P,Cook A,Rhodes S,Hopcroft L,Wheadon H,Whetton AD,Jørgensen HG,Bhatia R,Holyoake TL
Published figure using CD34 monoclonal antibody (Product # 17-0349-42) in Flow Cytometry
Thu Aug 28 00:00:00 UTC 2014
Comparison of human adipose-derived stem cells isolated from subcutaneous, omental, and intrathoracic adipose tissue depots for regenerative applications.
Stem cells translational medicine
Russo V,Yu C,Belliveau P,Hamilton A,Flynn LE
17-0349 was used in Flow cytometry/Cell sorting to determine that depot selection is an important factor to consider when applying ASCs in tissue-specific cell-based regenerative therapies.
Sat Feb 01 00:00:00 UTC 2014
Central role of conventional dendritic cells in regulation of bone marrow release and survival of neutrophils.
Journal of immunology (Baltimore, Md. : 1950)
Jiao J,Dragomir AC,Kocabayoglu P,Rahman AH,Chow A,Hashimoto D,Leboeuf M,Kraus T,Moran T,Carrasco-Avino G,Friedman SL,Merad M,Aloman C
17-0349 was used in Flow cytometry/Cell sorting to investigate the role of dendritic cells in neutrophil homeostasis, showing a central role in regulation of bone marrow release and survival of neutrophils.
Tue Apr 01 00:00:00 UTC 2014
Interaction of mesenchymal stem cells with fibroblast-like synoviocytes via cadherin-11 promotes angiogenesis by enhanced secretion of placental growth factor.
Journal of immunology (Baltimore, Md. : 1950)
Park SJ,Kim KJ,Kim WU,Cho CS
17-0349 was used in Flow cytometry/Cell sorting to study how cadherin-11 allows mesenchymal stem cells and fibroblast-like synoviocytes to interact, promoting angiogenesis.
Tue Apr 01 00:00:00 UTC 2014
Generation of X-CGD cells for vector evaluation from healthy donor CD34(+) HSCs by shRNA-mediated knock down of gp91(phox).
Molecular therapy. Methods & clinical development
Brendel C,Kaufmann KB,Krattenmacher A,Pahujani S,Grez M
17-0349 was used in Flow cytometry/Cell sorting to demonstrate a method for overcoming the paucity of X-CGD CD34+ cells, using a lentiviral mediated gp91phox knock down vector that abrogates gp91phox expression as well as NADPH oxidase activity in the myeloid progeny.
Wed May 27 00:00:00 UTC 2015
FOXF1 mediates mesenchymal stem cell fusion-induced reprogramming of lung cancer cells.
Oncotarget
Wei HJ,Nickoloff JA,Chen WH,Liu HY,Lo WC,Chang YT,Yang PC,Wu CW,Williams DF,Gelovani JG,Deng WP
17-0349 was used in Flow cytometry/Cell sorting to study the effect of fusion of mesenchymal stem cells with lung cancer cells, showing that fusion is mediated by FOXF1 and leads to a more benign state due to complementation of tumorigenic defects such as the restoration of p21 function.
Wed Oct 15 00:00:00 UTC 2014
Distinct fibroblast lineages determine dermal architecture in skin development and repair.
Nature
Driskell RR,Lichtenberger BM,Hoste E,Kretzschmar K,Simons BD,Charalambous M,Ferron SR,Herault Y,Pavlovic G,Ferguson-Smith AC,Watt FM
17-0349 was used in Flow cytometry/Cell sorting to explain why wounding is linked to formation of ECM-rich scar tissue that lacks hair follicles.
Thu Dec 12 00:00:00 UTC 2013
Terminal differentiation and loss of tumorigenicity of human cancers via pluripotency-based reprogramming.
Oncogene
Zhang X,Cruz FD,Terry M,Remotti F,Matushansky I
17-0349 was used in Flow cytometry/Cell sorting to investigate the potential of nuclear reprogramming as a broadly applicable therapeutic strategy for the treatment of cancer.
Thu May 02 00:00:00 UTC 2013
Human cardiac stem cells isolated from atrial appendages stably express c-kit.
PloS one
He JQ,Vu DM,Hunt G,Chugh A,Bhatnagar A,Bolli R
17-0349 was used in Flow cytometry/Cell sorting to investigate how a higher percentage of relative pure human CSCs with stable expression of c-kit+ could be obtained from atrial appendage specimens.
Mon Apr 09 00:00:00 UTC 2012