Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This antibody is a mouse monoclonal antibody raised against residues near the C terminus of human CDC6.
Immunogen sequence: QDLKKELKG FKTIMLNCMS LRTAQAVFPA IAQEICQEEV SRPAGKDMMR KLEKHMTAEK GPMIVLVLDE MDQLDSKGQD VLYTLFEWPW LSNSHLVLIG IANTLDLTDR ILPRLQAREK CKPQLLNFPP YTRNQIVTIL QDRLNQVSRD QVLDNAAVQF CARKVSAVSG DVRKALDVCR RAIEIVESDV KSQTILKPLS ECKSPSEPLI PKRVGLIHIS QVISEIDGNR MTLSQEGAQD SFPLQQKILV CSLMLLIRQL KIKEVTLGKL YEAYSKVCRK QQVAAVDQSE CLSLSGLLEA RGILGLKRNK ETRLTKVFFK IEEKEIEHAL KDKALIGNIL ATGLP (217-560 aa encoded by BC025232)
The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC18(S.pombe); Cdc18-related protein; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6 homolog; HsCdc18; HsCDC6; p62(cdc6)
Gene Aliases: CDC18L; CDC6; HsCDC18; HsCDC6
UniProt ID: (Human) Q99741, (Mouse) O89033
Entrez Gene ID: (Human) 990, (Mouse) 23834, (Rat) 360621
Molecular Function: replication origin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support