Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CORIN is a member of the type II transmembrane serine protease class of the trypsin superfamily. Members of this family are composed of multiple structurally distinct domains. CORIN converts pro-atrial natriuretic peptide to biologically active atrial natriuretic peptide, a cardiac hormone that regulates blood volume and pressure. This protein may also function as a pro-brain-type natriuretic peptide convertase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Atrial natriuretic peptide-converting enzyme; Corin; EC 3.4.21; Heart-specific serine proteinase ATC2; Low density lipoprotein receptor-related protein 4; low density lipoprotein-related protein 4; MGC119742; pro-ANP-convertase; Pro-ANP-converting enzyme; PRSC; serine peptidase; serine protease; Transmembrane protease serine 10
Gene Aliases: ATC2; AV273130; CORIN; CRN; Lrp4; PEE5; TMPRSS10
UniProt ID: (Human) Q9Y5Q5, (Mouse) Q9Z319, (Rat) Q80YN4
Entrez Gene ID: (Human) 10699, (Mouse) 53419, (Rat) 289596
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support