Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LWKGPSFDV QVGLHELLGH GSGKLFVQDE KGAFNFDQET VINPETGEQI QSWYRSGETW DSKFSTIASS YEECRAESVG LYLCLHPQVL EIFGFEGADA EDVIYVNWLN MVRAGLLALE FYTPEAFNWR QAHMQARFVI LRVLLEAGEG LVTITPTTGS DGRPDARVRL DRSKIRSVGK PALERFLRRL QVLKSTGDVA GGRALYEGYA TVTDAPPECF LTLRDTVLLR KESRKLIVQP NTRLEGSDVQ LLEYEASAAG LIRSFSERFP EDGPELEEIL TQLATADARF WKGPSEAPSG QA (437-737 aa encoded by BC007221)
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dipeptidyl aminopeptidase III; Dipeptidyl arylamidase III; Dipeptidyl peptidase 3; Dipeptidyl peptidase III; dipeptidyl-peptidase 3; dipeptidyl-peptidase III; dipeptidylpeptidase III; DPP III; Enkephalinase B; hCG_2016942
Gene Aliases: 4930533O14Rik; C86324; DPP3; DPPIII
UniProt ID: (Human) Q9NY33, (Mouse) Q99KK7
Entrez Gene ID: (Human) 10072, (Mouse) 75221
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support