Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: DNYEDDHFK SSHSNQTNHK PSPDQIIQPL LELDQNRSKL KLYIGHLTTL CHDRDPLILR GLTPPASYNL DDDQAAWENE LQKMTRGQLQ DELEKGERDN AELQEFANAI LQQIADHCPD ILEQVVNALE ESS (985-1116 aa encoded by B C132782)
The protein encoded by this gene is a member of a family of RIM-binding proteins. RIMs are active zone proteins that regulate neurotransmitter release. This gene has been found fused to the receptor-type tyrosine kinase gene RET by gene rearrangement due to the translocation t(10;12)(q11;p13). Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELKS/Rab6-interacting/CAST family member 1; ERC-1; FLJ31750; MGC12974; OTTHUMP00000160633; RAB6 interacting protein 2; Rab6-interacting protein 2
Gene Aliases: Cast2; ELKS; ERC-1; ERC1; KIAA1081; RAB6IP2
UniProt ID: (Human) Q8IUD2
Entrez Gene ID: (Human) 23085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support