Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSEGVDLIDI YADEEFNQDP EFNNTDQIDL YDDVLTATSQ PSDDRSSSTE PPPPVRQEPS PKPNNKTPAI LYTYSGLRNR RAAVYVGSFS WWTTDQQLIQ VIRSIGVYDV VELKFAENRA NGQSKGYAEV VVASENSVHK LLELLPGKVL NGEKVDVRPA TRQNLSQFEA QARKRECVRV PRGGIPPRAH SRDSSDSADG RATPSENLVP SSARVDKPPS VLPYFNRPPS ALPLMGLPPP PIPPPPPLSS SFGVPPPPPG IHYQHLMPPP PRLPPHLAVP PPGAIPPALH LNPAFFPPPN ATVGPPPDTY MKASAPYNHH GSRDSGPPPS TVSEAEFEDI MKRNRAISSS AISKAVSGAS AGDYSDAIET LLTAIAVIKQ SRVANDERCR VLISSLKDCL HGIEAKSYSV GASGSSSRKR HRSRERSPSR SRESSRRHRD LLHNEDRHDD YFQERNREHE RHRDRERDRH H
FLJ12529 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFIm59; cleavage and polyadenylation specific factor 7, 59kDa; Cleavage and polyadenylation specificity factor 59 kDa subunit; Cleavage and polyadenylation specificity factor subunit 7; Cleavage factor Im complex 59 kDa subunit; CPSF 59 kDa subunit; FLJ12529; pre mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor I, 59 kDa subunit; Pre-mRNA cleavage factor Im 59 kDa subunit
Gene Aliases: CFIm59; CPSF7
UniProt ID: (Human) Q8N684
Entrez Gene ID: (Human) 79869
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support