Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTLSTEMSDA SGLAEETDID VVGEGEDEED EEEEDDDEGG GGGPRLAVPA QRRRRRRSYA GEDELEDLEE EEDDDDILLA PPAGGSPAPP
This intronless gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in tumor formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Forkhead box protein D1; Forkhead, drosophila, homolog-like 8; forkhead-like 8; forkhead-related activator 4; Forkhead-related protein FKHL8; Forkhead-related transcription factor 4; FREAC-4
Gene Aliases: FKHL8; FOXD1; FREAC-4; FREAC4
UniProt ID: (Human) Q16676
Entrez Gene ID: (Human) 2297
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support