Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: IVTADGADFP MDSSNGNCRG ASSERCKCKP IRATQKTYFR NNYNYVIRAK VKEIKTKCHD VTAVVEVKEI LKSSLVNIPR DTVNLYTSSG CLCPPLNVNE EYIIMGYEDE ERSRLLLVEG SIAEKWKDRL GKKVKRWDMK LRHLGLSKSD SSNSDSTQSQ KSGRNSNPRQ ARN
The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FREOS1; Frezzled; Fritz; frizzled homolog-related; Frizzled-related protein 1; FrzB-1; FRZB1sFRP-3; Secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; sFRP-3; SFRP3frezzled
Gene Aliases: FIZ; FRE; frezzled; FRITZ; FRP; FRP-3; FRZB; FRZB-1; FRZB-PEN; FRZB1; FZRB; hFIZ; OS1; SFRP3; SRFP3
UniProt ID: (Human) Q92765, (Mouse) P97401
Entrez Gene ID: (Human) 2487, (Mouse) 20378
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support