Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody specifically recognizes the 75 kDa human HBS1L protein.
Immunogen sequence: SRRDKPSVE PVEEYDYEDL KESSNSVSNH QLSGFDQARL YSCLDHMREV LGDAVPDEIL IEAVLKNKFD VQKALSGVLE QDRVQSLKDK NEATVSTGKI AKGKPVDSQT SRSESEIVPK VAKMTVSGKK QTMGFEVPGV SSEENGHSFH TPQKGPPIED AIASSDVLET ASKSANPPHT IQASEEQSST PAPVKKSGKL RQQIDVKAEL EKRQGGKQLL N (43-262 aa encoded by BC001465)
This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eRF3 family member; ERF3-similar protein; ERFS; eukaryotic release factor 3; HBS1-like protein; HBS1-like translational GTPase; Hsp70 subfamily B suppressor 1-like protein
Gene Aliases: 2810035F15Rik; AI326327; EF-1a; eRF3c; ERFS; HBS1; HBS1L; HSPC276; KIAA1038
UniProt ID: (Human) Q9Y450, (Rat) Q6AXM7, (Mouse) Q69ZS7
Entrez Gene ID: (Human) 10767, (Rat) 293408, (Mouse) 56422
Molecular Function: translation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support