Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
For maximum recovery of product, centrifuge the vial prior to removing the cap.
The Slo3 channel is a novel potassium channel abundantly expressed in mammalian speramtocytes- tests have shown that it is expressed in both mouse and human testis. It represents a new and unique type of potassium channel that is regulated by both intracellular pH and membrane voltage. Because of its sensitivity to both pH and voltage, Slo3 may play a role in alkalization-mediated K(+) fluxes associated with sperm capacitation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-activated potassium channel subunit alpha-3; Calcium-activated potassium channel, subfamily M subunit alpha-3; KSper; mSlo3; pH-sensitive maxi potassium channel; Pore-forming subunit of the sperm-specific alkalization activated K(+) current; Potassium channel subfamily U member 1; potassium large conductance pH-sensitive channel, subfamily M, alpha member 3; Slowpoke homolog 3
Gene Aliases: Kcnma3; Kcnu1; Ksper; mSlo3; Slo3
UniProt ID: (Mouse) O54982
Entrez Gene ID: (Mouse) 16532
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support