Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Ki-67 Antibodies
            Now also available in recombinant format (Product # 740008T)

            Invitrogen

            Ki-67 Monoclonal Antibody (SolA15), eBioscience™

            Advanced Verification
            87 Published Figures
            414 References
            View all (222) Ki-67 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 14-5698-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            435.00
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite Ki-67 Monoclonal Antibody (SolA15), eBioscience™

            • Antibody Testing Data (8)
            • Published Figures (87)
            • Advanced Verification (2)
            Application
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 97

            Ki-67 Antibody (14-5698-82) in ICC/IF

            Immunofluorescence analysis of Ki67 was performed using 70% confluent log phase HeLa cells serum starved for 36 Hrs followed by serum release for 6 Hrs. The cells were fixed with 4% Paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 10 minutes at room temperature. The cells were labeled with Ki-67 Monoclonal Antibody (SolA15), eBioscience™ (Product # 14-5698-82) at 5 µg/mL in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Goat anti-Rat IgG (H+L) Cross-... View More
            View Product

            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            Ki-67 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            Ki-67 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (Paraffin) (ICC/IF, IHC (P))
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
            Ki-67 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Ki-67 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunocytochemistry (ICC/IF)
            Ki-67 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed) (IHC (P), IHC (PFA))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Ki-67 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
            Ki-67 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
            Ki-67 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            Ki-67 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            Ki-67 Antibody in Immunohistochemistry, Immunohistochemistry (Frozen) (IHC, IHC (F))
            Ki-67 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed) (IHC, IHC (PFA))
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody in Immunohistochemistry (IHC)
            Ki-67 Antibody
            Ki-67 Antibody

            Product Details

            14-5698-82

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            -
            View 2 publications 2 publications

            Immunohistochemistry (IHC)

            -
            View 199 publications 199 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            2.5-5 µg/mL
            View 17 publications 17 publications

            Immunohistochemistry (PFA fixed) (IHC (PFA))

            -
            View 5 publications 5 publications

            Immunohistochemistry (Frozen) (IHC (F))

            5 µg/mL
            View 11 publications 11 publications

            Immunocytochemistry (ICC/IF)

            1:100
            View 85 publications 85 publications

            Flow Cytometry (Flow)

            -
            View 93 publications 93 publications

            Immunoprecipitation (IP)

            -
            View 1 publication 1 publication

            Functional Assay (Functional)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Dog, Cynomolgus monkey, Human, Mouse, Non-human primate, Rat

            Published species

            Dog, Ferret, Human, Mouse, Mustelid, Pig, Rat

            Host/Isotype

            Rat / IgG2a, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            SolA15

            Conjugate

            Unconjugated
            • Alexa Fluor 488 
            • Alexa Fluor 532 
            • Alexa Fluor 660 
            • Alexa Fluor 700 
            • Alexa Fluor Plus 594 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 395
            • Brilliant UV 496
            • Brilliant UV 563
            • Brilliant UV 615
            • Brilliant UV 661
            • Brilliant UV 737
            • Brilliant UV 805
            • Brilliant Violet 421
            • Brilliant Violet 480
            • Brilliant Violet 605
            • Brilliant Violet 650
            • Brilliant Violet 711
            • Brilliant Violet 786
            • eFluor 450
            • eFluor 506
            • eFluor 570
            • eFluor 615
            • eFluor 660
            • FITC
            • PE 
            • PE-Cyanine5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-eFluor 710
            • Request custom conjugation

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_10854564

            Product Specific Information

            Description: The monoclonal antibody SolA15 recognizes mouse and rat Ki-67, a 300 kDa nuclear protein. Ki-67 is present during all active phases of the cell cycle (G1, S, G2, and mitosis), but is absent from resting cells (G0). Ki-67 is detected within the nucleus during interphase but redistributes to the chromosomes during mitosis. Ki-67 is used as a marker for determining the growth fraction of a given population of cells. In studies of tumor cells, the "Ki-67 labeling index" refers to the number of Ki-67 positive cells within the population and this is used to predict outcome of particular cancer types. Ki-67 has been shown to interact with the DNA-bound protein chromobox protein homolog 3 (CBX3) (heterochromatin).

            The SolA15 antibody also recognizes human, non-human primate and canine Ki-67.

            Applications Reported: This SolA15 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded (FFPE) tissue sections and frozen (paraformaldehyde fixed) sections and immunocytochemistry (ICC).

            Applications Tested: This SolA15 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded mouse skin with high or low pH antigen retrieval and immunocytochemistry on fixed and permeabilized C2C12 cells at less than or equal to 5 µg/mL. It is recommended that this antibody be carefully titrated for optimal performance in the assay of interest.

            Purity: Greater than 90%, as determined by SDS-PAGE.

            Aggregation: Less than 10%, as determined by HPLC.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            Ki-67 is a nuclear protein that is expressed during various stages in the cell cycle, particularly during late G1, S, G2, and M phases. The protein has a forkhead associated domain (FHA) through which it associates with euchromatin at the perichromosomal layer, the centromeric heterochromatin, and the nucleolus. Ki-67 is shown to have a cell cycle dependent topographical distribution with perinucleolar expression at G1, expression in the nuclear matrix at G2, and expression on the chromosomes during M phase. Ki-67 is commonly used as a proliferation marker because it is not detected in G0 cells, but increases steadily from G1 through mitosis. Ki-67 antibodies are useful in establishing the cell growing fraction in neoplasms. In neoplastic tissues, the prognostic value is comparable to the tritiated thymidine-labelling index. The correlation between low Ki-67 index and histologically low-grade tumors is strong. Ki-67 is routinely used as a neuronal marker of cell cycling and proliferation.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Bioinformatics

            Protein Aliases: Antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; unnamed protein product

            View more View less

            Gene Aliases: D630048A14Rik; Ki-67; Ki67; KIA; MIB-; MIB-1; MKI67; PPP1R105

            View more View less

            UniProt ID: (Human) P46013, (Mouse) E9PVX6

            View more View less

            Entrez Gene ID: (Dog) 100686578, (Human) 4288, (Cynomolgus monkey) 102135895, (Rat) 291234, (Mouse) 17345

            View more View less

            Function(s)

            protein binding ATP binding protein C-terminus binding poly(A) RNA binding molecular_function

            Process(es)

            DNA metabolic process meiotic nuclear division cell proliferation response to organic cyclic compound hyaluronan metabolic process organ regeneration cellular response to heat hepatocyte proliferation cholangiocyte proliferation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-5cpsf:80/100.66.75.64:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0