Product References
Poly-L-ornithine blocks the inhibitory effects of fibronectin on oligodendrocyte differentiation and promotes myelin repair.
Neural regeneration research
Xiong YJ,Soomro SH,Huang ZH,Yu PP,Ping J,Fu H
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Sat Apr 01 00:00:00 UTC 2023
Transformation of primary murine peritoneal mast cells by constitutive KIT activation is accompanied by loss of Cdkn2a/Arf expression.
Frontiers in immunology
Capellmann S,Sonntag R,Schüler H,Meurer SK,Gan L,Kauffmann M,Horn K,Königs-Werner H,Weiskirchen R,Liedtke C,Huber M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Apr 19 00:00:00 UTC 2023
B-cell antigen receptor expression and phosphatidylinositol 3-kinase signaling regulate genesis and maintenance of mouse chronic lymphocytic leukemia.
Haematologica
Schmid VK,Khadour A,Ahmed N,Brandl C,Nitschke L,Rajewsky K,Jumaa H,Hobeika E
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
Inhibition of LncRNA Vof-16 expression promotes nerve regeneration and functional recovery after spinal cord injury.
Neural regeneration research
Zhang XM,Zeng LN,Yang WY,Ding L,Chen KZ,Fu WJ,Zeng SQ,Liang YR,Chen GH,Wu HF
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Sat Jan 01 00:00:00 UTC 2022
Application of an instructive hydrogel accelerates re-epithelialization of xenografted human skin wounds.
Scientific reports
Sparks HD,Mandla S,Vizely K,Rosin N,Radisic M,Biernaskie J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Sat Aug 20 00:00:00 UTC 2022
Application of an instructive hydrogel accelerates re-epithelialization of xenografted human skin wounds.
Scientific reports
Sparks HD,Mandla S,Vizely K,Rosin N,Radisic M,Biernaskie J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Sat Aug 20 00:00:00 UTC 2022
Visualization of individual cell division history in complex tissues using iCOUNT.
Cell stem cell
Denoth-Lippuner A,Jaeger BN,Liang T,Royall LN,Chie SE,Buthey K,Machado D,Korobeynyk VI,Kruse M,Munz CM,Gerbaulet A,Simons BD,Jessberger S
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Thu Nov 04 00:00:00 UTC 2021
Epicardial placement of human MSC-loaded fibrin sealant films for heart failure: Preclinical efficacy and mechanistic data.
Molecular therapy : the journal of the American Society of Gene Therapy
Fields L,Ito T,Kobayashi K,Ichihara Y,Podaru MN,Hussain M,Yamashita K,Machado V,Lewis-McDougall F,Suzuki K
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Wed Aug 04 00:00:00 UTC 2021
Ageing promotes early T follicular helper cell differentiation by modulating expression of RBPJ.
Aging cell
Webb LMC,Fra-Bido S,Innocentin S,Matheson LS,Attaf N,Bignon A,Novarino J,Fazilleau N,Linterman MA
56-5698-82 was used in Flow Cytometry to show increased formation of Tfh precursors (pre-Tfh) but no associated increase in germinal centre (GC)-Tfh cells in aged mice, suggesting age-driven promotion of only early Tfh cell differentiation.
Fri Jan 01 00:00:00 UTC 2021
Heterogeneity of astrocytes: Electrophysiological properties of juxtavascular astrocytes before and after brain injury.
Glia
Götz S,Bribian A,López-Mascaraque L,Götz M,Grothe B,Kunz L
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
Heterogeneity of astrocytes: Electrophysiological properties of juxtavascular astrocytes before and after brain injury.
Glia
Götz S,Bribian A,López-Mascaraque L,Götz M,Grothe B,Kunz L
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
Environmental oxygen regulates astrocyte proliferation to guide angiogenesis during retinal development.
Development (Cambridge, England)
Perelli RM,O'Sullivan ML,Zarnick S,Kay JN
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Sat May 01 00:00:00 UTC 2021
HDAC1 Regulates Neuronal Differentiation.
Frontiers in molecular neuroscience
Nieto-Estevez V,Changarathil G,Adeyeye AO,Coppin MO,Kassim RS,Zhu J,Hsieh J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry (PFA fixed)
Tue Feb 01 00:00:00 UTC 2022
Macrophage LAMTOR1 Deficiency Prevents Dietary Obesity and Insulin Resistance Through Inflammation-Induced Energy Expenditure.
Frontiers in cell and developmental biology
Ying L,Zhang M,Ma X,Si Y,Li X,Su J,Yin J,Bao Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Jun 08 00:00:00 UTC 2021
Transcription factor old astrocyte specifically induced substance is a novel regulator of kidney fibrosis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Yamamoto A,Morioki H,Nakae T,Miyake Y,Harada T,Noda S,Mitsuoka S,Matsumoto K,Tomimatsu M,Kanemoto S,Tanaka S,Maeda M,Conway SJ,Imaizumi K,Fujio Y,Obana M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
HOXA11 plays critical roles in disease progression and response to cytarabine in AML.
Oncology reports
Fu JF,Shih LY,Yen TH
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Jul 01 00:00:00 UTC 2021
Thrombospondin-2 spatiotemporal expression in skeletal fractures.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Zondervan RL,Jenkins DC,Reicha JD,Hankenson KD
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri Jan 01 00:00:00 UTC 2021
MicroRNA-384 inhibits nasopharyngeal carcinoma growth and metastasis via binding to Smad5 and suppressing the Wnt/β-catenin axis.
Cytotechnology
Zeng X,Liao H,Wang F
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Apr 01 00:00:00 UTC 2021
Requirement of DNMT1 to orchestrate epigenomic reprogramming for NPM-ALK-driven lymphomagenesis.
Life science alliance
Redl E,Sheibani-Tezerji R,Cardona CJ,Hamminger P,Timelthaler G,Hassler MR,Zrimšek M,Lagger S,Dillinger T,Hofbauer L,Draganić K,Tiefenbacher A,Kothmayer M,Dietz CH,Ramsahoye BH,Kenner L,Bock C,Seiser C,Ellmeier W,Schweikert G,Egger G
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
A booster dose enhances immunogenicity of the COVID-19 vaccine candidate ChAdOx1 nCoV-19 in aged mice.
Med (New York, N.Y.)
Silva-Cayetano A,Foster WS,Innocentin S,Belij-Rammerstorfer S,Spencer AJ,Burton OT,Fra-Bidó S,Le Lee J,Thakur N,Conceicao C,Wright D,Barrett J,Evans-Bailey N,Noble C,Bailey D,Liston A,Gilbert SC,Lambe T,Linterman MA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Mar 12 00:00:00 UTC 2021
Age-related neurodegeneration and cognitive impairments of NRMT1 knockout mice are preceded by misregulation of RB and abnormal neural stem cell development.
Cell death & disease
Catlin JP,Marziali LN,Rein B,Yan Z,Feltri ML,Schaner Tooley CE
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Thu Oct 28 00:00:00 UTC 2021
Ten-eleven translocation protein 1 modulates medulloblastoma progression.
Genome biology
Kim H,Kang Y,Li Y,Chen L,Lin L,Johnson ND,Zhu D,Robinson MH,McSwain L,Barwick BG,Yuan X,Liao X,Zhao J,Zhang Z,Shu Q,Chen J,Allen EG,Kenney AM,Castellino RC,Van Meir EG,Conneely KN,Vertino PM,Jin P,Li J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Apr 29 00:00:00 UTC 2021
An mTORC1-dependent switch orchestrates the transition between mouse spermatogonial stem cells and clones of progenitor spermatogonia.
Cell reports
Suzuki S,McCarrey JR,Hermann BP
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Feb 16 00:00:00 UTC 2021
An mTORC1-dependent switch orchestrates the transition between mouse spermatogonial stem cells and clones of progenitor spermatogonia.
Cell reports
Suzuki S,McCarrey JR,Hermann BP
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Feb 16 00:00:00 UTC 2021
IL-1beta promotes the age-associated decline of beta cell function.
iScience
Böni-Schnetzler M,Méreau H,Rachid L,Wiedemann SJ,Schulze F,Trimigliozzi K,Meier DT,Donath MY
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Nov 19 00:00:00 UTC 2021
CDCP1 promotes compensatory renal growth by integrating Src and Met signaling.
Life science alliance
Kajiwara K,Yamano S,Aoki K,Okuzaki D,Matsumoto K,Okada M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Apr 01 00:00:00 UTC 2021
CXCR4 Regulates Temporal Differentiation via PRC1 Complex in Organogenesis of Epithelial Glands.
International journal of molecular sciences
Kim J,Lee SW,Park K
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Sun Jan 10 00:00:00 UTC 2021
Delta-like 4 is required for pulmonary vascular arborization and alveolarization in the developing lung.
JCI insight
Xia S,Menden HL,Townley N,Mabry SM,Johnston J,Nyp MF,Heruth DP,Korfhagen T,Sampath V
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Apr 08 00:00:00 UTC 2021
Adult neural stem cells have latent inflammatory potential that is kept suppressed by Tcf4 to facilitate adult neurogenesis.
Science advances
Shariq M,Sahasrabuddhe V,Krishna S,Radha S,Bellampalli R,Dwivedi A,Cheramangalam R,Reizis B,Hébert J,Ghosh HS
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Sat May 01 00:00:00 UTC 2021
Notch Signaling between Cerebellar Granule Cell Progenitors.
eNeuro
Adachi T,Miyashita S,Yamashita M,Shimoda M,Okonechnikov K,Chavez L,Kool M,Pfister SM,Inoue T,Kawauchi D,Hoshino M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Jul 02 00:00:00 UTC 2021
Developmental Role of Adenosine Kinase in the Cerebellum.
eNeuro
Gebril H,Wahba A,Zhou X,Lai T,Alharfoush E,DiCicco-Bloom E,Boison D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Jul 02 00:00:00 UTC 2021
Field size effects on DNA damage and proliferation in normal human cell populations irradiated with X-ray microbeams.
Scientific reports
Ojima M,Ito A,Usami N,Ohara M,Suzuki K,Kai M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri Mar 26 00:00:00 UTC 2021
Dissecting phenotypic transitions in metastatic disease via photoconversion-based isolation.
eLife
Sela Y,Li J,Kuri P,Merrell AJ,Li N,Lengner C,Rompolas P,Stanger BZ
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Feb 23 00:00:00 UTC 2021
NEIL3-deficiency increases gut permeability and contributes to a pro-atherogenic metabolic phenotype.
Scientific reports
Karlsen TR,Kong XY,Holm S,Quiles-Jiménez A,Dahl TB,Yang K,Sagen EL,Skarpengland T,S Øgaard JD,Holm K,Vestad B,Olsen MB,Aukrust P,Bjørås M,Hov JR,Halvorsen B,Gregersen I
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Tue Oct 05 00:00:00 UTC 2021
GTSE1 is possibly involved in the DNA damage repair and cisplatin resistance in osteosarcoma.
Journal of orthopaedic surgery and research
Xie C,Xiang W,Shen H,Shen J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Dec 07 00:00:00 UTC 2021
TWIK-1 BAC-GFP Transgenic Mice, an Animal Model for TWIK-1 Expression.
Cells
Kwon O,Yang H,Kim SC,Kim J,Sim J,Lee J,Hwang EM,Shim S,Park JY
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Oct 14 00:00:00 UTC 2021
Targeting Feedforward Loops Formed by Nuclear Receptor RORγ and Kinase PBK in mCRPC with Hyperactive AR Signaling.
Cancers
Zhang X,Huang Z,Wang J,Ma Z,Yang J,Corey E,Evans CP,Yu AM,Chen HW
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Apr 01 00:00:00 UTC 2021
Gene Signatures Detect Damaged Liver Sinusoidal Endothelial Cells in Chronic Liver Diseases.
Frontiers in medicine
Verhulst S,van Os EA,De Smet V,Eysackers N,Mannaerts I,van Grunsven LA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Wed Nov 10 00:00:00 UTC 2021
Imbalanced Activation of Wnt-/β-Catenin-Signaling in Liver Endothelium Alters Normal Sinusoidal Differentiation.
Frontiers in physiology
Koch PS,Sandorski K,Heil J,Schmid CD,Kürschner SW,Hoffmann J,Winkler M,Staniczek T,de la Torre C,Sticht C,Schledzewski K,Taketo MM,Trogisch FA,Heineke J,Géraud C,Goerdt S,Olsavszky V
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Oct 22 00:00:00 UTC 2021
An in vivo model allowing continuous observation of human vascular formation in the same animal over time.
Scientific reports
Tsukada Y,Muramatsu F,Hayashi Y,Inagaki C,Su H,Iba T,Kidoya H,Takakura N
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Jan 12 00:00:00 UTC 2021
Mutant ASXL1 induces age-related expansion of phenotypic hematopoietic stem cells through activation of Akt/mTOR pathway.
Nature communications
Fujino T,Goyama S,Sugiura Y,Inoue D,Asada S,Yamasaki S,Matsumoto A,Yamaguchi K,Isobe Y,Tsuchiya A,Shikata S,Sato N,Morinaga H,Fukuyama T,Tanaka Y,Fukushima T,Takeda R,Yamamoto K,Honda H,Nishimura EK,Furukawa Y,Shibata T,Abdel-Wahab O,Suematsu M,Kitamura T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Mar 23 00:00:00 UTC 2021
Heterologous arenavirus vector prime-boost overrules self-tolerance for efficient tumor-specific CD8 T cell attack.
Cell reports. Medicine
Bonilla WV,Kirchhammer N,Marx AF,Kallert SM,Krzyzaniak MA,Lu M,Darbre S,Schmidt S,Raguz J,Berka U,Vincenti I,Pauzuolis M,Kerber R,Hoepner S,Günther S,Magnus C,Merkler D,Orlinger KK,Zippelius A,Pinschewer DD
56-5698-82 was used in Flow Cytometry to establish an arenavirus-based immunotherapy regimen that allows reshuffling of immunodominance hierarchies and breaking self-directed tolerance for efficient tumor control.
Tue Mar 16 00:00:00 UTC 2021
Targeting Phosphatidylserine Enhances the Anti-tumor Response to Tumor-Directed Radiation Therapy in a Preclinical Model of Melanoma.
Cell reports
Budhu S,Giese R,Gupta A,Fitzgerald K,Zappasodi R,Schad S,Hirschhorn D,Campesato LF,De Henau O,Gigoux M,Liu C,Mazo G,Deng L,Barker CA,Wolchok JD,Merghoub T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
Shikonin Derivatives from Onsoma visianii Decrease Expression of Phosphorylated STAT3 in Leukemia Cells and Exert Antitumor Activity.
Nutrients
Todorovic Z,Milovanovic J,Arsenijevic D,Vukovic N,Vukic M,Arsenijevic A,Djurdjevic P,Milovanovic M,Arsenijevic N
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Mar 31 00:00:00 UTC 2021
Cross-Reactivity and Functionality of Approved Human Immune Checkpoint Blockers in Dogs.
Cancers
Pantelyushin S,Ranninger E,Guerrera D,Hutter G,Maake C,Markkanen E,Bettschart-Wolfensberger R,Rohrer Bley C,Läubli H,Vom Berg J
56-5698 was used in Flow cytometry/Cell sorting to assess suitability of seven FDA-approved human ICIs to target CTLA-4 or PD-1/PD-L1 in dogs.
Sat Feb 13 00:00:00 UTC 2021
Human Lung Stem Cell-Based Alveolospheres Provide Insights into SARS-CoV-2-Mediated Interferon Responses and Pneumocyte Dysfunction.
Cell stem cell
Katsura H,Sontake V,Tata A,Kobayashi Y,Edwards CE,Heaton BE,Konkimalla A,Asakura T,Mikami Y,Fritch EJ,Lee PJ,Heaton NS,Boucher RC,Randell SH,Baric RS,Tata PR
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Dec 03 00:00:00 UTC 2020
Human Lung Stem Cell-Based Alveolospheres Provide Insights into SARS-CoV-2-Mediated Interferon Responses and Pneumocyte Dysfunction.
Cell stem cell
Katsura H,Sontake V,Tata A,Kobayashi Y,Edwards CE,Heaton BE,Konkimalla A,Asakura T,Mikami Y,Fritch EJ,Lee PJ,Heaton NS,Boucher RC,Randell SH,Baric RS,Tata PR
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Dec 03 00:00:00 UTC 2020
Egr2-guided histone H2B monoubiquitination is required for peripheral nervous system myelination.
Nucleic acids research
Wüst HM,Wegener A,Fröb F,Hartwig AC,Wegwitz F,Kari V,Schimmel M,Tamm ER,Johnsen SA,Wegner M,Sock E
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri Sep 18 00:00:00 UTC 2020
Genetic manipulation of LKB1 elicits lethal metastatic prostate cancer.
The Journal of experimental medicine
Hermanova I,Zúñiga-García P,Caro-Maldonado A,Fernandez-Ruiz S,Salvador F,Martín-Martín N,Zabala-Letona A,Nuñez-Olle M,Torrano V,Camacho L,Lizcano JM,Talamillo A,Carreira S,Gurel B,Cortazar AR,Guiu M,López JI,Martinez-Romero A,Astobiza I,Valcarcel-Jimenez L,Lorente M,Arruabarrena-Aristorena A,Velasco G,Gomez-Muñoz A,Suárez-Cabrera C,Lodewijk I,Flores JM,Sutherland JD,Barrio R,de Bono JS,Paramio JM,Trka J,Graupera M,Gomis RR,Carracedo A
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Jun 01 00:00:00 UTC 2020
Epithelial Vegfa Specifies a Distinct Endothelial Population in the Mouse Lung.
Developmental cell
Vila Ellis L,Cain MP,Hutchison V,Flodby P,Crandall ED,Borok Z,Zhou B,Ostrin EJ,Wythe JD,Chen J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Mon Mar 09 00:00:00 UTC 2020
Administration of all-trans retinoic acid after experimental traumatic brain injury is brain protective.
British journal of pharmacology
Hummel R,Ulbrich S,Appel D,Li S,Hirnet T,Zander S,Bobkiewicz W,Gölz C,Schäfer MKE
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Sun Nov 01 00:00:00 UTC 2020
Multimodal single-cell analysis reveals distinct radioresistant stem-like and progenitor cell populations in murine glioma.
Glia
Alexander J,LaPlant QC,Pattwell SS,Szulzewsky F,Cimino PJ,Caruso FP,Pugliese P,Chen Z,Chardon F,Hill AJ,Spurrell C,Ahrendsen D,Pietras A,Starita LM,Hambardzumyan D,Iavarone A,Shendure J,Holland EC
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Dec 01 00:00:00 UTC 2020
FGF2 modulates simultaneously the mode, the rate of division and the growth fraction in cultures of radial glia.
Development (Cambridge, England)
Ledesma-Terrón M,Peralta-Cañadas N,Míguez DG
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri Jul 24 00:00:00 UTC 2020
NPC1 deficiency impairs cerebellar postnatal development of microglia and climbing fiber refinement in a mouse model of Niemann-Pick disease type C.
Development (Cambridge, England)
Boyle BR,Melli SE,Altreche RS,Padron ZM,Yousufzai FAK,Kim S,Vasquez MD,Carone DM,Carone BR,Soto I
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry (Frozen)
Mon Aug 03 00:00:00 UTC 2020
Novel enhanced GFP-positive congenic inbred strain establishment and application of tumor-bearing nude mouse model.
Cancer science
Lan Q,Chen Y,Dai C,Li S,Fei X,Dong J,Shen Y,Dai X,Lu Z,Liu B,Wang Q,Wang H,Zhou Z,Ji X,Wang Z,Huang Q
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Thu Oct 01 00:00:00 UTC 2020
Mycobacterium tuberculosis-infected alveolar epithelial cells modulate dendritic cell function through the HIF-1α-NOS2 axis.
Journal of leukocyte biology
Rodrigues TS,Alvarez ARP,Gembre AF,Forni MFPAD,de Melo BMS,Alves Filho JCF,Câmara NOS,Bonato VLD
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Radiation Induced Metabolic Alterations Associate With Tumor Aggressiveness and Poor Outcome in Glioblastoma.
Frontiers in oncology
Gupta K,Vuckovic I,Zhang S,Xiong Y,Carlson BL,Jacobs J,Olson I,Petterson XM,Macura SI,Sarkaria J,Burns TC
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Mon Sep 28 00:00:00 UTC 2020
Radiation Induced Metabolic Alterations Associate With Tumor Aggressiveness and Poor Outcome in Glioblastoma.
Frontiers in oncology
Gupta K,Vuckovic I,Zhang S,Xiong Y,Carlson BL,Jacobs J,Olson I,Petterson XM,Macura SI,Sarkaria J,Burns TC
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Mon Sep 28 00:00:00 UTC 2020
TNFSF15 facilitates human umbilical cord blood haematopoietic stem cell expansion by activating Notch signal pathway.
Journal of cellular and molecular medicine
Ding Y,Gao S,Shen J,Bai T,Yang M,Xu S,Gao Y,Zhang Z,Li L
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Agrin Promotes Limbal Stem Cell Proliferation and Corneal Wound Healing Through Hippo-Yap Signaling Pathway.
Investigative ophthalmology & visual science
Hou L,Fu W,Liu Y,Wang Q,Wang L,Huang Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Mon May 11 00:00:00 UTC 2020
Quinazolinone derivative BNUA-3 ameliorated [NDEA+2-AAF]-induced liver carcinogenesis in SD rats by modulating AhR-CYP1B1-Nrf2-Keap1 pathway.
Clinical and experimental pharmacology & physiology
Bose P,Siddique MUM,Acharya R,Jayaprakash V,Sinha BN,Lapenna A,Pattanayak SP
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Wed Jan 01 00:00:00 UTC 2020
New Insights into the Pathogenesis of Diabetic Nephropathy: Proximal Renal Tubules Are Primary Target of Oxidative Stress in Diabetic Kidney.
Acta histochemica et cytochemica
Haraguchi R,Kohara Y,Matsubayashi K,Kitazawa R,Kitazawa S
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Apr 28 00:00:00 UTC 2020
Elevated Serum Amino Acids Induce a Subpopulation of Alpha Cells to Initiate Pancreatic Neuroendocrine Tumor Formation.
Cell reports. Medicine
Smith DK,Kates L,Durinck S,Patel N,Stawiski EW,Kljavin N,Foreman O,Sipos B,Solloway MJ,Allan BB,Peterson AS
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Aug 25 00:00:00 UTC 2020
Aripiprazole reduces liver cell division.
PloS one
Pirc Marolt T,Kramar B,Bulc Rozman K,Šuput D,Milisav I
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Tue Dec 08 00:00:00 UTC 2020
Regeneration of the pulmonary vascular endothelium after viral pneumonia requires COUP-TF2.
Science advances
Zhao G,Weiner AI,Neupauer KM,de Mello Costa MF,Palashikar G,Adams-Tzivelekidis S,Mangalmurti NS,Vaughan AE
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Sun Nov 01 00:00:00 UTC 2020
Transcriptome analysis of basic fibroblast growth factor treated stem cells isolated from human exfoliated deciduous teeth.
Heliyon
Nowwarote N,Manokawinchoke J,Kanjana K,Fournier BPJ,Sukarawan W,Osathanon T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Mon Jun 01 00:00:00 UTC 2020
The effects of hedgehog ligand neutralising antibody 5E1 in a mouse model of endometriosis.
BMC research notes
Cousins FL,Farley JK,Kerrigan R,Mukherjee S,Darzi S,Gargett CE,Deane JA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri Sep 25 00:00:00 UTC 2020
B Cell Activating Factor (BAFF) Is Required for the Development of Intra-Renal Tertiary Lymphoid Organs in Experimental Kidney Transplantation in Rats.
International journal of molecular sciences
Steines L,Poth H,Herrmann M,Schuster A,Banas B,Bergler T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Wed Oct 28 00:00:00 UTC 2020
A discrete subtype of neural progenitor crucial for cortical folding in the gyrencephalic mammalian brain.
eLife
Matsumoto N,Tanaka S,Horiike T,Shinmyo Y,Kawasaki H
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Tue Apr 21 00:00:00 UTC 2020
A discrete subtype of neural progenitor crucial for cortical folding in the gyrencephalic mammalian brain.
eLife
Matsumoto N,Tanaka S,Horiike T,Shinmyo Y,Kawasaki H
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Tue Apr 21 00:00:00 UTC 2020
Changes in Liver Mechanical Properties and Water Diffusivity During Normal Pregnancy Are Driven by Cellular Hypertrophy.
Frontiers in physiology
Garczyńska K,Tzschätzsch H,Kühl AA,Morr AS,Lilaj L,Häckel A,Schellenberger E,Berndt N,Holzhütter HG,Braun J,Sack I,Guo J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Dec 18 00:00:00 UTC 2020
Interleukin-13 disrupts type 2 pneumocyte stem cell activity.
JCI insight
Glisinski KM,Schlobohm AJ,Paramore SV,Birukova A,Moseley MA,Foster MW,Barkauskas CE
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Jan 16 00:00:00 UTC 2020
Interleukin-13 disrupts type 2 pneumocyte stem cell activity.
JCI insight
Glisinski KM,Schlobohm AJ,Paramore SV,Birukova A,Moseley MA,Foster MW,Barkauskas CE
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Jan 16 00:00:00 UTC 2020
Chemosensitization of prostate cancer stem cells in mice by angiogenin and plexin-B2 inhibitors.
Communications biology
Li S,Goncalves KA,Lyu B,Yuan L,Hu GF
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Jan 15 00:00:00 UTC 2020
A single-cell transcriptomic and anatomic atlas of mouse dorsal raphe Pet1 neurons.
eLife
Okaty BW,Sturrock N,Escobedo Lozoya Y,Chang Y,Senft RA,Lyon KA,Alekseyenko OV,Dymecki SM
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Jun 22 00:00:00 UTC 2020
Follicular Regulatory T Cells Can Access the Germinal Center Independently of CXCR5.
Cell reports
Vanderleyden I,Fra-Bido SC,Innocentin S,Stebegg M,Okkenhaug H,Evans-Bailey N,Pierson W,Denton AE,Linterman MA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Jan 21 00:00:00 UTC 2020
Follicular Regulatory T Cells Can Access the Germinal Center Independently of CXCR5.
Cell reports
Vanderleyden I,Fra-Bido SC,Innocentin S,Stebegg M,Okkenhaug H,Evans-Bailey N,Pierson W,Denton AE,Linterman MA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Jan 21 00:00:00 UTC 2020
Metastasis-initiating cells induce and exploit a fibroblast niche to fuel malignant colonization of the lungs.
Nature communications
Pein M,Insua-Rodríguez J,Hongu T,Riedel A,Meier J,Wiedmann L,Decker K,Essers MAG,Sinn HP,Spaich S,Sütterlin M,Schneeweiss A,Trumpp A,Oskarsson T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Mar 20 00:00:00 UTC 2020
Heart-derived fibroblasts express LYPD-1 and negatively regulate angiogenesis in rat.
Regenerative therapy
Sakamoto S,Matsuura K,Masuda S,Hagiwara N,Shimizu T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Dec 01 00:00:00 UTC 2020
CCL2 and CXCL12 Derived from Mesenchymal Stromal Cells Cooperatively Polarize IL-10+ Tissue Macrophages to Mitigate Gut Injury.
Cell reports
Giri J,Das R,Nylen E,Chinnadurai R,Galipeau J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Feb 11 00:00:00 UTC 2020
Adenosine kinase inhibition promotes proliferation of neural stem cells after traumatic brain injury.
Brain communications
Gebril HM,Rose RM,Gesese R,Emond MP,Huo Y,Aronica E,Boison D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Apr 02 00:00:00 UTC 2021
Comparative Analysis of Age-Related Changes in Lacrimal Glands and Meibomian Glands of a C57BL/6 Male Mouse Model.
International journal of molecular sciences
Yoon CH,Ryu JS,Hwang HS,Kim MK
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Jun 11 00:00:00 UTC 2020
Microbiota-Sourced Purines Support Wound Healing and Mucous Barrier Function.
iScience
Lee JS,Wang RX,Goldberg MS,Clifford GP,Kao DJ,Colgan SP
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry (Paraffin)
Fri Jun 26 00:00:00 UTC 2020
CD103+ cDC1 and endogenous CD8+ T cells are necessary for improved CD40L-overexpressing CAR T cell antitumor function.
Nature communications
Kuhn NF,Lopez AV,Li X,Cai W,Daniyan AF,Brentjens RJ
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Dec 02 00:00:00 UTC 2020
Eradication of tumor growth by delivering novel photothermal selenium-coated tellurium nanoheterojunctions.
Science advances
Chen S,Xing C,Huang D,Zhou C,Ding B,Guo Z,Peng Z,Wang D,Zhu X,Liu S,Cai Z,Wu J,Zhao J,Wu Z,Zhang Y,Wei C,Yan Q,Wang H,Fan D,Liu L,Zhang H,Cao Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Wed Apr 01 00:00:00 UTC 2020
Treatment of mice with a ligand binding blocking anti-CD28 monoclonal antibody improves healing after myocardial infarction.
PloS one
Gladow N,Hollmann C,Ramos G,Frantz S,Kerkau T,Beyersdorf N,Hofmann U
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Jul 06 00:00:00 UTC 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Dec 17 00:00:00 UTC 2020
Sphingosine 1-phosphate-regulated transcriptomes in heterogenous arterial and lymphatic endothelium of the aorta.
eLife
Engelbrecht E,Levesque MV,He L,Vanlandewijck M,Nitzsche A,Niazi H,Kuo A,Singh SA,Aikawa M,Holton K,Proia RL,Kono M,Pu WT,Camerer E,Betsholtz C,Hla T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Feb 24 00:00:00 UTC 2020
Poor Concordance of Floxed Sequence Recombination in Single Neural Stem Cells: Implications for Cell Autonomous Studies.
eNeuro
Dause TJ,Kirby ED
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Jun 18 00:00:00 UTC 2021
Discrete populations of isotype-switched memory B lymphocytes are maintained in murine spleen and bone marrow.
Nature communications
Riedel R,Addo R,Ferreira-Gomes M,Heinz GA,Heinrich F,Kummer J,Greiff V,Schulz D,Klaeden C,Cornelis R,Menzel U,Kröger S,Stervbo U,Köhler R,Haftmann C,Kühnel S,Lehmann K,Maschmeyer P,McGrath M,Naundorf S,Hahne S,Sercan-Alp Ö,Siracusa F,Stefanowski J,Weber M,Westendorf K,Zimmermann J,Hauser AE,Reddy ST,Durek P,Chang HD,Mashreghi MF,Radbruch A
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Fri May 22 00:00:00 UTC 2020
Anti-commensal IgG Drives Intestinal Inflammation and Type 17 Immunity in Ulcerative Colitis.
Immunity
Castro-Dopico T,Dennison TW,Ferdinand JR,Mathews RJ,Fleming A,Clift D,Stewart BJ,Jing C,Strongili K,Labzin LI,Monk EJM,Saeb-Parsy K,Bryant CE,Clare S,Parkes M,Clatworthy MR
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Apr 16 00:00:00 UTC 2019
Anti-commensal IgG Drives Intestinal Inflammation and Type 17 Immunity in Ulcerative Colitis.
Immunity
Castro-Dopico T,Dennison TW,Ferdinand JR,Mathews RJ,Fleming A,Clift D,Stewart BJ,Jing C,Strongili K,Labzin LI,Monk EJM,Saeb-Parsy K,Bryant CE,Clare S,Parkes M,Clatworthy MR
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Apr 16 00:00:00 UTC 2019
Sensing of apoptotic cells through Axl causes lung basal cell proliferation in inflammatory diseases.
The Journal of experimental medicine
Fujino N,Brand OJ,Morgan DJ,Fujimori T,Grabiec AM,Jagger CP,Maciewicz RA,Yamada M,Itakura K,Sugiura H,Ichinose M,Hussell T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Sep 02 00:00:00 UTC 2019
Sensing of apoptotic cells through Axl causes lung basal cell proliferation in inflammatory diseases.
The Journal of experimental medicine
Fujino N,Brand OJ,Morgan DJ,Fujimori T,Grabiec AM,Jagger CP,Maciewicz RA,Yamada M,Itakura K,Sugiura H,Ichinose M,Hussell T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Sep 02 00:00:00 UTC 2019
JAK-STAT inhibition impairs K-RAS-driven lung adenocarcinoma progression.
International journal of cancer
Mohrherr J,Haber M,Breitenecker K,Aigner P,Moritsch S,Voronin V,Eferl R,Moriggl R,Stoiber D,Győrffy B,Brcic L,László V,Döme B,Moldvay J,Dezső K,Bilban M,Popper H,Moll HP,Casanova E
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Sun Dec 15 00:00:00 UTC 2019
Comprehensive and cell-type-based characterization of the dorsal midbrain during development.
Genes to cells : devoted to molecular & cellular mechanisms
Arimura N,Dewa KI,Okada M,Yanagawa Y,Taya SI,Hoshino M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Jan 01 00:00:00 UTC 2019
Absence of TGFβ signaling in retinal microglia induces retinal degeneration and exacerbates choroidal neovascularization.
eLife
Ma W,Silverman SM,Zhao L,Villasmil R,Campos MM,Amaral J,Wong WT
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Tue Jan 22 00:00:00 UTC 2019
Sustained expression of HeyL is critical for the proliferation of muscle stem cells in overloaded muscle.
eLife
Fukuda S,Kaneshige A,Kaji T,Noguchi YT,Takemoto Y,Zhang L,Tsujikawa K,Kokubo H,Uezumi A,Maehara K,Harada A,Ohkawa Y,Fukada SI
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Mon Sep 23 00:00:00 UTC 2019
Regulatory T-cells regulate neonatal heart regeneration by potentiating cardiomyocyte proliferation in a paracrine manner.
Theranostics
Li J,Yang KY,Tam RCY,Chan VW,Lan HY,Hori S,Zhou B,Lui KO
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri Aug 14 00:00:00 UTC 2020
Deficiency in the secreted protein Semaphorin3d causes abnormal parathyroid development in mice.
The Journal of biological chemistry
Singh A,Mia MM,Cibi DM,Arya AK,Bhadada SK,Singh MK
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri May 24 00:00:00 UTC 2019
Live imaging of alveologenesis in precision-cut lung slices reveals dynamic epithelial cell behaviour.
Nature communications
Akram KM,Yates LL,Mongey R,Rothery S,Gaboriau DCA,Sanderson J,Hind M,Griffiths M,Dean CH
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Mar 12 00:00:00 UTC 2019
Live imaging of alveologenesis in precision-cut lung slices reveals dynamic epithelial cell behaviour.
Nature communications
Akram KM,Yates LL,Mongey R,Rothery S,Gaboriau DCA,Sanderson J,Hind M,Griffiths M,Dean CH
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Mar 12 00:00:00 UTC 2019
A solid-phase transfection platform for arrayed CRISPR screens.
Molecular systems biology
Serçin Ö,Reither S,Roidos P,Ballin N,Palikyras S,Baginska A,Rein K,Llamazares M,Halavatyi A,Winter H,Muley T,Jurkowska RZ,Abdollahi A,Zenke FT,Neumann B,Mardin BR
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Sun Dec 01 00:00:00 UTC 2019
SORLA regulates endosomal trafficking and oncogenic fitness of HER2.
Nature communications
Pietilä M,Sahgal P,Peuhu E,Jäntti NZ,Paatero I,Närvä E,Al-Akhrass H,Lilja J,Georgiadou M,Andersen OM,Padzik A,Sihto H,Joensuu H,Blomqvist M,Saarinen I,Boström PJ,Taimen P,Ivaska J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue May 28 00:00:00 UTC 2019
Soluble TREM2 ameliorates pathological phenotypes by modulating microglial functions in an Alzheimer's disease model.
Nature communications
Zhong L,Xu Y,Zhuo R,Wang T,Wang K,Huang R,Wang D,Gao Y,Zhu Y,Sheng X,Chen K,Wang N,Zhu L,Can D,Marten Y,Shinohara M,Liu CC,Du D,Sun H,Wen L,Xu H,Bu G,Chen XF
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Mon Mar 25 00:00:00 UTC 2019
FCRL5+ Memory B Cells Exhibit Robust Recall Responses.
Cell reports
Kim CC,Baccarella AM,Bayat A,Pepper M,Fontana MF
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Apr 30 00:00:00 UTC 2019
Orphan Nuclear Receptor NR2F6 Suppresses T Follicular Helper Cell Accumulation through Regulation of IL-21.
Cell reports
Olson WJ,Jakic B,Labi V,Schoeler K,Kind M,Klepsch V,Baier G,Hermann-Kleiter N
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Sep 10 00:00:00 UTC 2019
Targeted Delivery of Stk25 Antisense Oligonucleotides to Hepatocytes Protects Mice Against Nonalcoholic Fatty Liver Disease.
Cellular and molecular gastroenterology and hepatology
Cansby E,Nuñez-Durán E,Magnusson E,Amrutkar M,Booten SL,Kulkarni NM,Svensson LT,Borén J,Marschall HU,Aghajan M,Mahlapuu M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Tue Jun 04 00:00:00 UTC 2019
Spatial Genome Re-organization between Fetal and Adult Hematopoietic Stem Cells.
Cell reports
Chen C,Yu W,Tober J,Gao P,He B,Lee K,Trieu T,Blobel GA,Speck NA,Tan K
56-5698 was used in Flow cytometry/Cell sorting to find that chromosomal compartments and topologically associating domains (TADs) are largely conserved between fetal and adult HSCs.
Tue Dec 17 00:00:00 UTC 2019
Pulsatile MEK Inhibition Improves Anti-tumor Immunity and T Cell Function in Murine Kras Mutant Lung Cancer.
Cell reports
Choi H,Deng J,Li S,Silk T,Dong L,Brea EJ,Houghton S,Redmond D,Zhong H,Boiarsky J,Akbay EA,Smith PD,Merghoub T,Wong KK,Wolchok JD
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Apr 16 00:00:00 UTC 2019
IL-17 inhibits CXCL9/10-mediated recruitment of CD8+ cytotoxic T cells and regulatory T cells to colorectal tumors.
Journal for immunotherapy of cancer
Chen J,Ye X,Pitmon E,Lu M,Wan J,Jellison ER,Adler AJ,Vella AT,Wang K
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Nov 27 00:00:00 UTC 2019
Keap1-Nrf2 System Plays an Important Role in Invariant Natural Killer T Cell Development and Homeostasis.
Cell reports
Pyaram K,Kumar A,Kim YH,Noel S,Reddy SP,Rabb H,Chang CH
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Apr 16 00:00:00 UTC 2019
Lymphoid Aggregates in the CNS of Progressive Multiple Sclerosis Patients Lack Regulatory T Cells.
Frontiers in immunology
Bell L,Lenhart A,Rosenwald A,Monoranu CM,Berberich-Siebelt F
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Oct 22 00:00:00 UTC 2020
Adrenergic nerve degeneration in bone marrow drives aging of the hematopoietic stem cell niche.
Nature medicine
Maryanovich M,Zahalka AH,Pierce H,Pinho S,Nakahara F,Asada N,Wei Q,Wang X,Ciero P,Xu J,Leftin A,Frenette PS
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Fri Jun 01 00:00:00 UTC 2018
Tumor Evolution and Drug Response in Patient-Derived Organoid Models of Bladder Cancer.
Cell
Lee SH,Hu W,Matulay JT,Silva MV,Owczarek TB,Kim K,Chua CW,Barlow LJ,Kandoth C,Williams AB,Bergren SK,Pietzak EJ,Anderson CB,Benson MC,Coleman JA,Taylor BS,Abate-Shen C,McKiernan JM,Al-Ahmadie H,Solit DB,Shen MM
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Thu Apr 05 00:00:00 UTC 2018
Modulation of Myelopoiesis Progenitors Is an Integral Component of Trained Immunity.
Cell
Mitroulis I,Ruppova K,Wang B,Chen LS,Grzybek M,Grinenko T,Eugster A,Troullinaki M,Palladini A,Kourtzelis I,Chatzigeorgiou A,Schlitzer A,Beyer M,Joosten LAB,Isermann B,Lesche M,Petzold A,Simons K,Henry I,Dahl A,Schultze JL,Wielockx B,Zamboni N,Mirtschink P,Coskun Ü,Hajishengallis G,Netea MG,Chavakis T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Jan 11 00:00:00 UTC 2018
A Metabolite-Triggered Tuft Cell-ILC2 Circuit Drives Small Intestinal Remodeling.
Cell
Schneider C,O'Leary CE,von Moltke J,Liang HE,Ang QY,Turnbaugh PJ,Radhakrishnan S,Pellizzon M,Ma A,Locksley RM
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Jul 12 00:00:00 UTC 2018
Fc Effector Function Contributes to the Activity of Human Anti-CTLA-4 Antibodies.
Cancer cell
Arce Vargas F,Furness AJS,Litchfield K,Joshi K,Rosenthal R,Ghorani E,Solomon I,Lesko MH,Ruef N,Roddie C,Henry JY,Spain L,Ben Aissa A,Georgiou A,Wong YNS,Smith M,Strauss D,Hayes A,Nicol D,O'Brien T,Mårtensson L,Ljungars A,Teige I,Frendéus B,Pule M,Marafioti T,Gore M,Larkin J,Turajlic S,Swanton C,Peggs KS,Quezada SA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Apr 09 00:00:00 UTC 2018
The Epigenetic State of PRDM16-Regulated Enhancers in Radial Glia Controls Cortical Neuron Position.
Neuron
Baizabal JM,Mistry M,García MT,Gómez N,Olukoya O,Tran D,Johnson MB,Walsh CA,Harwell CC
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Wed Jun 06 00:00:00 UTC 2018
Dual-targeting Wnt and uPA receptors using peptide conjugated ultra-small nanoparticle drug carriers inhibited cancer stem-cell phenotype in chemo-resistant breast cancer.
Biomaterials
Miller-Kleinhenz J,Guo X,Qian W,Zhou H,Bozeman EN,Zhu L,Ji X,Wang YA,Styblo T,O'Regan R,Mao H,Yang L
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Jan 01 00:00:00 UTC 2018
Dual-targeting Wnt and uPA receptors using peptide conjugated ultra-small nanoparticle drug carriers inhibited cancer stem-cell phenotype in chemo-resistant breast cancer.
Biomaterials
Miller-Kleinhenz J,Guo X,Qian W,Zhou H,Bozeman EN,Zhu L,Ji X,Wang YA,Styblo T,O'Regan R,Mao H,Yang L
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Jan 01 00:00:00 UTC 2018
Defining Lineage Potential and Fate Behavior of Precursors during Pancreas Development.
Developmental cell
Sznurkowska MK,Hannezo E,Azzarelli R,Rulands S,Nestorowa S,Hindley CJ,Nichols J,Göttgens B,Huch M,Philpott A,Simons BD
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Aug 06 00:00:00 UTC 2018
Defining Lineage Potential and Fate Behavior of Precursors during Pancreas Development.
Developmental cell
Sznurkowska MK,Hannezo E,Azzarelli R,Rulands S,Nestorowa S,Hindley CJ,Nichols J,Göttgens B,Huch M,Philpott A,Simons BD
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Aug 06 00:00:00 UTC 2018
MicroRNA-126 deficiency enhanced the activation and function of CD4+ T cells by elevating IRS-1 pathway.
Clinical and experimental immunology
Chu F,Hu Y,Zhou Y,Guo M,Lu J,Zheng W,Xu H,Zhao J,Xu L
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2018
Inhibition of histone deacetylase 1 ameliorates renal tubulointerstitial fibrosis via modulation of inflammation and extracellular matrix gene transcription in mice.
International journal of molecular medicine
Nguyễn-Thanh T,Kim D,Lee S,Kim W,Park SK,Kang KP
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Jan 01 00:00:00 UTC 2018
Vitreous Cytokine Expression and a Murine Model Suggest a Key Role of Microglia in the Inflammatory Response to Retinal Detachment.
Investigative ophthalmology & visual science
Kiang L,Ross BX,Yao J,Shanmugam S,Andrews CA,Hansen S,Besirli CG,Zacks DN,Abcouwer SF
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Jul 02 00:00:00 UTC 2018
C-Kit Cardiac Progenitor Cell Based Cell Sheet Improves Vascularization and Attenuates Cardiac Remodeling following Myocardial Infarction in Rats.
BioMed research international
Dergilev K,Tsokolaeva Z,Makarevich P,Beloglazova I,Zubkova E,Boldyreva M,Ratner E,Dyikanov D,Menshikov M,Ovchinnikov A,Ageev F,Parfyonova Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Jan 14 00:00:00 UTC 2019
Selective pharmacological inhibition of DDR1 prevents experimentally-induced glomerulonephritis in prevention and therapeutic regime.
Journal of translational medicine
Moll S,Yasui Y,Abed A,Murata T,Shimada H,Maeda A,Fukushima N,Kanamori M,Uhles S,Badi L,Cagarelli T,Formentini I,Drawnel F,Georges G,Bergauer T,Gasser R,Bonfil RD,Fridman R,Richter H,Funk J,Moeller MJ,Chatziantoniou C,Prunotto M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry (Paraffin)
Fri Jun 01 00:00:00 UTC 2018
Factors Within the Endoneurial Microenvironment Act to Suppress Tumorigenesis of MPNST.
Frontiers in cellular neuroscience
Stratton JA,Assinck P,Sinha S,Kumar R,Moulson A,Patrick N,Raharjo E,Chan JA,Midha R,Tetzlaff W,Biernaskie J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Thu Oct 01 00:00:00 UTC 2020
Lymphotoxin α fine-tunes T cell clonal deletion by regulating thymic entry of antigen-presenting cells.
Nature communications
Lopes N,Charaix J,Cédile O,Sergé A,Irla M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Mar 28 00:00:00 UTC 2018
ARTS mediates apoptosis and regeneration of the intestinal stem cell niche.
Nature communications
Koren E,Yosefzon Y,Ankawa R,Soteriou D,Jacob A,Nevelsky A,Ben-Yosef R,Bar-Sela G,Fuchs Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Fri Nov 02 00:00:00 UTC 2018
Pax6 Lengthens G1 Phase and Decreases Oscillating Cdk6 Levels in Murine Embryonic Cortical Progenitors.
Frontiers in cellular neuroscience
Mi D,Manuel M,Huang YT,Mason JO,Price DJ
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Mar 04 00:00:00 UTC 2021
The organic ester O,O'-diethyl-(S,S)-ethylenediamine-N,N'-di-2-(3-cyclohexyl)propanoate dihydrochloride attenuates murine breast cancer growth and metastasis.
Oncotarget
Jurisevic M,Arsenijevic A,Pantic J,Gajovic N,Milovanovic J,Milovanovic M,Poljarevic J,Sabo T,Vojvodic D,Radosavljevic GD,Arsenijevic N
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Fri Jun 15 00:00:00 UTC 2018
The transcription factor Klf5 is essential for intrahepatic biliary epithelial tissue remodeling after cholestatic liver injury.
The Journal of biological chemistry
Okada H,Yamada M,Kamimoto K,Kok CY,Kaneko K,Ema M,Miyajima A,Itoh T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Apr 27 00:00:00 UTC 2018
ATF3 Sustains IL-22-Induced STAT3 Phosphorylation to Maintain Mucosal Immunity Through Inhibiting Phosphatases.
Frontiers in immunology
Glal D,Sudhakar JN,Lu HH,Liu MC,Chiang HY,Liu YC,Cheng CF,Shui JW
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Sep 26 00:00:00 UTC 2019
Stem cell factor is selectively secreted by arterial endothelial cells in bone marrow.
Nature communications
Xu C,Gao X,Wei Q,Nakahara F,Zimmerman SE,Mar J,Frenette PS
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Fri Jun 22 00:00:00 UTC 2018
A somatic mutation in erythro-myeloid progenitors causes neurodegenerative disease.
Nature
Mass E,Jacome-Galarza CE,Blank T,Lazarov T,Durham BH,Ozkaya N,Pastore A,Schwabenland M,Chung YR,Rosenblum MK,Prinz M,Abdel-Wahab O,Geissmann F
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Sep 21 00:00:00 UTC 2017
Myeloid progenitor cluster formation drives emergency and leukaemic myelopoiesis.
Nature
Hérault A,Binnewies M,Leong S,Calero-Nieto FJ,Zhang SY,Kang YA,Wang X,Pietras EM,Chu SH,Barry-Holson K,Armstrong S,Göttgens B,Passegué E
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Apr 06 00:00:00 UTC 2017
De Novo Epigenetic Programs Inhibit PD-1 Blockade-Mediated T Cell Rejuvenation.
Cell
Ghoneim HE,Fan Y,Moustaki A,Abdelsamed HA,Dash P,Dogra P,Carter R,Awad W,Neale G,Thomas PG,Youngblood B
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Jun 29 00:00:00 UTC 2017
White Adipose Tissue Is a Reservoir for Memory T Cells and Promotes Protective Memory Responses to Infection.
Immunity
Han SJ,Glatman Zaretsky A,Andrade-Oliveira V,Collins N,Dzutsev A,Shaik J,Morais da Fonseca D,Harrison OJ,Tamoutounour S,Byrd AL,Smelkinson M,Bouladoux N,Bliska JB,Brenchley JM,Brodsky IE,Belkaid Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Dec 19 00:00:00 UTC 2017
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
Mutations in 5-methylcytosine oxidase TET2 and RhoA cooperatively disrupt T cell homeostasis.
The Journal of clinical investigation
Zang S,Li J,Yang H,Zeng H,Han W,Zhang J,Lee M,Moczygemba M,Isgandarova S,Yang Y,Zhou Y,Rao A,You MJ,Sun D,Huang Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2017
Egr2 and 3 control adaptive immune responses by temporally uncoupling expansion from T cell differentiation.
The Journal of experimental medicine
Miao T,Symonds ALJ,Singh R,Symonds JD,Ogbe A,Omodho B,Zhu B,Li S,Wang P
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Jun 05 00:00:00 UTC 2017
1810011o10 Rik Inhibits the Antitumor Effect of Intratumoral CD8+ T Cells through Suppression of Notch2 Pathway in a Murine Hepatocellular Carcinoma Model.
Frontiers in immunology
Dai K,Huang L,Huang YB,Chen ZB,Yang LH,Jiang YA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Wed Nov 20 00:00:00 UTC 2019
Adaptive Immune Responses to Zika Virus Are Important for Controlling Virus Infection and Preventing Infection in Brain and Testes.
Journal of immunology (Baltimore, Md. : 1950)
Winkler CW,Myers LM,Woods TA,Messer RJ,Carmody AB,McNally KL,Scott DP,Hasenkrug KJ,Best SM,Peterson KE
56-5698 was used in Flow cytometry/Cell sorting to study the response of the adaptive immune system to ZIKV infection and determine its role in regulating viral infection and preventing disease.
Mon May 01 00:00:00 UTC 2017
The Ror1 receptor tyrosine kinase plays a critical role in regulating satellite cell proliferation during regeneration of injured muscle.
The Journal of biological chemistry
Kamizaki K,Doi R,Hayashi M,Saji T,Kanagawa M,Toda T,Fukada SI,Ho HH,Greenberg ME,Endo M,Minami Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Fri Sep 22 00:00:00 UTC 2017
Histone variant H2A.J accumulates in senescent cells and promotes inflammatory gene expression.
Nature communications
Contrepois K,Coudereau C,Benayoun BA,Schuler N,Roux PF,Bischof O,Courbeyrette R,Carvalho C,Thuret JY,Ma Z,Derbois C,Nevers MC,Volland H,Redon CE,Bonner WM,Deleuze JF,Wiel C,Bernard D,Snyder MP,Rübe CE,Olaso R,Fenaille F,Mann C
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Wed May 10 00:00:00 UTC 2017
IL-4 as a Repurposed Biological Drug for Myocardial Infarction through Augmentation of Reparative Cardiac Macrophages: Proof-of-Concept Data in Mice.
Scientific reports
Shintani Y,Ito T,Fields L,Shiraishi M,Ichihara Y,Sato N,Podaru M,Kainuma S,Tanaka H,Suzuki K
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Mon Jul 31 00:00:00 UTC 2017
Neonatal pancreatic pericytes support β-cell proliferation.
Molecular metabolism
Epshtein A,Rachi E,Sakhneny L,Mizrachi S,Baer D,Landsman L
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Sun Oct 01 00:00:00 UTC 2017
Hyaluronan and TLR4 promote surfactant-protein-C-positive alveolar progenitor cell renewal and prevent severe pulmonary fibrosis in mice.
Nature medicine
Liang J,Zhang Y,Xie T,Liu N,Chen H,Geng Y,Kurkciyan A,Mena JM,Stripp BR,Jiang D,Noble PW
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunocytochemistry
Tue Nov 01 00:00:00 UTC 2016
Image-based detection and targeting of therapy resistance in pancreatic adenocarcinoma.
Nature
Fox RG,Lytle NK,Jaquish DV,Park FD,Ito T,Bajaj J,Koechlein CS,Zimdahl B,Yano M,Kopp J,Kritzik M,Sicklick J,Sander M,Grandgenett PM,Hollingsworth MA,Shibata S,Pizzo D,Valasek M,Sasik R,Scadeng M,Okano H,Kim Y,MacLeod AR,Lowy AM,Reya T
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Thu Jun 16 00:00:00 UTC 2016
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Thu Oct 06 00:00:00 UTC 2016
Lineage-specific enhancers activate self-renewal genes in macrophages and embryonic stem cells.
Science (New York, N.Y.)
Soucie EL,Weng Z,Geirsdóttir L,Molawi K,Maurizio J,Fenouil R,Mossadegh-Keller N,Gimenez G,VanHille L,Beniazza M,Favret J,Berruyer C,Perrin P,Hacohen N,Andrau JC,Ferrier P,Dubreuil P,Sidow A,Sieweke MH
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Fri Feb 12 00:00:00 UTC 2016
RANKL/RANK control Brca1 mutation- .
Cell research
Sigl V,Owusu-Boaitey K,Joshi PA,Kavirayani A,Wirnsberger G,Novatchkova M,Kozieradzki I,Schramek D,Edokobi N,Hersl J,Sampson A,Odai-Afotey A,Lazaro C,Gonzalez-Suarez E,Pujana MA,Cimba F,Heyn H,Vidal E,Cruickshank J,Berman H,Sarao R,Ticevic M,Uribesalgo I,Tortola L,Rao S,Tan Y,Pfeiler G,Lee EY,Bago-Horvath Z,Kenner L,Popper H,Singer C,Khokha R,Jones LP,Penninger JM
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Jul 01 00:00:00 UTC 2016
Foxp3 and Toll-like receptor signaling balance Treg cell anabolic metabolism for suppression.
Nature immunology
Gerriets VA,Kishton RJ,Johnson MO,Cohen S,Siska PJ,Nichols AG,Warmoes MO,de Cubas AA,MacIver NJ,Locasale JW,Turka LA,Wells AD,Rathmell JC
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2016
Oligodendrocyte death results in immune-mediated CNS demyelination.
Nature neuroscience
Traka M,Podojil JR,McCarthy DP,Miller SD,Popko B
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2016
Schwann cell proliferation and differentiation that is induced by ferulic acid through MEK1/ERK1/2 signalling promotes peripheral nerve remyelination following crush injury in rats.
Experimental and therapeutic medicine
Zhu X,Li K,Guo X,Wang J,Xiang Y
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Thu Sep 01 00:00:00 UTC 2016
Whole Chromosome Instability induces senescence and promotes SASP.
Scientific reports
Andriani GA,Almeida VP,Faggioli F,Mauro M,Tsai WL,Santambrogio L,Maslov A,Gadina M,Campisi J,Vijg J,Montagna C
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Wed Oct 12 00:00:00 UTC 2016
Whole Chromosome Instability induces senescence and promotes SASP.
Scientific reports
Andriani GA,Almeida VP,Faggioli F,Mauro M,Tsai WL,Santambrogio L,Maslov A,Gadina M,Campisi J,Vijg J,Montagna C
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Wed Oct 12 00:00:00 UTC 2016
Suppression of ischemia in arterial occlusive disease by JNK-promoted native collateral artery development.
eLife
Ramo K,Sugamura K,Craige S,Keaney JF,Davis RJ
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Aug 09 00:00:00 UTC 2016
The cell proliferation antigen Ki-67 organises heterochromatin.
eLife
Sobecki M,Mrouj K,Camasses A,Parisis N,Nicolas E,Llères D,Gerbe F,Prieto S,Krasinska L,David A,Eguren M,Birling MC,Urbach S,Hem S,Déjardin J,Malumbres M,Jay P,Dulic V,Lafontaine DLj,Feil R,Fisher D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
The cell proliferation antigen Ki-67 organises heterochromatin.
eLife
Sobecki M,Mrouj K,Camasses A,Parisis N,Nicolas E,Llères D,Gerbe F,Prieto S,Krasinska L,David A,Eguren M,Birling MC,Urbach S,Hem S,Déjardin J,Malumbres M,Jay P,Dulic V,Lafontaine DLj,Feil R,Fisher D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
The cell proliferation antigen Ki-67 organises heterochromatin.
eLife
Sobecki M,Mrouj K,Camasses A,Parisis N,Nicolas E,Llères D,Gerbe F,Prieto S,Krasinska L,David A,Eguren M,Birling MC,Urbach S,Hem S,Déjardin J,Malumbres M,Jay P,Dulic V,Lafontaine DLj,Feil R,Fisher D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
The cell proliferation antigen Ki-67 organises heterochromatin.
eLife
Sobecki M,Mrouj K,Camasses A,Parisis N,Nicolas E,Llères D,Gerbe F,Prieto S,Krasinska L,David A,Eguren M,Birling MC,Urbach S,Hem S,Déjardin J,Malumbres M,Jay P,Dulic V,Lafontaine DLj,Feil R,Fisher D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
The cell proliferation antigen Ki-67 organises heterochromatin.
eLife
Sobecki M,Mrouj K,Camasses A,Parisis N,Nicolas E,Llères D,Gerbe F,Prieto S,Krasinska L,David A,Eguren M,Birling MC,Urbach S,Hem S,Déjardin J,Malumbres M,Jay P,Dulic V,Lafontaine DLj,Feil R,Fisher D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
The cell proliferation antigen Ki-67 organises heterochromatin.
eLife
Sobecki M,Mrouj K,Camasses A,Parisis N,Nicolas E,Llères D,Gerbe F,Prieto S,Krasinska L,David A,Eguren M,Birling MC,Urbach S,Hem S,Déjardin J,Malumbres M,Jay P,Dulic V,Lafontaine DLj,Feil R,Fisher D
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
Growth and metastasis of lung adenocarcinoma is potentiated by BMP4-mediated immunosuppression.
Oncoimmunology
Chen L,Yi X,Goswami S,Ahn YH,Roybal JD,Yang Y,Diao L,Peng D,Peng D,Fradette JJ,Wang J,Byers LA,Kurie JM,Ullrich SE,Qin FX,Gibbons DL
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
Heterogeneous fibroblasts underlie age-dependent tertiary lymphoid tissues in the kidney.
JCI insight
Sato Y,Mii A,Hamazaki Y,Fujita H,Nakata H,Masuda K,Nishiyama S,Shibuya S,Haga H,Ogawa O,Shimizu A,Narumiya S,Kaisho T,Arita M,Yanagisawa M,Miyasaka M,Sharma K,Minato N,Kawamoto H,Yanagita M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Thu Jul 21 00:00:00 UTC 2016
Detection of Cell Proliferation Markers by Immunofluorescence Staining and Microscopy Imaging in Paraffin-Embedded Tissue Sections.
Current protocols in molecular biology
Eminaga S,Teekakirikul P,Seidman CE,Seidman JG
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Fri Jul 01 00:00:00 UTC 2016
Immune response modulation by Galectin-1 in a transgenic model of neuroblastoma.
Oncoimmunology
Büchel G,Schulte JH,Harrison L,Batzke K,Schüller U,Hansen W,Schramm A
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Sun May 01 00:00:00 UTC 2016
The stress kinase GCN2 does not mediate suppression of antitumor T cell responses by tryptophan catabolism in experimental melanomas.
Oncoimmunology
Sonner JK,Deumelandt K,Ott M,Thomé CM,Rauschenbach KJ,Schulz S,Munteanu B,Mohapatra S,Adam I,Hofer AC,Feuerer M,Opitz CA,Hopf C,Wick W,Platten M
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
Epithelial-to-mesenchymal transition induces cell cycle arrest and parenchymal damage in renal fibrosis.
Nature medicine
Lovisa S,LeBleu VS,Tampe B,Sugimoto H,Vadnagara K,Carstens JL,Wu CC,Hagos Y,Burckhardt BC,Pentcheva-Hoang T,Nischal H,Allison JP,Zeisberg M,Kalluri R
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Tue Sep 01 00:00:00 UTC 2015
Low levels of endogenous or X-ray-induced DNA double-strand breaks activate apoptosis in adult neural stem cells.
Journal of cell science
Barazzuol L,Rickett N,Ju L,Jeggo PA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Thu Oct 01 00:00:00 UTC 2015
The adhesion G protein-coupled receptor GPR56 is a cell-autonomous regulator of oligodendrocyte development.
Nature communications
Giera S,Deng Y,Luo R,Ackerman SD,Mogha A,Monk KR,Ying Y,Jeong SJ,Makinodan M,Bialas AR,Chang BS,Stevens B,Corfas G,Piao X
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Wed Jan 21 00:00:00 UTC 2015
Mammary Stem Cells and Tumor-Initiating Cells Are More Resistant to Apoptosis and Exhibit Increased DNA Repair Activity in Response to DNA Damage.
Stem cell reports
Chang CH,Zhang M,Rajapakshe K,Coarfa C,Edwards D,Huang S,Rosen JM
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Sep 08 00:00:00 UTC 2015
NF-κB-induced microRNA-31 promotes epidermal hyperplasia by repressing protein phosphatase 6 in psoriasis.
Nature communications
Yan S,Xu Z,Lou F,Zhang L,Ke F,Bai J,Liu Z,Liu J,Wang H,Zhu H,Sun Y,Cai W,Gao Y,Su B,Li Q,Yang X,Yu J,Lai Y,Yu XZ,Zheng Y,Shen N,Chin YE,Wang H
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunohistochemistry
Fri Jul 03 00:00:00 UTC 2015
Alveolar progenitor and stem cells in lung development, renewal and cancer.
Nature
Desai TJ,Brownfield DG,Krasnow MA
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Thu Mar 13 00:00:00 UTC 2014
Metastasis is regulated via microRNA-200/ZEB1 axis control of tumour cell PD-L1 expression and intratumoral immunosuppression.
Nature communications
Chen L,Gibbons DL,Goswami S,Cortez MA,Ahn YH,Byers LA,Zhang X,Yi X,Dwyer D,Lin W,Diao L,Wang J,Roybal J,Patel M,Ungewiss C,Peng D,Antonia S,Mediavilla-Varela M,Robertson G,Suraokar M,Welsh JW,Erez B,Wistuba II,Chen L,Peng D,Wang S,Ullrich SE,Heymach JV,Kurie JM,Qin FX
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Oct 28 00:00:00 UTC 2014
Dedifferentiation of committed epithelial cells into stem cells in vivo.
Nature
Tata PR,Mou H,Pardo-Saganta A,Zhao R,Prabhu M,Law BM,Vinarsky V,Cho JL,Breton S,Sahay A,Medoff BD,Rajagopal J
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Thu Nov 14 00:00:00 UTC 2013
IL-1R signaling in dendritic cells replaces pattern-recognition receptors in promoting CD8⁺ T cell responses to influenza A virus.
Nature immunology
Pang IK,Ichinohe T,Iwasaki A
Fri Mar 01 00:00:00 UTC 2013
Primate B-1 cells generate antigen-specific B cell responses to T cell-independent type 2 antigens.
Journal of immunology (Baltimore, Md. : 1950)
Yammani RD,Haas KM
Mon Apr 01 00:00:00 UTC 2013
Small intestine inflammation in Roquin-mutant and Roquin-deficient mice.
PloS one
Schaefer JS,Montufar-Solis D,Nakra N,Vigneswaran N,Klein JR
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Flow Cytometry
Tue Sep 03 00:00:00 UTC 2013
Myeloid cells expressing VEGF and arginase-1 following uptake of damaged retinal pigment epithelium suggests potential mechanism that drives the onset of choroidal angiogenesis in mice.
PloS one
Liu J,Copland DA,Horie S,Wu WK,Chen M,Xu Y,Paul Morgan B,Mack M,Xu H,Nicholson LB,Dick AD
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Thu Jan 15 00:00:00 UTC 2015
Myeloid cells expressing VEGF and arginase-1 following uptake of damaged retinal pigment epithelium suggests potential mechanism that drives the onset of choroidal angiogenesis in mice.
PloS one
Liu J,Copland DA,Horie S,Wu WK,Chen M,Xu Y,Paul Morgan B,Mack M,Xu H,Nicholson LB,Dick AD
Published figure using Ki-67 monoclonal antibody (Product # 56-5698-82) in Immunofluorescence
Thu Jan 15 00:00:00 UTC 2015