Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Contact Us
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • MRPS18B Antibodies

            Proteintech

            MRPS18B Polyclonal Antibody

            53 References
            View all (8) MRPS18B antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # 16139-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Product not available in your region
            150 µL
             
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite MRPS18B Polyclonal Antibody

            • Antibody Testing Data (18)
            Application
            MRPS18B Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 18

            MRPS18B Antibody (16139-1-AP) in ICC/IF

            Immunofluorescent analysis of HepG2 cells using 16139-1-AP (MRPS18B antibody) at dilution of 1:50 andand Rhodamine-labeled goat anti-rabbit IGG (red).
            View Product

            MRPS18B Antibody in Immunocytochemistry (ICC/IF)
            MRPS18B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MRPS18B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MRPS18B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MRPS18B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MRPS18B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MRPS18B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Western Blot (WB)
            MRPS18B Antibody in Immunoprecipitation (IP)

            Product Details

            16139-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            View 45 publications 45 publications

            Immunohistochemistry (IHC)

            -
            View 2 publications 2 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:20-1:200
            View 3 publications 3 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            View 1 publication 1 publication

            Not applicable (N/A)

            -
            View 2 publications 2 publications
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            MRPS18B Fusion Protein Ag9198 (1-258 aa encoded by BC005373)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.38 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            This antibody recognizes specifically the 29 kDa MRPS18B protein.

            Immunogen sequence: MAASVLNTV LRRLPMLSLF RGSHRVQVPL QTLCTKAPSE EDSLSSVPIS PYKDEPWKYL ESEEYQERYG SRPVWADYRR NHKGGVPPQR TRKTCIRRNK VVGNPCPICR DHKLHVDFRN VKLLEQFVCA HTGIIFYAPY TGVCVKQHKR LTQAIQKARD HGLLIYHIPQ VEPRDLDFST SHGAVSATPP APTLVSSDPW YPWYNWKQPP ERELSRLRRL YQGHLQEESG PPPESMPKMP PRTPAEASST GQTGPQSAL (1-258 aa encoded by BC005373 )

            Target Information

            Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: 28S ribosomal protein S18-2, mitochondrial; 28S ribosomal protein S18b, mitochondrial; mitochondrial ribosomal protein S18-2; MRP-S18-2; MRP-S18-b; mrps18-b; S18mt-b; Small ribosomal subunit protein bS18b; Small ribosomal subunit protein mS40

            View more View less

            Gene Aliases: 2400002C15Rik; C6orf14; HSPC183; HumanS18a; MRP-S18-2; MRPS18-2; MRPS18B; PTD017; S18amt

            View more View less

            UniProt ID: (Human) Q9Y676, (Mouse) Q99N84

            View more View less

            Entrez Gene ID: (Human) 28973, (Rat) 294230, (Mouse) 66973

            View more View less

            Function(s)

            structural constituent of ribosome ribosomal protein

            Process(es)

            translation mitochondrial translation mitochondrial translational elongation mitochondrial translational termination
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            • Social Media
            • Contact Us
            • Report a Site Issue
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            argentina flag icon
            Argentina

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-pdf9t:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0