Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LTNLEGIYNS ETEKLRSDLE RLQLSEEEAK VATGRVLSLQ EEIAKLRKDL EQTRSEKKCI EEHADRYKQE TEQLVSNLKE ENTLLKQEKE ALNHRIVQQA KEMTETMEKK LVEETKQLEL DLNDERLRYQ NLLNEFSRLE ERYDDLKEEM TLMVHVPKPG HKRTDSTHSS NESEYIFSSE IAEMEDIPSR TEEPSEKKVP L
This gene is one of three myosin V heavy-chain genes, belonging to the myosin gene superfamily. Myosin V is a class of actin-based motor proteins involved in cytoplasmic vesicle transport and anchorage, spindle-pole alignment and mRNA translocation. The protein encoded by this gene is abundant in melanocytes and nerve cells. Mutations in this gene cause Griscelli syndrome type-1 (GS1), Griscelli syndrome type-3 (GS3) and neuroectodermal melanolysosomal disease, or Elejalde disease. Multiple alternatively spliced transcript variants encoding different isoforms have been reported, but the full-length nature of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dilute lethal-20J protein; Dilute myosin heavy chain, non-muscle; dilute-opisthotonus; flailer; myosin 5a; Myosin heavy chain 12; myosin V; myosin VA (heavy chain 12, myoxin); myosin, heavy polypeptide kinase; Myosin-12; myosin-Va; Myoxin; OTTMUSP00000017756; OTTMUSP00000019176; OTTMUSP00000046608; Unconventional myosin-Va
Gene Aliases: 9630007J19Rik; AI413174; AI661011; D; d-120J; Dbv; Dilute; Dop; flail; flr; GS1; MVa; MYH12; MYO5; MYO5A; MyoVA; MYR12; Sev-1
UniProt ID: (Human) Q9Y4I1, (Mouse) Q99104, (Rat) Q9QYF3
Entrez Gene ID: (Human) 4644, (Mouse) 17918, (Rat) 25017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support