Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Reconstitute in 100 µL of MQ water. Centrifuge to remove any insoluble material.
Specificity of this antibody: NPY6R.
Neuropeptide Y Receptor Y6 (NPY6R) is a receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibits adenylate cyclase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuropeptide Y receptor type 6; NPY6-R; Pancreatic polypeptide receptor 2; PP2
Gene Aliases: Npy5r; Npy6r; Ppyr2; Y2b
UniProt ID: (Mouse) Q61212
Entrez Gene ID: (Mouse) 18169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support