Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MKTLQSTLL LLLLVPLIKP APPTQQDSRI IYDYGTDNFE ESIFSQDYED KYLDGKNIKE KETVIIPNEK SLQLQKDEAI TPLPPKKEND EMPTCLLCVC LSGSVYCEEV DIDAVPPLPK ESAYLYARFN KIKKLTAKDF ADIPNLRRLD FTGNLIEDIE DGTFSKLSLL EELSLAENQL LKLPVLPPKL TLFNAKYNKI KSRGIKANAF KKLNNLTFLY LDHNALESVP LNLPESLRVI HLQFNNIASI TDDTFCKAND TSYIRDRIEE IRLEGNPIVL GKHPNSFICL KRLPIGSYF (1-298 aa encoded by BC037273 )
Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta. Mimecan is a small proteoglycan which contains tandem leucine-rich repeats. Mimecan is encoded by the OGN gene, a member of the small leucine-rich proteoglycan (SLRP) family of proteins. Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta (TGF-beta-1 or TGF-beta-2) and may regulate osteoblast differentiation. High expression of mimecan protein may be associated with elevated heart left ventricular mass. Alternative splicing results in multiple transcript variants of mimecan. Alternatively spliced transcript variants that encode mimecan have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: corneal keratan sulfate proteoglycan; Mimecan; mimecan proteoglycan; OIF; Osteoglycin; osteoglycin/mimecan; Osteoinductive factor
Gene Aliases: OG; OGN; OIF; SLRR3A
UniProt ID: (Human) P20774
Entrez Gene ID: (Pig) 106509723, (Human) 4969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support