Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: RHTKKTHSQ ELMKESLQTG DLLSTFHTIS PSFQLKAAAL PPFPLGASAQ NGLASSLPAE VHSLTLSPPE QAAQPMQPLP ESLASLHPSV SPGSPPPPLP NHKYNTTSTS YSPLASLPLK ADTKGFCNIS LFEDLPLQEP QSPQKLNPGF DLAKGNAGKV NLPKELPADA VNLTIPASLD LSPLLGFWQL PPPATQNTFG NSTLALGPGE SLPHRLSCLG QQQQEPPLAM GTVSLGQLPL PPIPHVFSAG TGSAILPHFH HAFR (201-463 aa encoded by B C074814)
This gene encodes a C2H2 zinc finger protein with transactivation and DNA-binding activities. It has been shown to have anti-proliferative properties, and thus thought to function as a tumor suppressor. In addition, overexpression of this gene during fetal development is believed to underlie the rare disorder, transient neonatal diabetes mellitus (TNDM). This gene is imprinted, with preferential expression of the paternal allele in many tissues, however, biallelic expression has been noted in peripheral blood leucocytes. A recent study reports that tissue-specific imprinting results from variable utilization of monoallelic and biallelic promoters. Many transcript variants differing in the 5' UTR and encoding two different isoforms, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lost on transformation 1; LOT-1; PLAG-like 1; pleiomorphic adenoma gene-like 1; Pleiomorphic adenoma-like protein 1; Tumor suppressor ZAC; Zinc finger protein PLAGL1; zinc finger protein regulator of apoptosis and cell cycle arrest
Gene Aliases: LOT1; PLAGL1; ZAC; ZAC1
UniProt ID: (Human) Q9UM63
Entrez Gene ID: (Human) 5325, (Mouse) 22634
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support