Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Sequence of this protein is as follows: MDKQNSQMNA SHPETNLPVG YPPQYPPTAF QGPPGYSGYP GPQVSYPPPP AGHSGPGPAG FPVPNQPVYN QPVYNQPVGA AGVPWMPAPQ PPLNCPPGLE YLSQIDQILI HQQIELLEVL TGFETNNKYE IKNSFGQRVY FAAEDTDCCT RNCCGPSRPF TLRIIDNMGQ EVITLERPLR CSSCCCPCCL QEIEIQAPPG VPIGYVIQTW HPCLPKFTIQ NEKREDVLKI SGPCVVCSCC GDVDFEIKSL DEQCVVGKIS KHWTGILREA FTDADNFGIQ FPLDLDVKMK AVMIGACFLI DFMFFESTGS QEQKSGVW
The calcium-dependent plasma membrane protein phospholipid scramblase 1 (PLSCR1) contributes to the transbilayer movement of phosphatidylserine and other membrane phospholipids upon influx of calcium into the cytosol. This movement results in plasma membrane phospholipid remodelling and surface exposure of phosphatidylserine in injured or apoptotic cells, which leads to cell death. Interferons and other cytokines induce expression of PLSCR1, implying that PLSCR1 also functions in cytokine signaling pathways. EGF stilmulation results in tyrosine phosphorylation of PLSCR1 on tyrosines 69 and 74, which allows it to interact with Shc, and thereby connecting Src kinase activation to stimulation of the EGF receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ca(2+)-dependent phospholipid scramblase 1; Erythrocyte phospholipid scramblase; Mg(2+)-dependent nuclease; MmTRA1b; NOR1; Phospholipid scramblase 1; PL scramblase 1; Transplantability-associated protein 1
Gene Aliases: MmTRA1a; MMTRA1B; Nor1; PLSCR1; Tra1; Tra1a; Tra1b; Tras1; Tras2
UniProt ID: (Human) O15162, (Mouse) Q9JJ00
Entrez Gene ID: (Human) 5359, (Mouse) 22038
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support