Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHALLVNHYM KGGFYPRGGS SQIAFHTIPV IQRAGGAVLT KATVQSVLLD SAGKACGVSV KKGHELVNIY CPIVVSNAGL FNTYEHLLPG NARCLPGVKQ QLGTVRPGLG MTSVFICLRG TKEDLHLPST NYYVYYDTDM DQAMERYVSM PREEAAEHIP LLFFAFPSAK DPTWEDRFPG GECDCRIPTH QPVLSGCSPR CLLRG
Retinol saturase carrying out the saturation of the 13-14 double bond of all-trans-retinol to produce all-trans-13,14-dihydroretinol. Has activity toward all-trans-retinol as substrate. Does not use all-trans-retinoic acid nor 9-cis, 11-cis or 13-cis-retinol isomers as substrates. May play a role in the metabolism of vitamin A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: All-trans-13,14-dihydroretinol saturase; All-trans-retinol 13,14-reductase; PPAR-alpha-regulated and starvation-induced gene protein; retinol saturase (all-trans-retinol 13,14-reductase); RetSat
Gene Aliases: PPSIG; RETSAT; UNQ439/PRO872
UniProt ID: (Human) Q6NUM9
Entrez Gene ID: (Human) 54884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support